Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128462714-128463205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB8
Entrez Gene ID
165721
Gene Name
DnaJ heat shock protein family (Hsp40) member B8
Gene Aliases
CT156, DJ6
UniGene
Hs.518241
Gene Chromosome Location
Chr.3: 128462432-128467248 on Build GRCh38
Transcripts
NM_153330.4 XM_006713519.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1670546 c.376C>T p.R126W Chr.3: 128462870-128462870 on Build GRCh38
3927352 c.476G>A p.G159D Chr.3: 128462770-128462770 on Build GRCh38
6647153 c.47delC p.P16fs*115 Chr.3: 128463199-128463199 on Build GRCh38
1616857 c.377G>A p.R126Q Chr.3: 128462869-128462869 on Build GRCh38
4940471 c.190C>T p.L64L Chr.3: 128463056-128463056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974384342 Chr.3: 128463159-128463159 on Build GRCh38
rs763571430 Chr.3: 128462842-128462842 on Build GRCh38
rs992661568 Chr.3: 128463101-128463101 on Build GRCh38
rs528682453 Chr.3: 128463067-128463067 on Build GRCh38
rs534460713 Chr.3: 128463002-128463002 on Build GRCh38
rs781035565 Chr.3: 128462971-128462971 on Build GRCh38
rs868039013 Chr.3: 128463074-128463077 on Build GRCh38
rs755099183 Chr.3: 128462940-128462940 on Build GRCh38
rs779641429 Chr.3: 128463136-128463136 on Build GRCh38
rs201743640 Chr.3: 128463103-128463103 on Build GRCh38
rs118145329 Chr.3: 128462739-128462739 on Build GRCh38
rs756117119 Chr.3: 128462865-128462865 on Build GRCh38
rs753974991 Chr.3: 128462868-128462868 on Build GRCh38
rs143657415 Chr.3: 128462849-128462849 on Build GRCh38
rs757171782 Chr.3: 128463011-128463011 on Build GRCh38
rs764723777 Chr.3: 128462843-128462843 on Build GRCh38
rs756204923 Chr.3: 128462793-128462793 on Build GRCh38
rs766637147 Chr.3: 128463053-128463053 on Build GRCh38
rs140270817 Chr.3: 128463120-128463120 on Build GRCh38
rs755533418 Chr.3: 128463019-128463019 on Build GRCh38
rs34694670 Chr.3: 128462991-128462991 on Build GRCh38
rs753882660 Chr.3: 128462938-128462938 on Build GRCh38
rs765817483 Chr.3: 128462773-128462773 on Build GRCh38
rs751552556 Chr.3: 128463106-128463106 on Build GRCh38
rs371803176 Chr.3: 128462978-128462978 on Build GRCh38
rs778186479 Chr.3: 128463161-128463161 on Build GRCh38
rs988268838 Chr.3: 128463057-128463057 on Build GRCh38
rs149526532 Chr.3: 128463026-128463026 on Build GRCh38
rs143882241 Chr.3: 128463062-128463062 on Build GRCh38
rs202073990 Chr.3: 128462869-128462869 on Build GRCh38
rs925364258 Chr.3: 128462836-128462836 on Build GRCh38
rs142768827 Chr.3: 128463121-128463121 on Build GRCh38
rs758063580 Chr.3: 128462957-128462957 on Build GRCh38
rs746861310 Chr.3: 128462965-128462965 on Build GRCh38
rs200983717 Chr.3: 128463003-128463003 on Build GRCh38
rs368623876 Chr.3: 128462907-128462907 on Build GRCh38
rs748790484 Chr.3: 128463025-128463025 on Build GRCh38
rs763172592 Chr.3: 128462796-128462796 on Build GRCh38
rs750443437 Chr.3: 128462790-128462790 on Build GRCh38
rs140613138 Chr.3: 128462883-128462883 on Build GRCh38
rs367931490 Chr.3: 128462966-128462966 on Build GRCh38
rs547558895 Chr.3: 128462890-128462890 on Build GRCh38
rs199590562 Chr.3: 128463047-128463047 on Build GRCh38
rs146534960 Chr.3: 128462901-128462901 on Build GRCh38
rs147724414 Chr.3: 128462755-128462755 on Build GRCh38
rs774578927 Chr.3: 128462847-128462847 on Build GRCh38
rs868550382 Chr.3: 128462889-128462889 on Build GRCh38
rs775605264 Chr.3: 128463076-128463076 on Build GRCh38
rs768380102 Chr.3: 128462806-128462806 on Build GRCh38
rs375757137 Chr.3: 128462985-128462985 on Build GRCh38
rs569985047 Chr.3: 128462945-128462945 on Build GRCh38
rs375861387 Chr.3: 128462774-128462774 on Build GRCh38
rs770510421 Chr.3: 128462888-128462888 on Build GRCh38
rs748041270 Chr.3: 128462813-128462813 on Build GRCh38
rs747545072 Chr.3: 128463164-128463164 on Build GRCh38
rs61758422 Chr.3: 128462944-128462944 on Build GRCh38
rs371954073 Chr.3: 128462853-128462853 on Build GRCh38
rs771129968 Chr.3: 128462994-128462994 on Build GRCh38
rs759465873 Chr.3: 128462997-128462997 on Build GRCh38
rs140895506 Chr.3: 128462775-128462775 on Build GRCh38
rs768468155 Chr.3: 128462746-128462746 on Build GRCh38
rs956793235 Chr.3: 128462840-128462840 on Build GRCh38
rs771814098 Chr.3: 128462761-128462761 on Build GRCh38
rs764560694 Chr.3: 128462776-128462776 on Build GRCh38
rs770048373 Chr.3: 128463183-128463183 on Build GRCh38
rs140964428 Chr.3: 128463175-128463175 on Build GRCh38
rs1025782669 Chr.3: 128462752-128462752 on Build GRCh38
rs780012430 Chr.3: 128462742-128462742 on Build GRCh38
rs777252621 Chr.3: 128462880-128462880 on Build GRCh38
rs141309633 Chr.3: 128462929-128462929 on Build GRCh38
rs758085602 Chr.3: 128463111-128463113 on Build GRCh38
rs746470754 Chr.3: 128462820-128462820 on Build GRCh38
rs766457778 Chr.3: 128462867-128462867 on Build GRCh38
rs748553089 Chr.3: 128462734-128462734 on Build GRCh38
rs747873989 Chr.3: 128462878-128462878 on Build GRCh38
rs2659690 Chr.3: 128462934-128462934 on Build GRCh38
rs757410955 Chr.3: 128462786-128462786 on Build GRCh38
rs138090733 Chr.3: 128462984-128462984 on Build GRCh38
rs774940330 Chr.3: 128462903-128462903 on Build GRCh38
rs776851510 Chr.3: 128462996-128462996 on Build GRCh38
rs201620402 Chr.3: 128462933-128462933 on Build GRCh38
rs896205201 Chr.3: 128463178-128463178 on Build GRCh38
rs1012668058 Chr.3: 128462919-128462919 on Build GRCh38
rs1026507026 Chr.3: 128462751-128462751 on Build GRCh38
rs778557299 Chr.3: 128462807-128462807 on Build GRCh38
rs780929244 Chr.3: 128463014-128463014 on Build GRCh38
rs534274395 Chr.3: 128462870-128462870 on Build GRCh38
rs1056817540 Chr.3: 128462874-128462874 on Build GRCh38
rs200457178 Chr.3: 128463015-128463015 on Build GRCh38
rs761664766 Chr.3: 128463112-128463112 on Build GRCh38
rs766008763 Chr.3: 128463150-128463150 on Build GRCh38
rs779596027 Chr.3: 128463020-128463020 on Build GRCh38
rs757298895 Chr.3: 128462735-128462735 on Build GRCh38
rs760272118 Chr.3: 128462772-128462772 on Build GRCh38
rs145860432 Chr.3: 128462817-128462817 on Build GRCh38
rs753376929 Chr.3: 128463150-128463150 on Build GRCh38
rs113196104 Chr.3: 128462877-128462877 on Build GRCh38
rs763005953 Chr.3: 128463091-128463091 on Build GRCh38
rs751750410 Chr.3: 128462782-128462782 on Build GRCh38
rs148940950 Chr.3: 128462841-128462841 on Build GRCh38
rs35948511 Chr.3: 128462789-128462789 on Build GRCh38
rs112059249 Chr.3: 128463004-128463004 on Build GRCh38
rs915237255 Chr.3: 128462778-128462778 on Build GRCh38
rs370064517 Chr.3: 128463176-128463176 on Build GRCh38
rs369051408 Chr.3: 128463046-128463046 on Build GRCh38
rs769956876 Chr.3: 128463069-128463069 on Build GRCh38
rs772967326 Chr.3: 128462818-128462818 on Build GRCh38
rs751391450 Chr.3: 128463010-128463010 on Build GRCh38
rs147375075 Chr.3: 128463160-128463160 on Build GRCh38
rs200955342 Chr.3: 128462738-128462738 on Build GRCh38
rs201512803 Chr.3: 128463127-128463127 on Build GRCh38
rs199602765 Chr.3: 128463126-128463126 on Build GRCh38
rs767445904 Chr.3: 128463113-128463113 on Build GRCh38
rs967998451 Chr.3: 128463042-128463042 on Build GRCh38
rs749226045 Chr.3: 128462805-128462805 on Build GRCh38
rs766654127 Chr.3: 128462935-128462935 on Build GRCh38
rs374581480 Chr.3: 128462749-128462749 on Build GRCh38
rs960848602 Chr.3: 128462757-128462757 on Build GRCh38
rs150389103 Chr.3: 128462979-128462979 on Build GRCh38
rs374415635 Chr.3: 128462879-128462879 on Build GRCh38
rs201659373 Chr.3: 128463061-128463061 on Build GRCh38
rs745442143 Chr.3: 128462894-128462894 on Build GRCh38
rs546175418 Chr.3: 128463088-128463088 on Build GRCh38
rs141737255 Chr.3: 128462960-128462960 on Build GRCh38
rs772593121 Chr.3: 128462767-128462767 on Build GRCh38
rs757567730 Chr.3: 128463170-128463170 on Build GRCh38
rs762040488 Chr.3: 128462916-128462916 on Build GRCh38
rs746255807 Chr.3: 128463181-128463181 on Build GRCh38

View Full Product Details