Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128463628-128464122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB8
Entrez Gene ID
165721
Gene Name
DnaJ heat shock protein family (Hsp40) member B8
Gene Aliases
CT156, DJ6
UniGene
Hs.518241
Gene Chromosome Location
Chr.3: 128462432-128467248 on Build GRCh38
Transcripts
NM_153330.4 XM_006713519.3 NR_037890.1

Gene Symbol
DNAJB8-AS1
Entrez Gene ID
285224
Gene Name
DNAJB8 antisense RNA 1
Gene Aliases
-
UniGene
Hs.683815
Gene Chromosome Location
Chr.3: 128463594-128472317 on Build GRCh38
Transcripts
NM_153330.4 XM_006713519.3 NR_037890.1

SNP Detail

SNP ID Chromosome Location
rs1003997553 Chr.3: 128463905-128463905 on Build GRCh38
rs567190542 Chr.3: 128463859-128463859 on Build GRCh38
rs953139389 Chr.3: 128463966-128463966 on Build GRCh38
rs555846708 Chr.3: 128463928-128463928 on Build GRCh38
rs546012355 Chr.3: 128464028-128464028 on Build GRCh38
rs558130582 Chr.3: 128463797-128463797 on Build GRCh38
rs1024051503 Chr.3: 128463677-128463677 on Build GRCh38
rs975813551 Chr.3: 128463765-128463765 on Build GRCh38
rs575318548 Chr.3: 128463975-128463975 on Build GRCh38
rs570869268 Chr.3: 128463890-128463890 on Build GRCh38
rs1028683838 Chr.3: 128463965-128463965 on Build GRCh38
rs1001476945 Chr.3: 128463887-128463887 on Build GRCh38
rs564203657 Chr.3: 128464032-128464032 on Build GRCh38
rs756841871 Chr.3: 128463680-128463680 on Build GRCh38
rs970403177 Chr.3: 128463713-128463713 on Build GRCh38
rs905173840 Chr.3: 128463872-128463872 on Build GRCh38
rs528059610 Chr.3: 128463663-128463663 on Build GRCh38
rs557118478 Chr.3: 128463806-128463806 on Build GRCh38
rs761592744 Chr.3: 128463907-128463907 on Build GRCh38
rs935466105 Chr.3: 128463790-128463790 on Build GRCh38
rs1027543739 Chr.3: 128463895-128463895 on Build GRCh38
rs59916512 Chr.3: 128463861-128463861 on Build GRCh38
rs966874498 Chr.3: 128463972-128463972 on Build GRCh38
rs147827797 Chr.3: 128463793-128463793 on Build GRCh38
rs537529224 Chr.3: 128463775-128463775 on Build GRCh38
rs989678477 Chr.3: 128463751-128463751 on Build GRCh38
rs528431665 Chr.3: 128464076-128464076 on Build GRCh38
rs1043939903 Chr.3: 128463785-128463785 on Build GRCh38
rs1033438455 Chr.3: 128463732-128463732 on Build GRCh38
rs758263397 Chr.3: 128464031-128464031 on Build GRCh38
rs569303979 Chr.3: 128463789-128463789 on Build GRCh38
rs1024848326 Chr.3: 128463691-128463691 on Build GRCh38
rs185834589 Chr.3: 128463948-128463948 on Build GRCh38
rs918064592 Chr.3: 128464010-128464010 on Build GRCh38
rs899259696 Chr.3: 128463831-128463831 on Build GRCh38
rs140974225 Chr.3: 128464077-128464077 on Build GRCh38
rs567811275 Chr.3: 128463749-128463749 on Build GRCh38
rs546216742 Chr.3: 128463681-128463681 on Build GRCh38
rs1027511225 Chr.3: 128463688-128463688 on Build GRCh38
rs930749747 Chr.3: 128463835-128463835 on Build GRCh38
rs920613971 Chr.3: 128464049-128464049 on Build GRCh38
rs528994310 Chr.3: 128463764-128463764 on Build GRCh38
rs980372500 Chr.3: 128463728-128463728 on Build GRCh38
rs1001895907 Chr.3: 128463926-128463926 on Build GRCh38
rs905204317 Chr.3: 128464039-128464039 on Build GRCh38
rs796480587 Chr.3: 128463837-128463837 on Build GRCh38
rs544679214 Chr.3: 128463950-128463950 on Build GRCh38
rs539797076 Chr.3: 128463927-128463927 on Build GRCh38
rs887733308 Chr.3: 128464069-128464069 on Build GRCh38
rs778690503 Chr.3: 128463756-128463756 on Build GRCh38
rs369467436 Chr.3: 128463798-128463798 on Build GRCh38
rs16847447 Chr.3: 128463973-128463973 on Build GRCh38
rs921590145 Chr.3: 128463767-128463767 on Build GRCh38
rs1024858156 Chr.3: 128463906-128463906 on Build GRCh38
rs779735167 Chr.3: 128463867-128463867 on Build GRCh38
rs1049011685 Chr.3: 128464066-128464066 on Build GRCh38
rs543240494 Chr.3: 128463825-128463825 on Build GRCh38
rs931269251 Chr.3: 128464047-128464047 on Build GRCh38
rs966102409 Chr.3: 128463763-128463763 on Build GRCh38
rs753848050 Chr.3: 128463762-128463762 on Build GRCh38
rs896606405 Chr.3: 128463802-128463802 on Build GRCh38

View Full Product Details