Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128479342-128479845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA2
Entrez Gene ID
2624
Gene Name
GATA binding protein 2
Gene Aliases
DCML, IMD21, MONOMAC, NFE1B
UniGene
Hs.367725
Gene Chromosome Location
Chr.3: 128479422-128493187 on Build GRCh38
Transcripts
NM_001145661.1 NM_001145662.1 NM_032638.4

SNP Detail

SNP ID Chromosome Location
rs75618111 Chr.3: 128479591-128479591 on Build GRCh38
rs142582404 Chr.3: 128479790-128479790 on Build GRCh38
rs1026996375 Chr.3: 128479483-128479483 on Build GRCh38
rs927129253 Chr.3: 128479614-128479614 on Build GRCh38
rs1041362073 Chr.3: 128479679-128479679 on Build GRCh38
rs182456056 Chr.3: 128479671-128479671 on Build GRCh38
rs917330795 Chr.3: 128479564-128479564 on Build GRCh38
rs754227748 Chr.3: 128479474-128479474 on Build GRCh38
rs937263419 Chr.3: 128479655-128479655 on Build GRCh38
rs1019397802 Chr.3: 128479744-128479744 on Build GRCh38
rs77807344 Chr.3: 128479585-128479585 on Build GRCh38
rs141209260 Chr.3: 128479776-128479776 on Build GRCh38
rs1018430940 Chr.3: 128479438-128479438 on Build GRCh38
rs757792126 Chr.3: 128479575-128479575 on Build GRCh38
rs886979728 Chr.3: 128479714-128479714 on Build GRCh38
rs951332173 Chr.3: 128479503-128479503 on Build GRCh38
rs746310055 Chr.3: 128479706-128479706 on Build GRCh38
rs771679984 Chr.3: 128479724-128479724 on Build GRCh38
rs1028607561 Chr.3: 128479365-128479365 on Build GRCh38
rs995754448 Chr.3: 128479475-128479475 on Build GRCh38
rs914250693 Chr.3: 128479669-128479669 on Build GRCh38
rs1008836392 Chr.3: 128479424-128479424 on Build GRCh38
rs905132525 Chr.3: 128479601-128479601 on Build GRCh38
rs979354797 Chr.3: 128479450-128479450 on Build GRCh38
rs982905636 Chr.3: 128479573-128479573 on Build GRCh38
rs992962946 Chr.3: 128479523-128479523 on Build GRCh38
rs534739996 Chr.3: 128479383-128479383 on Build GRCh38
rs891310174 Chr.3: 128479399-128479399 on Build GRCh38
rs934751542 Chr.3: 128479517-128479517 on Build GRCh38
rs1054264379 Chr.3: 128479662-128479662 on Build GRCh38
rs748202430 Chr.3: 128479416-128479416 on Build GRCh38
rs11552327 Chr.3: 128479672-128479672 on Build GRCh38
rs1048705252 Chr.3: 128479702-128479702 on Build GRCh38
rs1024969919 Chr.3: 128479783-128479783 on Build GRCh38
rs1027217308 Chr.3: 128479773-128479773 on Build GRCh38
rs1007968865 Chr.3: 128479750-128479750 on Build GRCh38
rs940720520 Chr.3: 128479743-128479743 on Build GRCh38
rs981717592 Chr.3: 128479613-128479613 on Build GRCh38
rs970826848 Chr.3: 128479596-128479596 on Build GRCh38
rs867394821 Chr.3: 128479657-128479657 on Build GRCh38
rs899961260 Chr.3: 128479756-128479756 on Build GRCh38
rs777938092 Chr.3: 128479470-128479470 on Build GRCh38
rs1004450512 Chr.3: 128479715-128479715 on Build GRCh38
rs992939743 Chr.3: 128479818-128479818 on Build GRCh38
rs890899099 Chr.3: 128479701-128479701 on Build GRCh38
rs886057922 Chr.3: 128479476-128479476 on Build GRCh38
rs371779462 Chr.3: 128479652-128479652 on Build GRCh38
rs778707164 Chr.3: 128479396-128479396 on Build GRCh38
rs568410851 Chr.3: 128479782-128479782 on Build GRCh38
rs34069018 Chr.3: 128479542-128479543 on Build GRCh38
rs570968383 Chr.3: 128479751-128479751 on Build GRCh38
rs535278189 Chr.3: 128479775-128479775 on Build GRCh38
rs971261577 Chr.3: 128479788-128479788 on Build GRCh38
rs866904284 Chr.3: 128479597-128479597 on Build GRCh38
rs995772875 Chr.3: 128479757-128479757 on Build GRCh38
rs191422571 Chr.3: 128479646-128479646 on Build GRCh38
rs886057921 Chr.3: 128479475-128479475 on Build GRCh38

View Full Product Details