Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129001452-129001952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCC1
Entrez Gene ID
79825
Gene Name
EF-hand and coiled-coil domain containing 1
Gene Aliases
C3orf73, CCDC48
UniGene
Hs.134807
Gene Chromosome Location
Chr.3: 129001604-129040742 on Build GRCh38
Transcripts
NM_024768.2 XM_005247781.3 XM_011513163.2 XM_011513050.2 XM_017006939.1 XM_017006940.1 XM_017006941.1 XM_017006942.1 XM_017006943.1 XM_017006945.1 XM_017006946.1 XM_017006947.1 XM_017006952.1

Gene Symbol
KIAA1257
Entrez Gene ID
57501
Gene Name
KIAA1257
Gene Aliases
-
UniGene
Hs.518247
Gene Chromosome Location
Chr.3: 128910186-129032431 on Build GRCh38
Transcripts
NM_024768.2 XM_005247781.3 XM_011513163.2 XM_011513050.2 XM_017006939.1 XM_017006940.1 XM_017006941.1 XM_017006942.1 XM_017006943.1 XM_017006945.1 XM_017006946.1 XM_017006947.1 XM_017006952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4656387 c.239G>A p.R80Q Chr.3: 129001867-129001867 on Build GRCh38
4886265 c.301G>T p.G101C Chr.3: 129001929-129001929 on Build GRCh38
4522588 c.116G>A p.G39E Chr.3: 129001744-129001744 on Build GRCh38
3765053 c.16A>G p.T6A Chr.3: 129001644-129001644 on Build GRCh38
1199984 c.31G>A p.G11S Chr.3: 129001659-129001659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959906792 Chr.3: 129001684-129001684 on Build GRCh38
rs547077945 Chr.3: 129001514-129001514 on Build GRCh38
rs999675082 Chr.3: 129001525-129001525 on Build GRCh38
rs746043608 Chr.3: 129001841-129001842 on Build GRCh38
rs1052566857 Chr.3: 129001795-129001795 on Build GRCh38
rs934857536 Chr.3: 129001561-129001561 on Build GRCh38
rs368676820 Chr.3: 129001737-129001737 on Build GRCh38
rs570602884 Chr.3: 129001654-129001654 on Build GRCh38
rs961800230 Chr.3: 129001623-129001623 on Build GRCh38
rs964553875 Chr.3: 129001728-129001728 on Build GRCh38
rs991820304 Chr.3: 129001699-129001699 on Build GRCh38
rs535167034 Chr.3: 129001663-129001663 on Build GRCh38
rs934733163 Chr.3: 129001919-129001919 on Build GRCh38
rs903977916 Chr.3: 129001520-129001520 on Build GRCh38
rs902074396 Chr.3: 129001567-129001567 on Build GRCh38
rs947381450 Chr.3: 129001517-129001517 on Build GRCh38
rs889401195 Chr.3: 129001878-129001878 on Build GRCh38
rs750291278 Chr.3: 129001843-129001843 on Build GRCh38
rs1016410806 Chr.3: 129001619-129001619 on Build GRCh38
rs1051120059 Chr.3: 129001867-129001867 on Build GRCh38
rs1052632291 Chr.3: 129001528-129001528 on Build GRCh38
rs754537184 Chr.3: 129001757-129001757 on Build GRCh38
rs569885346 Chr.3: 129001590-129001590 on Build GRCh38
rs751472647 Chr.3: 129001787-129001787 on Build GRCh38
rs915863998 Chr.3: 129001708-129001708 on Build GRCh38
rs574809471 Chr.3: 129001726-129001726 on Build GRCh38
rs530314755 Chr.3: 129001524-129001524 on Build GRCh38
rs751195044 Chr.3: 129001671-129001671 on Build GRCh38
rs755811461 Chr.3: 129001877-129001877 on Build GRCh38
rs553070355 Chr.3: 129001675-129001675 on Build GRCh38
rs767185316 Chr.3: 129001823-129001823 on Build GRCh38
rs775534884 Chr.3: 129001754-129001754 on Build GRCh38
rs771224733 Chr.3: 129001713-129001713 on Build GRCh38
rs948126598 Chr.3: 129001503-129001503 on Build GRCh38
rs938612511 Chr.3: 129001476-129001476 on Build GRCh38
rs761627107 Chr.3: 129001815-129001815 on Build GRCh38
rs971841621 Chr.3: 129001635-129001635 on Build GRCh38
rs1028623359 Chr.3: 129001604-129001604 on Build GRCh38
rs902020584 Chr.3: 129001802-129001802 on Build GRCh38
rs776877258 Chr.3: 129001716-129001716 on Build GRCh38
rs924726770 Chr.3: 129001883-129001883 on Build GRCh38
rs72977188 Chr.3: 129001611-129001611 on Build GRCh38
rs370208626 Chr.3: 129001548-129001548 on Build GRCh38
rs779914968 Chr.3: 129001925-129001925 on Build GRCh38
rs559331172 Chr.3: 129001522-129001522 on Build GRCh38
rs921283972 Chr.3: 129001501-129001501 on Build GRCh38
rs773592981 Chr.3: 129001649-129001649 on Build GRCh38
rs147900895 Chr.3: 129001505-129001505 on Build GRCh38
rs866425772 Chr.3: 129001923-129001923 on Build GRCh38
rs999506884 Chr.3: 129001582-129001593 on Build GRCh38
rs554907927 Chr.3: 129001753-129001753 on Build GRCh38
rs898853214 Chr.3: 129001620-129001620 on Build GRCh38
rs764026856 Chr.3: 129001761-129001761 on Build GRCh38
rs997770367 Chr.3: 129001595-129001595 on Build GRCh38
rs1031495612 Chr.3: 129001584-129001595 on Build GRCh38
rs1006289070 Chr.3: 129001896-129001896 on Build GRCh38
rs1871951 Chr.3: 129001644-129001644 on Build GRCh38
rs896606767 Chr.3: 129001829-129001847 on Build GRCh38
rs768267208 Chr.3: 129001811-129001811 on Build GRCh38
rs1044190686 Chr.3: 129001507-129001507 on Build GRCh38
rs953156652 Chr.3: 129001607-129001607 on Build GRCh38
rs748862254 Chr.3: 129001637-129001637 on Build GRCh38
rs916569327 Chr.3: 129001497-129001497 on Build GRCh38
rs866616254 Chr.3: 129001924-129001924 on Build GRCh38
rs144678686 Chr.3: 129001481-129001481 on Build GRCh38
rs747342896 Chr.3: 129001682-129001682 on Build GRCh38

View Full Product Details