Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129252198-129252704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPG1
Entrez Gene ID
22820
Gene Name
coatomer protein complex subunit gamma 1
Gene Aliases
COPG
UniGene
Hs.518250
Gene Chromosome Location
Chr.3: 129249575-129277773 on Build GRCh38
Transcripts
NM_016128.3 XM_011512549.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5892856 c.232C>T p.Q78* Chr.3: 129252683-129252683 on Build GRCh38
5551208 c.132T>C p.C44C Chr.3: 129252322-129252322 on Build GRCh38
1536490 c.210C>T p.F70F Chr.3: 129252661-129252661 on Build GRCh38
6238073 c.146C>A p.T49N Chr.3: 129252336-129252336 on Build GRCh38
7162908 c.162C>T p.L54L Chr.3: 129252352-129252352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759319201 Chr.3: 129252595-129252595 on Build GRCh38
rs142574517 Chr.3: 129252340-129252340 on Build GRCh38
rs781314015 Chr.3: 129252282-129252282 on Build GRCh38
rs753279792 Chr.3: 129252256-129252256 on Build GRCh38
rs540554352 Chr.3: 129252250-129252250 on Build GRCh38
rs538955620 Chr.3: 129252520-129252520 on Build GRCh38
rs187497575 Chr.3: 129252366-129252366 on Build GRCh38
rs948862880 Chr.3: 129252532-129252532 on Build GRCh38
rs1052814599 Chr.3: 129252296-129252296 on Build GRCh38
rs113059124 Chr.3: 129252375-129252375 on Build GRCh38
rs749959471 Chr.3: 129252233-129252233 on Build GRCh38
rs747664234 Chr.3: 129252243-129252243 on Build GRCh38
rs571530812 Chr.3: 129252517-129252517 on Build GRCh38
rs149206482 Chr.3: 129252649-129252649 on Build GRCh38
rs772816543 Chr.3: 129252335-129252335 on Build GRCh38
rs745991108 Chr.3: 129252285-129252285 on Build GRCh38
rs771924432 Chr.3: 129252584-129252584 on Build GRCh38
rs374018706 Chr.3: 129252368-129252368 on Build GRCh38
rs145702032 Chr.3: 129252321-129252321 on Build GRCh38
rs750855746 Chr.3: 129252367-129252368 on Build GRCh38
rs758779886 Chr.3: 129252259-129252259 on Build GRCh38
rs575927643 Chr.3: 129252644-129252644 on Build GRCh38
rs780109873 Chr.3: 129252386-129252386 on Build GRCh38
rs891622460 Chr.3: 129252562-129252562 on Build GRCh38
rs754913505 Chr.3: 129252653-129252653 on Build GRCh38
rs770794831 Chr.3: 129252453-129252453 on Build GRCh38
rs764263230 Chr.3: 129252630-129252630 on Build GRCh38
rs764553632 Chr.3: 129252631-129252631 on Build GRCh38
rs904416621 Chr.3: 129252548-129252548 on Build GRCh38
rs775353146 Chr.3: 129252303-129252303 on Build GRCh38
rs1006847141 Chr.3: 129252369-129252369 on Build GRCh38
rs1049700117 Chr.3: 129252490-129252490 on Build GRCh38
rs112659808 Chr.3: 129252416-129252416 on Build GRCh38
rs779523763 Chr.3: 129252237-129252237 on Build GRCh38
rs1053330527 Chr.3: 129252561-129252561 on Build GRCh38
rs553960844 Chr.3: 129252580-129252580 on Build GRCh38
rs746106771 Chr.3: 129252377-129252377 on Build GRCh38
rs779697701 Chr.3: 129252621-129252622 on Build GRCh38
rs1000047713 Chr.3: 129252550-129252550 on Build GRCh38
rs752142619 Chr.3: 129252635-129252635 on Build GRCh38
rs917466076 Chr.3: 129252458-129252458 on Build GRCh38
rs143244455 Chr.3: 129252676-129252676 on Build GRCh38
rs755518343 Chr.3: 129252234-129252234 on Build GRCh38
rs535955599 Chr.3: 129252623-129252623 on Build GRCh38
rs111368177 Chr.3: 129252435-129252435 on Build GRCh38
rs926847893 Chr.3: 129252380-129252380 on Build GRCh38
rs114266469 Chr.3: 129252354-129252354 on Build GRCh38
rs904201078 Chr.3: 129252273-129252273 on Build GRCh38
rs369603395 Chr.3: 129252667-129252667 on Build GRCh38
rs769889570 Chr.3: 129252287-129252287 on Build GRCh38
rs750474907 Chr.3: 129252669-129252669 on Build GRCh38
rs766000841 Chr.3: 129252339-129252339 on Build GRCh38
rs371399914 Chr.3: 129252597-129252597 on Build GRCh38
rs776342417 Chr.3: 129252628-129252628 on Build GRCh38
rs372813357 Chr.3: 129252642-129252642 on Build GRCh38
rs60958881 Chr.3: 129252477-129252477 on Build GRCh38
rs768532565 Chr.3: 129252412-129252412 on Build GRCh38
rs371120121 Chr.3: 129252315-129252315 on Build GRCh38
rs376964607 Chr.3: 129252648-129252648 on Build GRCh38
rs748131137 Chr.3: 129252658-129252658 on Build GRCh38
rs761512466 Chr.3: 129252332-129252332 on Build GRCh38
rs4927953 Chr.3: 129252270-129252270 on Build GRCh38
rs774172924 Chr.3: 129252574-129252574 on Build GRCh38
rs201775145 Chr.3: 129252367-129252367 on Build GRCh38
rs184520282 Chr.3: 129252588-129252588 on Build GRCh38
rs75128085 Chr.3: 129252403-129252403 on Build GRCh38
rs778675521 Chr.3: 129252655-129252655 on Build GRCh38
rs778006635 Chr.3: 129252265-129252265 on Build GRCh38
rs764510012 Chr.3: 129252355-129252355 on Build GRCh38
rs758750587 Chr.3: 129252405-129252405 on Build GRCh38
rs368150983 Chr.3: 129252381-129252381 on Build GRCh38
rs368399189 Chr.3: 129252337-129252337 on Build GRCh38
rs935685497 Chr.3: 129252615-129252615 on Build GRCh38
rs554697382 Chr.3: 129252643-129252643 on Build GRCh38
rs771118633 Chr.3: 129252276-129252276 on Build GRCh38
rs932293281 Chr.3: 129252457-129252458 on Build GRCh38
rs956978389 Chr.3: 129252360-129252360 on Build GRCh38
rs761901616 Chr.3: 129252646-129252646 on Build GRCh38
rs772552568 Chr.3: 129252392-129252392 on Build GRCh38
rs767124070 Chr.3: 129252334-129252334 on Build GRCh38
rs774188206 Chr.3: 129252538-129252538 on Build GRCh38
rs762888021 Chr.3: 129252314-129252314 on Build GRCh38

View Full Product Details