Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129254414-129254937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPG1
Entrez Gene ID
22820
Gene Name
coatomer protein complex subunit gamma 1
Gene Aliases
COPG
UniGene
Hs.518250
Gene Chromosome Location
Chr.3: 129249575-129277773 on Build GRCh38
Transcripts
NM_016128.3 XM_011512549.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6224153 c.376C>T p.R126* Chr.3: 129254720-129254720 on Build GRCh38
1752894 c.393C>T p.I131I Chr.3: 129254737-129254737 on Build GRCh38
7297576 c.372C>T p.A124A Chr.3: 129254716-129254716 on Build GRCh38
1038049 c.373G>A p.V125M Chr.3: 129254717-129254717 on Build GRCh38
4929220 c.393C>A p.I131I Chr.3: 129254737-129254737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941711032 Chr.3: 129254911-129254911 on Build GRCh38
rs866174619 Chr.3: 129254870-129254870 on Build GRCh38
rs764260447 Chr.3: 129254732-129254732 on Build GRCh38
rs573184194 Chr.3: 129254570-129254570 on Build GRCh38
rs138650108 Chr.3: 129254713-129254713 on Build GRCh38
rs138065578 Chr.3: 129254680-129254680 on Build GRCh38
rs755690142 Chr.3: 129254687-129254687 on Build GRCh38
rs765539221 Chr.3: 129254665-129254665 on Build GRCh38
rs542106949 Chr.3: 129254862-129254862 on Build GRCh38
rs747668981 Chr.3: 129254649-129254649 on Build GRCh38
rs942681645 Chr.3: 129254698-129254698 on Build GRCh38
rs1039806034 Chr.3: 129254894-129254894 on Build GRCh38
rs781690771 Chr.3: 129254793-129254793 on Build GRCh38
rs1008367974 Chr.3: 129254551-129254551 on Build GRCh38
rs573994393 Chr.3: 129254847-129254847 on Build GRCh38
rs756402486 Chr.3: 129254888-129254888 on Build GRCh38
rs767199731 Chr.3: 129254685-129254685 on Build GRCh38
rs4927913 Chr.3: 129254904-129254904 on Build GRCh38
rs370947387 Chr.3: 129254623-129254623 on Build GRCh38
rs139016263 Chr.3: 129254716-129254716 on Build GRCh38
rs928843432 Chr.3: 129254898-129254898 on Build GRCh38
rs1040000329 Chr.3: 129254559-129254559 on Build GRCh38
rs763276285 Chr.3: 129254721-129254721 on Build GRCh38
rs926512977 Chr.3: 129254439-129254439 on Build GRCh38
rs912542615 Chr.3: 129254881-129254881 on Build GRCh38
rs753484584 Chr.3: 129254757-129254757 on Build GRCh38
rs900111513 Chr.3: 129254579-129254579 on Build GRCh38
rs80019867 Chr.3: 129254755-129254755 on Build GRCh38
rs376130585 Chr.3: 129254705-129254705 on Build GRCh38
rs567971939 Chr.3: 129254751-129254751 on Build GRCh38
rs775727605 Chr.3: 129254720-129254720 on Build GRCh38
rs911464929 Chr.3: 129254440-129254440 on Build GRCh38
rs748961361 Chr.3: 129254706-129254706 on Build GRCh38
rs771281531 Chr.3: 129254636-129254636 on Build GRCh38
rs937359901 Chr.3: 129254478-129254478 on Build GRCh38
rs1033982986 Chr.3: 129254574-129254574 on Build GRCh38
rs181612246 Chr.3: 129254498-129254498 on Build GRCh38
rs957828963 Chr.3: 129254828-129254828 on Build GRCh38
rs144931788 Chr.3: 129254712-129254712 on Build GRCh38
rs570175744 Chr.3: 129254714-129254714 on Build GRCh38
rs1006173795 Chr.3: 129254598-129254598 on Build GRCh38
rs373414971 Chr.3: 129254616-129254616 on Build GRCh38
rs779833800 Chr.3: 129254484-129254484 on Build GRCh38
rs1024692249 Chr.3: 129254669-129254669 on Build GRCh38
rs374157933 Chr.3: 129254882-129254882 on Build GRCh38
rs894132946 Chr.3: 129254575-129254575 on Build GRCh38
rs370723106 Chr.3: 129254750-129254750 on Build GRCh38
rs754427143 Chr.3: 129254707-129254707 on Build GRCh38
rs1026683587 Chr.3: 129254601-129254601 on Build GRCh38
rs774626666 Chr.3: 129254746-129254746 on Build GRCh38
rs186352729 Chr.3: 129254809-129254809 on Build GRCh38
rs763081148 Chr.3: 129254677-129254677 on Build GRCh38
rs1048636107 Chr.3: 129254762-129254762 on Build GRCh38
rs773661001 Chr.3: 129254632-129254632 on Build GRCh38
rs775609547 Chr.3: 129254674-129254674 on Build GRCh38
rs150896652 Chr.3: 129254823-129254823 on Build GRCh38
rs780920921 Chr.3: 129254891-129254891 on Build GRCh38
rs139359785 Chr.3: 129254865-129254865 on Build GRCh38
rs4927914 Chr.3: 129254820-129254820 on Build GRCh38
rs1055573615 Chr.3: 129254905-129254905 on Build GRCh38
rs751471194 Chr.3: 129254681-129254681 on Build GRCh38
rs373449238 Chr.3: 129254633-129254633 on Build GRCh38
rs989366040 Chr.3: 129254837-129254837 on Build GRCh38
rs778344234 Chr.3: 129254486-129254486 on Build GRCh38
rs112007954 Chr.3: 129254783-129254783 on Build GRCh38
rs540073837 Chr.3: 129254794-129254794 on Build GRCh38
rs541769033 Chr.3: 129254517-129254517 on Build GRCh38
rs1042209160 Chr.3: 129254715-129254715 on Build GRCh38
rs745823493 Chr.3: 129254499-129254499 on Build GRCh38
rs530666916 Chr.3: 129254609-129254609 on Build GRCh38
rs868834886 Chr.3: 129254530-129254530 on Build GRCh38
rs897708544 Chr.3: 129254804-129254804 on Build GRCh38
rs986880038 Chr.3: 129254653-129254653 on Build GRCh38
rs777112673 Chr.3: 129254640-129254640 on Build GRCh38
rs386665750 Chr.3: 129254904-129254906 on Build GRCh38
rs887342417 Chr.3: 129254915-129254915 on Build GRCh38
rs995743048 Chr.3: 129254595-129254595 on Build GRCh38
rs1018756348 Chr.3: 129254849-129254849 on Build GRCh38
rs770080152 Chr.3: 129254717-129254717 on Build GRCh38
rs771847926 Chr.3: 129254697-129254697 on Build GRCh38
rs1016584318 Chr.3: 129254604-129254604 on Build GRCh38
rs75765157 Chr.3: 129254838-129254838 on Build GRCh38
rs750279239 Chr.3: 129254686-129254686 on Build GRCh38
rs750357594 Chr.3: 129254754-129254754 on Build GRCh38
rs951075282 Chr.3: 129254615-129254615 on Build GRCh38
rs1048689802 Chr.3: 129254908-129254908 on Build GRCh38
rs778374543 Chr.3: 129254765-129254765 on Build GRCh38
rs551837614 Chr.3: 129254626-129254626 on Build GRCh38
rs370883000 Chr.3: 129254774-129254774 on Build GRCh38
rs931433097 Chr.3: 129254906-129254906 on Build GRCh38
rs565255993 Chr.3: 129254562-129254562 on Build GRCh38
rs545563909 Chr.3: 129254883-129254883 on Build GRCh38
rs142551391 Chr.3: 129254682-129254682 on Build GRCh38
rs759657651 Chr.3: 129254643-129254643 on Build GRCh38
rs1002503014 Chr.3: 129254557-129254557 on Build GRCh38
rs778462445 Chr.3: 129254708-129254708 on Build GRCh38
rs906933041 Chr.3: 129254518-129254518 on Build GRCh38
rs146419907 Chr.3: 129254472-129254472 on Build GRCh38
rs114317779 Chr.3: 129254761-129254761 on Build GRCh38
rs1041467736 Chr.3: 129254501-129254501 on Build GRCh38

View Full Product Details