Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130376523-130377030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A5
Entrez Gene ID
256076
Gene Name
collagen type VI alpha 5
Gene Aliases
COL29A1, VWA4
UniGene
Hs.205403
Gene Chromosome Location
Chr.3: 130345516-130484846 on Build GRCh38
Transcripts
NM_001278298.1 NM_153264.6 NR_022012.2 XM_011512621.2 XM_011512622.2 XM_011512623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4546159 c.393G>A p.R131R Chr.3: 130376562-130376562 on Build GRCh38
3060658 c.486G>A p.V162V Chr.3: 130376655-130376655 on Build GRCh38
7277103 c.480C>T p.D160D Chr.3: 130376649-130376649 on Build GRCh38
4157258 c.618C>A p.I206I Chr.3: 130376787-130376787 on Build GRCh38
5664187 c.521A>T p.E174V Chr.3: 130376690-130376690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780772048 Chr.3: 130376817-130376817 on Build GRCh38
rs765332150 Chr.3: 130376862-130376862 on Build GRCh38
rs759025488 Chr.3: 130376677-130376677 on Build GRCh38
rs1008843080 Chr.3: 130376970-130376970 on Build GRCh38
rs746721835 Chr.3: 130376765-130376765 on Build GRCh38
rs528892434 Chr.3: 130376666-130376666 on Build GRCh38
rs540088028 Chr.3: 130376918-130376918 on Build GRCh38
rs868668620 Chr.3: 130376599-130376599 on Build GRCh38
rs562362622 Chr.3: 130376655-130376655 on Build GRCh38
rs533903220 Chr.3: 130376630-130376630 on Build GRCh38
rs1041461048 Chr.3: 130376708-130376708 on Build GRCh38
rs550542345 Chr.3: 130376672-130376672 on Build GRCh38
rs934146720 Chr.3: 130376593-130376593 on Build GRCh38
rs778942743 Chr.3: 130376745-130376745 on Build GRCh38
rs557460823 Chr.3: 130376651-130376651 on Build GRCh38
rs190182061 Chr.3: 130376667-130376667 on Build GRCh38
rs539563406 Chr.3: 130376934-130376934 on Build GRCh38
rs528630596 Chr.3: 130376597-130376597 on Build GRCh38
rs757760527 Chr.3: 130376648-130376648 on Build GRCh38
rs566688640 Chr.3: 130376827-130376827 on Build GRCh38
rs986985859 Chr.3: 130376870-130376870 on Build GRCh38
rs746079551 Chr.3: 130376562-130376562 on Build GRCh38
rs765935042 Chr.3: 130376620-130376620 on Build GRCh38
rs575359773 Chr.3: 130376915-130376915 on Build GRCh38
rs768593324 Chr.3: 130376591-130376591 on Build GRCh38
rs746057781 Chr.3: 130376780-130376800 on Build GRCh38
rs772162967 Chr.3: 130376851-130376851 on Build GRCh38
rs777760382 Chr.3: 130376650-130376650 on Build GRCh38
rs760223815 Chr.3: 130376618-130376618 on Build GRCh38
rs779952685 Chr.3: 130376568-130376568 on Build GRCh38
rs567509738 Chr.3: 130376888-130376888 on Build GRCh38
rs949275728 Chr.3: 130376925-130376925 on Build GRCh38
rs577358826 Chr.3: 130376880-130376880 on Build GRCh38
rs752654130 Chr.3: 130376752-130376752 on Build GRCh38
rs777627967 Chr.3: 130376871-130376871 on Build GRCh38
rs115250607 Chr.3: 130376755-130376755 on Build GRCh38
rs773758829 Chr.3: 130376831-130376831 on Build GRCh38
rs748582738 Chr.3: 130376937-130376937 on Build GRCh38
rs747996709 Chr.3: 130376602-130376602 on Build GRCh38
rs755092854 Chr.3: 130376740-130376740 on Build GRCh38
rs556929903 Chr.3: 130376903-130376903 on Build GRCh38
rs555376244 Chr.3: 130376872-130376872 on Build GRCh38
rs771851852 Chr.3: 130376612-130376612 on Build GRCh38
rs114265933 Chr.3: 130376977-130376977 on Build GRCh38
rs193128156 Chr.3: 130376940-130376940 on Build GRCh38
rs150453992 Chr.3: 130376908-130376908 on Build GRCh38
rs933460620 Chr.3: 130376627-130376627 on Build GRCh38
rs1000801978 Chr.3: 130376935-130376935 on Build GRCh38
rs772943230 Chr.3: 130376671-130376671 on Build GRCh38
rs771904641 Chr.3: 130376668-130376668 on Build GRCh38
rs773587730 Chr.3: 130376938-130376938 on Build GRCh38
rs775088362 Chr.3: 130376825-130376825 on Build GRCh38
rs757039801 Chr.3: 130376886-130376886 on Build GRCh38
rs972150963 Chr.3: 130376583-130376583 on Build GRCh38
rs772653188 Chr.3: 130376830-130376830 on Build GRCh38
rs1030535985 Chr.3: 130376959-130376959 on Build GRCh38
rs749397417 Chr.3: 130376680-130376680 on Build GRCh38
rs938482632 Chr.3: 130376958-130376958 on Build GRCh38
rs749284239 Chr.3: 130376578-130376578 on Build GRCh38
rs553610181 Chr.3: 130376920-130376920 on Build GRCh38
rs754058364 Chr.3: 130376851-130376851 on Build GRCh38
rs781367899 Chr.3: 130376865-130376865 on Build GRCh38
rs569020480 Chr.3: 130376737-130376737 on Build GRCh38
rs1012715883 Chr.3: 130376976-130376976 on Build GRCh38
rs866090604 Chr.3: 130376573-130376573 on Build GRCh38
rs777628497 Chr.3: 130376760-130376760 on Build GRCh38
rs551636493 Chr.3: 130376818-130376818 on Build GRCh38
rs772891373 Chr.3: 130376615-130376615 on Build GRCh38
rs1009197582 Chr.3: 130376955-130376955 on Build GRCh38
rs535519733 Chr.3: 130376902-130376902 on Build GRCh38
rs184907722 Chr.3: 130376993-130376993 on Build GRCh38
rs1047591278 Chr.3: 130376910-130376910 on Build GRCh38
rs993426238 Chr.3: 130376987-130376987 on Build GRCh38
rs769647962 Chr.3: 130376594-130376595 on Build GRCh38
rs766737025 Chr.3: 130376848-130376848 on Build GRCh38
rs958867862 Chr.3: 130376773-130376773 on Build GRCh38
rs768028937 Chr.3: 130376713-130376713 on Build GRCh38
rs769473553 Chr.3: 130376820-130376820 on Build GRCh38
rs868003709 Chr.3: 130376907-130376907 on Build GRCh38
rs770744901 Chr.3: 130376799-130376799 on Build GRCh38
rs775035423 Chr.3: 130376695-130376695 on Build GRCh38
rs756254872 Chr.3: 130376566-130376566 on Build GRCh38
rs758150812 Chr.3: 130376792-130376792 on Build GRCh38
rs6787083 Chr.3: 130376649-130376649 on Build GRCh38
rs564327718 Chr.3: 130376574-130376574 on Build GRCh38
rs140689933 Chr.3: 130376864-130376864 on Build GRCh38
rs774149097 Chr.3: 130376598-130376598 on Build GRCh38
rs1044906027 Chr.3: 130376927-130376927 on Build GRCh38
rs745462929 Chr.3: 130376712-130376712 on Build GRCh38
rs759707071 Chr.3: 130376858-130376858 on Build GRCh38
rs775678864 Chr.3: 130376862-130376863 on Build GRCh38
rs763518867 Chr.3: 130376626-130376626 on Build GRCh38
rs769415870 Chr.3: 130376998-130376999 on Build GRCh38
rs752096481 Chr.3: 130376636-130376636 on Build GRCh38
rs919303318 Chr.3: 130376874-130376874 on Build GRCh38
rs779039342 Chr.3: 130376751-130376753 on Build GRCh38
rs754088234 Chr.3: 130376738-130376738 on Build GRCh38
rs760967939 Chr.3: 130376730-130376730 on Build GRCh38
rs770423427 Chr.3: 130376674-130376674 on Build GRCh38
rs1013379929 Chr.3: 130376732-130376732 on Build GRCh38
rs775963389 Chr.3: 130376624-130376624 on Build GRCh38
rs761014662 Chr.3: 130376847-130376847 on Build GRCh38
rs954479652 Chr.3: 130376968-130376968 on Build GRCh38

View Full Product Details