Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 132327879-132328374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACPP
Entrez Gene ID
55
Gene Name
acid phosphatase, prostate
Gene Aliases
5'-NT, ACP-3, ACP3
UniGene
Hs.433060
Gene Chromosome Location
Chr.3: 132317367-132368765 on Build GRCh38
Transcripts
NM_001099.4 NM_001134194.1 NM_001292037.1 XM_011512946.1 XM_011512947.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3587553 c.133G>A p.G45R Chr.3: 132328279-132328279 on Build GRCh38
7245552 c.185C>G p.S62* Chr.3: 132328331-132328331 on Build GRCh38
3587555 c.160C>T p.P54S Chr.3: 132328306-132328306 on Build GRCh38
1670572 c.203G>A p.G68D Chr.3: 132328349-132328349 on Build GRCh38
3914948 c.140G>A p.R47Q Chr.3: 132328286-132328286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757458583 Chr.3: 132328013-132328013 on Build GRCh38
rs779150269 Chr.3: 132328038-132328038 on Build GRCh38
rs377523594 Chr.3: 132328319-132328319 on Build GRCh38
rs117192307 Chr.3: 132327922-132327922 on Build GRCh38
rs1036276983 Chr.3: 132328071-132328071 on Build GRCh38
rs372279879 Chr.3: 132328179-132328180 on Build GRCh38
rs370760828 Chr.3: 132328324-132328324 on Build GRCh38
rs754914285 Chr.3: 132328032-132328032 on Build GRCh38
rs987275146 Chr.3: 132328331-132328331 on Build GRCh38
rs78007362 Chr.3: 132328196-132328196 on Build GRCh38
rs753385336 Chr.3: 132328241-132328241 on Build GRCh38
rs907352936 Chr.3: 132328069-132328069 on Build GRCh38
rs773049144 Chr.3: 132328303-132328303 on Build GRCh38
rs758944793 Chr.3: 132328246-132328246 on Build GRCh38
rs867400909 Chr.3: 132328284-132328284 on Build GRCh38
rs957616180 Chr.3: 132327907-132327907 on Build GRCh38
rs893179405 Chr.3: 132328180-132328180 on Build GRCh38
rs753337865 Chr.3: 132328327-132328327 on Build GRCh38
rs919208419 Chr.3: 132328075-132328075 on Build GRCh38
rs202047976 Chr.3: 132328350-132328350 on Build GRCh38
rs1051619785 Chr.3: 132328131-132328131 on Build GRCh38
rs780954030 Chr.3: 132328145-132328145 on Build GRCh38
rs552223731 Chr.3: 132327921-132327921 on Build GRCh38
rs965839810 Chr.3: 132328285-132328285 on Build GRCh38
rs933126737 Chr.3: 132328106-132328106 on Build GRCh38
rs1039675832 Chr.3: 132328166-132328166 on Build GRCh38
rs1018855837 Chr.3: 132328280-132328280 on Build GRCh38
rs781475900 Chr.3: 132328273-132328273 on Build GRCh38
rs774465017 Chr.3: 132328295-132328295 on Build GRCh38
rs958716550 Chr.3: 132327956-132327956 on Build GRCh38
rs759069721 Chr.3: 132328333-132328333 on Build GRCh38
rs532849099 Chr.3: 132327949-132327949 on Build GRCh38
rs200755203 Chr.3: 132328288-132328288 on Build GRCh38
rs974512425 Chr.3: 132328286-132328286 on Build GRCh38
rs764660360 Chr.3: 132328340-132328340 on Build GRCh38
rs375142881 Chr.3: 132328232-132328232 on Build GRCh38
rs772008636 Chr.3: 132328302-132328302 on Build GRCh38
rs942792809 Chr.3: 132328144-132328144 on Build GRCh38
rs908876766 Chr.3: 132328006-132328006 on Build GRCh38
rs746133481 Chr.3: 132328274-132328274 on Build GRCh38
rs368525246 Chr.3: 132328185-132328186 on Build GRCh38
rs928377505 Chr.3: 132327952-132327952 on Build GRCh38
rs757622848 Chr.3: 132328269-132328269 on Build GRCh38
rs768346500 Chr.3: 132328327-132328329 on Build GRCh38
rs940892811 Chr.3: 132328001-132328001 on Build GRCh38
rs75763236 Chr.3: 132328197-132328197 on Build GRCh38
rs752020438 Chr.3: 132328256-132328256 on Build GRCh38
rs746185287 Chr.3: 132328122-132328122 on Build GRCh38
rs778231613 Chr.3: 132328248-132328248 on Build GRCh38
rs749361161 Chr.3: 132328291-132328291 on Build GRCh38
rs891187524 Chr.3: 132328137-132328137 on Build GRCh38
rs760299722 Chr.3: 132328246-132328247 on Build GRCh38
rs907552652 Chr.3: 132327990-132327990 on Build GRCh38
rs967519287 Chr.3: 132327939-132327939 on Build GRCh38
rs147157265 Chr.3: 132328186-132328186 on Build GRCh38
rs566257634 Chr.3: 132328104-132328104 on Build GRCh38
rs974460236 Chr.3: 132327906-132327906 on Build GRCh38
rs981611162 Chr.3: 132327951-132327951 on Build GRCh38
rs547814873 Chr.3: 132327968-132327968 on Build GRCh38
rs765865589 Chr.3: 132328233-132328233 on Build GRCh38
rs556435640 Chr.3: 132327925-132327925 on Build GRCh38
rs903476919 Chr.3: 132328242-132328242 on Build GRCh38
rs374816958 Chr.3: 132328353-132328353 on Build GRCh38
rs780034526 Chr.3: 132328289-132328289 on Build GRCh38
rs747998417 Chr.3: 132328349-132328349 on Build GRCh38
rs933244215 Chr.3: 132328329-132328329 on Build GRCh38
rs761779572 Chr.3: 132328301-132328301 on Build GRCh38
rs768803310 Chr.3: 132328294-132328294 on Build GRCh38
rs1011618276 Chr.3: 132328186-132328186 on Build GRCh38
rs751582238 Chr.3: 132327976-132327976 on Build GRCh38
rs530483320 Chr.3: 132327909-132327909 on Build GRCh38

View Full Product Details