Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 132434446-132434941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC13
Entrez Gene ID
23317
Gene Name
DnaJ heat shock protein family (Hsp40) member C13
Gene Aliases
PARK21, RME8
UniGene
Hs.12707
Gene Chromosome Location
Chr.3: 132417527-132539032 on Build GRCh38
Transcripts
NM_015268.3 XM_005247245.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1616924 c.9A>G p.I3M Chr.3: 132434559-132434559 on Build GRCh38
5894786 c.62G>A p.R21K Chr.3: 132434612-132434612 on Build GRCh38
1038394 c.3G>T p.M1I Chr.3: 132434553-132434553 on Build GRCh38
1419065 c.62delG p.K23fs*21 Chr.3: 132434612-132434612 on Build GRCh38
1038395 c.10A>T p.I4F Chr.3: 132434560-132434560 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971425709 Chr.3: 132434836-132434836 on Build GRCh38
rs764310391 Chr.3: 132434602-132434602 on Build GRCh38
rs980329566 Chr.3: 132434838-132434838 on Build GRCh38
rs374504522 Chr.3: 132434865-132434865 on Build GRCh38
rs772803277 Chr.3: 132434663-132434663 on Build GRCh38
rs753166828 Chr.3: 132434570-132434570 on Build GRCh38
rs760212351 Chr.3: 132434668-132434668 on Build GRCh38
rs10662251 Chr.3: 132434489-132434490 on Build GRCh38
rs900593346 Chr.3: 132434488-132434488 on Build GRCh38
rs971604037 Chr.3: 132434499-132434499 on Build GRCh38
rs751869200 Chr.3: 132434603-132434603 on Build GRCh38
rs756954400 Chr.3: 132434529-132434529 on Build GRCh38
rs183429966 Chr.3: 132434908-132434908 on Build GRCh38
rs781468753 Chr.3: 132434621-132434621 on Build GRCh38
rs371828302 Chr.3: 132434518-132434518 on Build GRCh38
rs751327288 Chr.3: 132434509-132434510 on Build GRCh38
rs765358292 Chr.3: 132434645-132434646 on Build GRCh38
rs374320101 Chr.3: 132434618-132434618 on Build GRCh38
rs756219690 Chr.3: 132434629-132434629 on Build GRCh38
rs1049169621 Chr.3: 132434793-132434793 on Build GRCh38
rs201871185 Chr.3: 132434485-132434486 on Build GRCh38
rs927562973 Chr.3: 132434851-132434851 on Build GRCh38
rs989217465 Chr.3: 132434646-132434646 on Build GRCh38
rs542434759 Chr.3: 132434698-132434698 on Build GRCh38
rs906438143 Chr.3: 132434538-132434538 on Build GRCh38
rs886470357 Chr.3: 132434516-132434516 on Build GRCh38
rs956161214 Chr.3: 132434469-132434469 on Build GRCh38
rs772680510 Chr.3: 132434554-132434554 on Build GRCh38
rs927478844 Chr.3: 132434540-132434540 on Build GRCh38
rs749179677 Chr.3: 132434636-132434636 on Build GRCh38
rs956068012 Chr.3: 132434772-132434773 on Build GRCh38
rs922923684 Chr.3: 132434890-132434890 on Build GRCh38
rs191648146 Chr.3: 132434671-132434671 on Build GRCh38
rs747876962 Chr.3: 132434654-132434654 on Build GRCh38
rs750494611 Chr.3: 132434653-132434654 on Build GRCh38
rs554037554 Chr.3: 132434625-132434625 on Build GRCh38
rs1003384423 Chr.3: 132434549-132434549 on Build GRCh38
rs532134373 Chr.3: 132434877-132434877 on Build GRCh38
rs758798825 Chr.3: 132434594-132434594 on Build GRCh38
rs761357679 Chr.3: 132434525-132434525 on Build GRCh38
rs548737717 Chr.3: 132434626-132434628 on Build GRCh38
rs1039170978 Chr.3: 132434471-132434471 on Build GRCh38
rs996047962 Chr.3: 132434680-132434680 on Build GRCh38
rs758831832 Chr.3: 132434664-132434665 on Build GRCh38
rs771602655 Chr.3: 132434658-132434658 on Build GRCh38
rs977725681 Chr.3: 132434725-132434725 on Build GRCh38
rs765757323 Chr.3: 132434562-132434562 on Build GRCh38
rs199617861 Chr.3: 132434636-132434637 on Build GRCh38
rs1049541177 Chr.3: 132434515-132434515 on Build GRCh38
rs142883324 Chr.3: 132434488-132434489 on Build GRCh38
rs922094945 Chr.3: 132434772-132434772 on Build GRCh38
rs368925770 Chr.3: 132434878-132434878 on Build GRCh38
rs768552598 Chr.3: 132434645-132434645 on Build GRCh38
rs757446714 Chr.3: 132434637-132434637 on Build GRCh38
rs930720837 Chr.3: 132434494-132434494 on Build GRCh38
rs72990500 Chr.3: 132434768-132434768 on Build GRCh38
rs780144226 Chr.3: 132434631-132434631 on Build GRCh38
rs774011664 Chr.3: 132434653-132434653 on Build GRCh38
rs760085443 Chr.3: 132434556-132434556 on Build GRCh38
rs35680953 Chr.3: 132434490-132434491 on Build GRCh38
rs1024302437 Chr.3: 132434827-132434827 on Build GRCh38
rs917256489 Chr.3: 132434476-132434476 on Build GRCh38
rs886520220 Chr.3: 132434796-132434796 on Build GRCh38
rs1033477597 Chr.3: 132434616-132434616 on Build GRCh38
rs930792501 Chr.3: 132434787-132434787 on Build GRCh38
rs1045725280 Chr.3: 132434858-132434858 on Build GRCh38
rs1039984605 Chr.3: 132434634-132434634 on Build GRCh38
rs560775183 Chr.3: 132434730-132434730 on Build GRCh38
rs150159097 Chr.3: 132434615-132434615 on Build GRCh38
rs966428289 Chr.3: 132434708-132434708 on Build GRCh38
rs908044978 Chr.3: 132434876-132434876 on Build GRCh38
rs759326297 Chr.3: 132434622-132434623 on Build GRCh38
rs940704494 Chr.3: 132434846-132434846 on Build GRCh38
rs944927371 Chr.3: 132434754-132434754 on Build GRCh38
rs1050796928 Chr.3: 132434695-132434695 on Build GRCh38
rs543152074 Chr.3: 132434808-132434808 on Build GRCh38
rs1040139371 Chr.3: 132434888-132434888 on Build GRCh38

View Full Product Details