Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 132453493-132453997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC13
Entrez Gene ID
23317
Gene Name
DnaJ heat shock protein family (Hsp40) member C13
Gene Aliases
PARK21, RME8
UniGene
Hs.12707
Gene Chromosome Location
Chr.3: 132417527-132539032 on Build GRCh38
Transcripts
NM_015268.3 XM_005247245.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
228366 c.751G>C p.V251L Chr.3: 132453605-132453605 on Build GRCh38
6647248 c.801G>A p.P267P Chr.3: 132453655-132453655 on Build GRCh38
5995200 c.779C>T p.T260I Chr.3: 132453633-132453633 on Build GRCh38
7365067 c.749C>T p.P250L Chr.3: 132453603-132453603 on Build GRCh38
7417096 c.734C>A p.P245H Chr.3: 132453494-132453494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773709434 Chr.3: 132453562-132453562 on Build GRCh38
rs533887358 Chr.3: 132453947-132453947 on Build GRCh38
rs754276059 Chr.3: 132453589-132453589 on Build GRCh38
rs1051811683 Chr.3: 132453672-132453672 on Build GRCh38
rs937175337 Chr.3: 132453814-132453814 on Build GRCh38
rs79808031 Chr.3: 132453583-132453583 on Build GRCh38
rs953678210 Chr.3: 132453880-132453880 on Build GRCh38
rs769650097 Chr.3: 132453531-132453531 on Build GRCh38
rs765507211 Chr.3: 132453599-132453599 on Build GRCh38
rs757375779 Chr.3: 132453634-132453634 on Build GRCh38
rs949374216 Chr.3: 132453780-132453780 on Build GRCh38
rs1019996151 Chr.3: 132453527-132453527 on Build GRCh38
rs368364734 Chr.3: 132453582-132453582 on Build GRCh38
rs1004381480 Chr.3: 132453793-132453793 on Build GRCh38
rs532780555 Chr.3: 132453677-132453677 on Build GRCh38
rs547164536 Chr.3: 132453819-132453819 on Build GRCh38
rs761262838 Chr.3: 132453576-132453576 on Build GRCh38
rs185762958 Chr.3: 132453832-132453832 on Build GRCh38
rs1055665941 Chr.3: 132453822-132453822 on Build GRCh38
rs978429964 Chr.3: 132453881-132453881 on Build GRCh38
rs776664452 Chr.3: 132453521-132453521 on Build GRCh38
rs1053398963 Chr.3: 132453666-132453666 on Build GRCh38
rs146473569 Chr.3: 132453944-132453944 on Build GRCh38
rs139307487 Chr.3: 132453588-132453589 on Build GRCh38
rs760034142 Chr.3: 132453727-132453727 on Build GRCh38
rs569244389 Chr.3: 132453848-132453848 on Build GRCh38
rs964352467 Chr.3: 132453849-132453849 on Build GRCh38
rs540403662 Chr.3: 132453556-132453556 on Build GRCh38
rs997039970 Chr.3: 132453903-132453903 on Build GRCh38
rs190110375 Chr.3: 132453735-132453735 on Build GRCh38
rs370652045 Chr.3: 132453946-132453946 on Build GRCh38
rs181188175 Chr.3: 132453829-132453829 on Build GRCh38
rs888735043 Chr.3: 132453790-132453790 on Build GRCh38
rs185229166 Chr.3: 132453729-132453729 on Build GRCh38
rs964574934 Chr.3: 132453573-132453573 on Build GRCh38
rs774006453 Chr.3: 132453642-132453642 on Build GRCh38
rs543905953 Chr.3: 132453603-132453603 on Build GRCh38
rs892968021 Chr.3: 132453826-132453826 on Build GRCh38
rs902858399 Chr.3: 132453842-132453842 on Build GRCh38
rs573676097 Chr.3: 132453591-132453591 on Build GRCh38
rs777960726 Chr.3: 132453619-132453619 on Build GRCh38
rs1027206185 Chr.3: 132453757-132453757 on Build GRCh38
rs768213999 Chr.3: 132453683-132453683 on Build GRCh38
rs771512627 Chr.3: 132453705-132453705 on Build GRCh38
rs188129203 Chr.3: 132453938-132453938 on Build GRCh38
rs193143491 Chr.3: 132453655-132453655 on Build GRCh38
rs1020305526 Chr.3: 132453845-132453845 on Build GRCh38
rs551107263 Chr.3: 132453844-132453844 on Build GRCh38
rs1046488674 Chr.3: 132453706-132453706 on Build GRCh38
rs1046068328 Chr.3: 132453797-132453797 on Build GRCh38
rs549716405 Chr.3: 132453754-132453754 on Build GRCh38
rs200373363 Chr.3: 132453654-132453654 on Build GRCh38
rs752160880 Chr.3: 132453726-132453729 on Build GRCh38
rs777180636 Chr.3: 132453720-132453720 on Build GRCh38
rs374949489 Chr.3: 132453694-132453694 on Build GRCh38
rs1028093032 Chr.3: 132453875-132453875 on Build GRCh38
rs769775577 Chr.3: 132453648-132453648 on Build GRCh38
rs761316203 Chr.3: 132453697-132453697 on Build GRCh38
rs928539753 Chr.3: 132453802-132453802 on Build GRCh38
rs558102985 Chr.3: 132453543-132453543 on Build GRCh38
rs7613098 Chr.3: 132453551-132453551 on Build GRCh38
rs908589492 Chr.3: 132453963-132453963 on Build GRCh38
rs759114138 Chr.3: 132453732-132453732 on Build GRCh38
rs555490967 Chr.3: 132453967-132453967 on Build GRCh38
rs539705635 Chr.3: 132453525-132453525 on Build GRCh38
rs758519786 Chr.3: 132453609-132453609 on Build GRCh38
rs751616278 Chr.3: 132453628-132453628 on Build GRCh38
rs920073492 Chr.3: 132453617-132453617 on Build GRCh38
rs528818930 Chr.3: 132453795-132453795 on Build GRCh38
rs781283510 Chr.3: 132453641-132453641 on Build GRCh38

View Full Product Details