Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 132602360-132602794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD11
Entrez Gene ID
84129
Gene Name
acyl-CoA dehydrogenase family member 11
Gene Aliases
ACAD-11
UniGene
Hs.441378
Gene Chromosome Location
Chr.3: 132558138-132660131 on Build GRCh38
Transcripts
NM_032169.4 NR_132426.1 NR_132427.1 NR_132428.1 NM_016557.3 NM_178445.2 NR_037804.1

Gene Symbol
ACKR4
Entrez Gene ID
51554
Gene Name
atypical chemokine receptor 4
Gene Aliases
CC-CKR-11, CCBP2, CCR-11, CCR10, CCR11, CCRL1, CCX CKR, CCX-CKR, CKR-11, PPR1, VSHK1
UniGene
Hs.310512
Gene Chromosome Location
Chr.3: 132597237-132602644 on Build GRCh38
Transcripts
NM_032169.4 NR_132426.1 NR_132427.1 NR_132428.1 NM_016557.3 NM_178445.2 NR_037804.1

Gene Symbol
NPHP3-ACAD11
Entrez Gene ID
100532724
Gene Name
NPHP3-ACAD11 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.511991
Gene Chromosome Location
Chr.3: 132558138-132722459 on Build GRCh38
Transcripts
NM_032169.4 NR_132426.1 NR_132427.1 NR_132428.1 NM_016557.3 NM_178445.2 NR_037804.1

SNP Detail

SNP ID Chromosome Location
rs961643575 Chr.3: 132602665-132602665 on Build GRCh38
rs535880535 Chr.3: 132602559-132602559 on Build GRCh38
rs1015477673 Chr.3: 132602430-132602430 on Build GRCh38
rs557291993 Chr.3: 132602567-132602567 on Build GRCh38
rs62291492 Chr.3: 132602398-132602398 on Build GRCh38
rs952501001 Chr.3: 132602395-132602395 on Build GRCh38
rs559259964 Chr.3: 132602679-132602679 on Build GRCh38
rs1001298017 Chr.3: 132602559-132602560 on Build GRCh38
rs932324524 Chr.3: 132602625-132602625 on Build GRCh38
rs754149111 Chr.3: 132602582-132602582 on Build GRCh38
rs958668517 Chr.3: 132602742-132602742 on Build GRCh38
rs991454887 Chr.3: 132602749-132602749 on Build GRCh38
rs796247302 Chr.3: 132602761-132602761 on Build GRCh38
rs909889528 Chr.3: 132602639-132602639 on Build GRCh38
rs775170434 Chr.3: 132602451-132602451 on Build GRCh38
rs779037054 Chr.3: 132602757-132602757 on Build GRCh38
rs982084992 Chr.3: 132602469-132602469 on Build GRCh38
rs971054739 Chr.3: 132602432-132602432 on Build GRCh38
rs1004011931 Chr.3: 132602673-132602673 on Build GRCh38
rs991509009 Chr.3: 132602517-132602517 on Build GRCh38
rs1034126274 Chr.3: 132602561-132602561 on Build GRCh38
rs1041330730 Chr.3: 132602591-132602591 on Build GRCh38
rs899483469 Chr.3: 132602612-132602612 on Build GRCh38
rs1003852378 Chr.3: 132602702-132602702 on Build GRCh38
rs564262883 Chr.3: 132602743-132602743 on Build GRCh38
rs148663258 Chr.3: 132602472-132602472 on Build GRCh38
rs914669378 Chr.3: 132602547-132602547 on Build GRCh38
rs944280588 Chr.3: 132602589-132602589 on Build GRCh38
rs891454200 Chr.3: 132602541-132602541 on Build GRCh38
rs1049524729 Chr.3: 132602431-132602431 on Build GRCh38
rs142180732 Chr.3: 132602764-132602764 on Build GRCh38
rs1033575034 Chr.3: 132602722-132602722 on Build GRCh38
rs75518936 Chr.3: 132602509-132602509 on Build GRCh38
rs575538001 Chr.3: 132602636-132602636 on Build GRCh38
rs906394630 Chr.3: 132602686-132602686 on Build GRCh38
rs539045453 Chr.3: 132602650-132602650 on Build GRCh38
rs182981967 Chr.3: 132602713-132602713 on Build GRCh38
rs984160707 Chr.3: 132602635-132602635 on Build GRCh38
rs973024106 Chr.3: 132602671-132602671 on Build GRCh38

View Full Product Details