Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 134546167-134546678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP63
Entrez Gene ID
80254
Gene Name
centrosomal protein 63
Gene Aliases
SCKL6
UniGene
Hs.443301
Gene Chromosome Location
Chr.3: 134485733-134629607 on Build GRCh38
Transcripts
NM_001042383.1 NM_001042384.1 NM_001042400.1 NM_025180.3 XM_005247795.4 XM_005247797.3 XM_005247799.3 XM_006713760.3 XM_011513194.2 XM_011513197.2 XM_017007244.1 XM_017007245.1 XM_017007246.1 XM_017007247.1 XM_017007248.1 XM_017007249.1 XM_017007250.1 XM_017007251.1 XM_017007252.1 XM_017007253.1 XM_017007254.1 XM_017007255.1 XM_017007256.1 XM_017007257.1 XM_017007258.1 XM_017007259.1 XM_017007260.1 XM_017007261.1 XM_017007262.1 XM_017007263.1 XM_017007264.1 XM_017007265.1 XM_017007266.1 XM_017007267.1 XM_017007268.1 XM_017007269.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7232440 c.925A>T p.I309L Chr.3: 134546284-134546284 on Build GRCh38
4931430 c.894G>T p.K298N Chr.3: 134546253-134546253 on Build GRCh38
7186598 c.925A>G p.I309V Chr.3: 134546284-134546284 on Build GRCh38
1038669 c.811G>T p.E271* Chr.3: 134546170-134546170 on Build GRCh38
398217 c.823G>T p.A275S Chr.3: 134546182-134546182 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751775921 Chr.3: 134546268-134546268 on Build GRCh38
rs765275868 Chr.3: 134546225-134546225 on Build GRCh38
rs71139539 Chr.3: 134546376-134546377 on Build GRCh38
rs781407073 Chr.3: 134546245-134546249 on Build GRCh38
rs143098200 Chr.3: 134546350-134546350 on Build GRCh38
rs960037320 Chr.3: 134546643-134546643 on Build GRCh38
rs973387357 Chr.3: 134546319-134546319 on Build GRCh38
rs773159512 Chr.3: 134546338-134546338 on Build GRCh38
rs750447493 Chr.3: 134546261-134546261 on Build GRCh38
rs903527592 Chr.3: 134546547-134546547 on Build GRCh38
rs753630215 Chr.3: 134546190-134546190 on Build GRCh38
rs781687192 Chr.3: 134546291-134546291 on Build GRCh38
rs979807595 Chr.3: 134546210-134546210 on Build GRCh38
rs778387697 Chr.3: 134546304-134546304 on Build GRCh38
rs969801726 Chr.3: 134546199-134546199 on Build GRCh38
rs547682772 Chr.3: 134546336-134546336 on Build GRCh38
rs147417466 Chr.3: 134546365-134546365 on Build GRCh38
rs865834445 Chr.3: 134546493-134546493 on Build GRCh38
rs111445863 Chr.3: 134546451-134546451 on Build GRCh38
rs549119355 Chr.3: 134546638-134546638 on Build GRCh38
rs976003122 Chr.3: 134546211-134546211 on Build GRCh38
rs980004250 Chr.3: 134546273-134546273 on Build GRCh38
rs890526010 Chr.3: 134546393-134546393 on Build GRCh38
rs527852053 Chr.3: 134546228-134546228 on Build GRCh38
rs1015476291 Chr.3: 134546650-134546650 on Build GRCh38
rs377061183 Chr.3: 134546224-134546224 on Build GRCh38
rs901710813 Chr.3: 134546571-134546571 on Build GRCh38
rs1035467445 Chr.3: 134546557-134546557 on Build GRCh38
rs144258361 Chr.3: 134546275-134546275 on Build GRCh38
rs528017525 Chr.3: 134546437-134546437 on Build GRCh38
rs1053267416 Chr.3: 134546458-134546458 on Build GRCh38
rs770182568 Chr.3: 134546300-134546300 on Build GRCh38
rs1051832806 Chr.3: 134546381-134546381 on Build GRCh38
rs749793131 Chr.3: 134546309-134546309 on Build GRCh38
rs1001192545 Chr.3: 134546468-134546468 on Build GRCh38
rs752662493 Chr.3: 134546333-134546333 on Build GRCh38
rs1017295525 Chr.3: 134546570-134546570 on Build GRCh38
rs980172940 Chr.3: 134546413-134546413 on Build GRCh38
rs536806788 Chr.3: 134546463-134546463 on Build GRCh38
rs761730966 Chr.3: 134546191-134546191 on Build GRCh38
rs1043788087 Chr.3: 134546545-134546545 on Build GRCh38
rs374330136 Chr.3: 134546644-134546644 on Build GRCh38
rs200936751 Chr.3: 134546205-134546205 on Build GRCh38
rs148333590 Chr.3: 134546332-134546332 on Build GRCh38
rs1053339438 Chr.3: 134546445-134546445 on Build GRCh38
rs142766824 Chr.3: 134546264-134546264 on Build GRCh38
rs774992680 Chr.3: 134546327-134546327 on Build GRCh38
rs755395532 Chr.3: 134546228-134546230 on Build GRCh38
rs748584464 Chr.3: 134546292-134546292 on Build GRCh38
rs760249099 Chr.3: 134546331-134546331 on Build GRCh38
rs573181710 Chr.3: 134546341-134546341 on Build GRCh38
rs960269083 Chr.3: 134546234-134546234 on Build GRCh38
rs947629186 Chr.3: 134546490-134546490 on Build GRCh38
rs771558556 Chr.3: 134546318-134546318 on Build GRCh38
rs548108649 Chr.3: 134546450-134546450 on Build GRCh38
rs997324853 Chr.3: 134546648-134546648 on Build GRCh38
rs1007298403 Chr.3: 134546548-134546548 on Build GRCh38

View Full Product Details