Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138010084-138010590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN18
Entrez Gene ID
51208
Gene Name
claudin 18
Gene Aliases
SFTA5, SFTPJ
UniGene
Hs.655324
Gene Chromosome Location
Chr.3: 137998816-138033652 on Build GRCh38
Transcripts
NM_001002026.2 NM_016369.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
279922 c.145C>T p.L49F Chr.3: 138010370-138010370 on Build GRCh38
291861 c.160G>A p.V54M Chr.3: 138010385-138010385 on Build GRCh38
1726259 c.58G>A p.G20S Chr.3: 138010283-138010283 on Build GRCh38
7324372 c.30G>A p.A10A Chr.3: 138010255-138010255 on Build GRCh38
1726260 c.59G>T p.G20V Chr.3: 138010284-138010284 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768644447 Chr.3: 138010330-138010330 on Build GRCh38
rs750864362 Chr.3: 138010193-138010193 on Build GRCh38
rs1013655724 Chr.3: 138010168-138010168 on Build GRCh38
rs747439048 Chr.3: 138010204-138010204 on Build GRCh38
rs912287789 Chr.3: 138010393-138010393 on Build GRCh38
rs376378448 Chr.3: 138010341-138010341 on Build GRCh38
rs781764259 Chr.3: 138010283-138010283 on Build GRCh38
rs939773412 Chr.3: 138010400-138010400 on Build GRCh38
rs753279758 Chr.3: 138010468-138010468 on Build GRCh38
rs906212531 Chr.3: 138010439-138010439 on Build GRCh38
rs761875755 Chr.3: 138010253-138010253 on Build GRCh38
rs201924367 Chr.3: 138010333-138010333 on Build GRCh38
rs763088441 Chr.3: 138010450-138010450 on Build GRCh38
rs372756802 Chr.3: 138010442-138010442 on Build GRCh38
rs1051588476 Chr.3: 138010152-138010152 on Build GRCh38
rs761451503 Chr.3: 138010435-138010436 on Build GRCh38
rs78868067 Chr.3: 138010482-138010482 on Build GRCh38
rs1055476916 Chr.3: 138010500-138010500 on Build GRCh38
rs371070256 Chr.3: 138010357-138010357 on Build GRCh38
rs1044710640 Chr.3: 138010396-138010396 on Build GRCh38
rs755675744 Chr.3: 138010211-138010211 on Build GRCh38
rs776132884 Chr.3: 138010378-138010379 on Build GRCh38
rs933221408 Chr.3: 138010343-138010343 on Build GRCh38
rs951589103 Chr.3: 138010291-138010291 on Build GRCh38
rs201678067 Chr.3: 138010352-138010352 on Build GRCh38
rs761927288 Chr.3: 138010345-138010345 on Build GRCh38
rs757411997 Chr.3: 138010187-138010187 on Build GRCh38
rs764171839 Chr.3: 138010248-138010248 on Build GRCh38
rs374810800 Chr.3: 138010209-138010209 on Build GRCh38
rs909983834 Chr.3: 138010294-138010294 on Build GRCh38
rs776623998 Chr.3: 138010251-138010251 on Build GRCh38
rs2289767 Chr.3: 138010142-138010142 on Build GRCh38
rs372332058 Chr.3: 138010504-138010504 on Build GRCh38
rs752124309 Chr.3: 138010275-138010275 on Build GRCh38
rs375872728 Chr.3: 138010389-138010389 on Build GRCh38
rs769881298 Chr.3: 138010421-138010421 on Build GRCh38
rs761088746 Chr.3: 138010332-138010334 on Build GRCh38
rs1033513415 Chr.3: 138010154-138010154 on Build GRCh38
rs775088096 Chr.3: 138010309-138010309 on Build GRCh38
rs570192498 Chr.3: 138010533-138010533 on Build GRCh38
rs374173675 Chr.3: 138010373-138010373 on Build GRCh38
rs746317426 Chr.3: 138010236-138010242 on Build GRCh38
rs866539095 Chr.3: 138010307-138010307 on Build GRCh38
rs771118569 Chr.3: 138010461-138010461 on Build GRCh38
rs958268174 Chr.3: 138010247-138010247 on Build GRCh38
rs894968354 Chr.3: 138010503-138010503 on Build GRCh38
rs748868334 Chr.3: 138010292-138010292 on Build GRCh38
rs746970570 Chr.3: 138010321-138010321 on Build GRCh38
rs933252466 Chr.3: 138010110-138010110 on Build GRCh38
rs771924320 Chr.3: 138010301-138010301 on Build GRCh38
rs374110052 Chr.3: 138010407-138010407 on Build GRCh38
rs373633928 Chr.3: 138010192-138010192 on Build GRCh38
rs771733842 Chr.3: 138010227-138010227 on Build GRCh38
rs764118969 Chr.3: 138010176-138010176 on Build GRCh38
rs768188619 Chr.3: 138010378-138010378 on Build GRCh38
rs748956270 Chr.3: 138010214-138010214 on Build GRCh38
rs781145268 Chr.3: 138010418-138010418 on Build GRCh38
rs753226794 Chr.3: 138010379-138010379 on Build GRCh38
rs144157849 Chr.3: 138010300-138010300 on Build GRCh38
rs1053547704 Chr.3: 138010182-138010182 on Build GRCh38
rs529393617 Chr.3: 138010509-138010509 on Build GRCh38
rs1013809212 Chr.3: 138010519-138010519 on Build GRCh38
rs149036233 Chr.3: 138010140-138010140 on Build GRCh38
rs774612228 Chr.3: 138010153-138010153 on Build GRCh38
rs1032346447 Chr.3: 138010216-138010216 on Build GRCh38
rs750376314 Chr.3: 138010450-138010456 on Build GRCh38
rs768414784 Chr.3: 138010409-138010409 on Build GRCh38
rs760479633 Chr.3: 138010244-138010244 on Build GRCh38
rs200350564 Chr.3: 138010391-138010391 on Build GRCh38
rs558942553 Chr.3: 138010282-138010282 on Build GRCh38
rs201156499 Chr.3: 138010384-138010384 on Build GRCh38
rs745725595 Chr.3: 138010226-138010226 on Build GRCh38
rs760849544 Chr.3: 138010175-138010175 on Build GRCh38
rs145987155 Chr.3: 138010420-138010420 on Build GRCh38
rs780656234 Chr.3: 138010203-138010203 on Build GRCh38
rs765498189 Chr.3: 138010259-138010259 on Build GRCh38
rs759969828 Chr.3: 138010377-138010377 on Build GRCh38
rs764586364 Chr.3: 138010436-138010437 on Build GRCh38
rs975424557 Chr.3: 138010318-138010318 on Build GRCh38
rs758674451 Chr.3: 138010202-138010202 on Build GRCh38
rs775529414 Chr.3: 138010241-138010241 on Build GRCh38
rs753511405 Chr.3: 138010337-138010337 on Build GRCh38
rs373852186 Chr.3: 138010271-138010271 on Build GRCh38
rs749967723 Chr.3: 138010486-138010486 on Build GRCh38
rs772583544 Chr.3: 138010291-138010292 on Build GRCh38
rs530659595 Chr.3: 138010463-138010463 on Build GRCh38
rs754081703 Chr.3: 138010186-138010186 on Build GRCh38
rs760019263 Chr.3: 138010464-138010464 on Build GRCh38
rs766775832 Chr.3: 138010367-138010367 on Build GRCh38
rs756859479 Chr.3: 138010382-138010382 on Build GRCh38
rs376611413 Chr.3: 138010408-138010408 on Build GRCh38
rs1054052863 Chr.3: 138010368-138010368 on Build GRCh38
rs895013344 Chr.3: 138010156-138010156 on Build GRCh38
rs750585890 Chr.3: 138010268-138010268 on Build GRCh38
rs764807195 Chr.3: 138010485-138010485 on Build GRCh38
rs763856935 Chr.3: 138010355-138010356 on Build GRCh38
rs368088796 Chr.3: 138010215-138010215 on Build GRCh38
rs756808279 Chr.3: 138010297-138010297 on Build GRCh38
rs1011897932 Chr.3: 138010198-138010198 on Build GRCh38
rs774923871 Chr.3: 138010183-138010184 on Build GRCh38
rs757910641 Chr.3: 138010495-138010495 on Build GRCh38
rs777363929 Chr.3: 138010212-138010212 on Build GRCh38
rs775953600 Chr.3: 138010296-138010296 on Build GRCh38

View Full Product Details