Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138023516-138024010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN18
Entrez Gene ID
51208
Gene Name
claudin 18
Gene Aliases
SFTA5, SFTPJ
UniGene
Hs.655324
Gene Chromosome Location
Chr.3: 137998816-138033652 on Build GRCh38
Transcripts
NM_001002026.2 NM_016369.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
81026 c.238C>A p.R80R Chr.3: 138023675-138023675 on Build GRCh38
3784338 c.273C>T p.A91A Chr.3: 138023710-138023710 on Build GRCh38
1718423 c.239G>A p.R80Q Chr.3: 138023676-138023676 on Build GRCh38
6855380 c.313C>A p.R105S Chr.3: 138023750-138023750 on Build GRCh38
6163543 c.283C>T p.L95L Chr.3: 138023720-138023720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773920904 Chr.3: 138023780-138023780 on Build GRCh38
rs761116930 Chr.3: 138023675-138023675 on Build GRCh38
rs759037089 Chr.3: 138023784-138023784 on Build GRCh38
rs574364248 Chr.3: 138023815-138023815 on Build GRCh38
rs894314610 Chr.3: 138023738-138023738 on Build GRCh38
rs777032007 Chr.3: 138023767-138023767 on Build GRCh38
rs544779988 Chr.3: 138023986-138023986 on Build GRCh38
rs139920646 Chr.3: 138023691-138023691 on Build GRCh38
rs529685225 Chr.3: 138023799-138023799 on Build GRCh38
rs767208390 Chr.3: 138023800-138023800 on Build GRCh38
rs112935391 Chr.3: 138023866-138023866 on Build GRCh38
rs779620883 Chr.3: 138023613-138023613 on Build GRCh38
rs558186375 Chr.3: 138023690-138023690 on Build GRCh38
rs529867138 Chr.3: 138023572-138023572 on Build GRCh38
rs35071725 Chr.3: 138023875-138023876 on Build GRCh38
rs200466463 Chr.3: 138023751-138023751 on Build GRCh38
rs781557344 Chr.3: 138023859-138023859 on Build GRCh38
rs748604223 Chr.3: 138023861-138023861 on Build GRCh38
rs1051145533 Chr.3: 138023727-138023727 on Build GRCh38
rs758527566 Chr.3: 138023848-138023848 on Build GRCh38
rs780462927 Chr.3: 138023854-138023854 on Build GRCh38
rs749210958 Chr.3: 138023712-138023712 on Build GRCh38
rs780912402 Chr.3: 138023634-138023634 on Build GRCh38
rs543312707 Chr.3: 138023818-138023818 on Build GRCh38
rs369419550 Chr.3: 138023556-138023556 on Build GRCh38
rs746093722 Chr.3: 138023730-138023730 on Build GRCh38
rs569480686 Chr.3: 138023638-138023638 on Build GRCh38
rs777755431 Chr.3: 138023650-138023650 on Build GRCh38
rs114998965 Chr.3: 138023699-138023699 on Build GRCh38
rs749380019 Chr.3: 138023651-138023651 on Build GRCh38
rs144886047 Chr.3: 138023701-138023701 on Build GRCh38
rs762329918 Chr.3: 138023774-138023774 on Build GRCh38
rs754564499 Chr.3: 138023692-138023692 on Build GRCh38
rs751179167 Chr.3: 138023614-138023614 on Build GRCh38
rs774747197 Chr.3: 138023659-138023659 on Build GRCh38
rs746038647 Chr.3: 138023670-138023670 on Build GRCh38
rs199724224 Chr.3: 138023680-138023680 on Build GRCh38
rs779079104 Chr.3: 138023717-138023717 on Build GRCh38
rs769692054 Chr.3: 138023643-138023643 on Build GRCh38
rs369712344 Chr.3: 138023750-138023750 on Build GRCh38
rs147784282 Chr.3: 138023547-138023547 on Build GRCh38
rs769032958 Chr.3: 138023766-138023766 on Build GRCh38
rs767157044 Chr.3: 138023696-138023696 on Build GRCh38
rs777615642 Chr.3: 138023711-138023711 on Build GRCh38
rs949699149 Chr.3: 138023686-138023686 on Build GRCh38
rs142630531 Chr.3: 138023618-138023618 on Build GRCh38
rs747380065 Chr.3: 138023764-138023764 on Build GRCh38
rs762406591 Chr.3: 138023689-138023689 on Build GRCh38
rs958693403 Chr.3: 138023577-138023577 on Build GRCh38
rs952434619 Chr.3: 138023552-138023552 on Build GRCh38
rs772579355 Chr.3: 138023671-138023671 on Build GRCh38
rs549770826 Chr.3: 138023855-138023855 on Build GRCh38
rs151166278 Chr.3: 138023776-138023776 on Build GRCh38
rs764565705 Chr.3: 138023676-138023676 on Build GRCh38
rs1018194869 Chr.3: 138023584-138023584 on Build GRCh38
rs752318671 Chr.3: 138023801-138023801 on Build GRCh38
rs753733515 Chr.3: 138023821-138023821 on Build GRCh38
rs192161007 Chr.3: 138023567-138023567 on Build GRCh38
rs887999939 Chr.3: 138023964-138023964 on Build GRCh38
rs1024029112 Chr.3: 138023982-138023982 on Build GRCh38
rs375347495 Chr.3: 138023845-138023845 on Build GRCh38
rs775929081 Chr.3: 138023672-138023672 on Build GRCh38
rs377074436 Chr.3: 138023716-138023716 on Build GRCh38
rs556809984 Chr.3: 138023829-138023829 on Build GRCh38
rs1004206091 Chr.3: 138023765-138023765 on Build GRCh38
rs778989278 Chr.3: 138023839-138023839 on Build GRCh38
rs1026557536 Chr.3: 138023551-138023551 on Build GRCh38

View Full Product Details