Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138163687-138164139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBR1
Entrez Gene ID
51163
Gene Name
debranching RNA lariats 1
Gene Aliases
-
UniGene
Hs.477700
Gene Chromosome Location
Chr.3: 138160988-138174949 on Build GRCh38
Transcripts
NM_016216.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1038992 c.732G>T p.Q244H Chr.3: 138163841-138163841 on Build GRCh38
5434438 c.793C>T p.Q265* Chr.3: 138163780-138163780 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767970705 Chr.3: 138163906-138163907 on Build GRCh38
rs919460286 Chr.3: 138163985-138163985 on Build GRCh38
rs906926717 Chr.3: 138163831-138163831 on Build GRCh38
rs145130716 Chr.3: 138164030-138164030 on Build GRCh38
rs375069331 Chr.3: 138163879-138163879 on Build GRCh38
rs2622733 Chr.3: 138163910-138163910 on Build GRCh38
rs779512592 Chr.3: 138163819-138163819 on Build GRCh38
rs771020405 Chr.3: 138163777-138163777 on Build GRCh38
rs892282429 Chr.3: 138163787-138163787 on Build GRCh38
rs769824503 Chr.3: 138163885-138163885 on Build GRCh38
rs745777240 Chr.3: 138163924-138163924 on Build GRCh38
rs575779048 Chr.3: 138163715-138163715 on Build GRCh38
rs577970013 Chr.3: 138163923-138163923 on Build GRCh38
rs906172844 Chr.3: 138164093-138164093 on Build GRCh38
rs763276806 Chr.3: 138163768-138163768 on Build GRCh38
rs544377797 Chr.3: 138163921-138163921 on Build GRCh38
rs748093704 Chr.3: 138163741-138163741 on Build GRCh38
rs768020164 Chr.3: 138163788-138163788 on Build GRCh38
rs966294379 Chr.3: 138163933-138163933 on Build GRCh38
rs964929705 Chr.3: 138163941-138163941 on Build GRCh38
rs774803255 Chr.3: 138163778-138163778 on Build GRCh38
rs909604528 Chr.3: 138164044-138164044 on Build GRCh38
rs141555371 Chr.3: 138163806-138163806 on Build GRCh38
rs755063993 Chr.3: 138163897-138163899 on Build GRCh38
rs529580688 Chr.3: 138164047-138164047 on Build GRCh38
rs749857302 Chr.3: 138163794-138163794 on Build GRCh38
rs777940515 Chr.3: 138163891-138163891 on Build GRCh38
rs2724712 Chr.3: 138164107-138164107 on Build GRCh38
rs939313181 Chr.3: 138164105-138164105 on Build GRCh38
rs377091967 Chr.3: 138163863-138163863 on Build GRCh38
rs767556869 Chr.3: 138163933-138163933 on Build GRCh38
rs1044712977 Chr.3: 138164085-138164085 on Build GRCh38
rs367901122 Chr.3: 138163793-138163793 on Build GRCh38
rs759763750 Chr.3: 138163780-138163780 on Build GRCh38
rs558173849 Chr.3: 138163922-138163922 on Build GRCh38
rs555237675 Chr.3: 138164031-138164035 on Build GRCh38
rs774373355 Chr.3: 138163908-138163908 on Build GRCh38
rs754886465 Chr.3: 138163833-138163833 on Build GRCh38
rs6810238 Chr.3: 138164058-138164058 on Build GRCh38
rs374734939 Chr.3: 138163823-138163823 on Build GRCh38
rs145527166 Chr.3: 138163862-138163862 on Build GRCh38
rs199674725 Chr.3: 138163749-138163749 on Build GRCh38
rs73867020 Chr.3: 138164002-138164002 on Build GRCh38
rs769776897 Chr.3: 138163958-138163958 on Build GRCh38
rs761162405 Chr.3: 138163789-138163789 on Build GRCh38
rs771200581 Chr.3: 138163899-138163899 on Build GRCh38
rs773404197 Chr.3: 138163764-138163764 on Build GRCh38

View Full Product Details