Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138497761-138498264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP70
Entrez Gene ID
80321
Gene Name
centrosomal protein 70
Gene Aliases
BITE
UniGene
Hs.531962
Gene Chromosome Location
Chr.3: 138494339-138594383 on Build GRCh38
Transcripts
NM_001288964.1 NM_001288965.1 NM_001288967.1 NM_001320598.1 NM_001320599.1 NM_024491.3 XM_005247805.3 XM_017007270.1 XM_017007271.1 XM_017007272.1 XM_017007273.1 XM_017007274.1 XM_017007275.1 XM_017007276.1 XM_017007277.1 XM_017007278.1 XM_017007279.1 XM_017007280.1 XM_017007281.1 XM_017007282.1 XM_017007283.1 XM_017007284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2156410 c.1699C>T p.Q567* Chr.3: 138498064-138498064 on Build GRCh38
479492 c.1720C>T p.L574F Chr.3: 138498043-138498043 on Build GRCh38
5590176 c.1728C>T p.I576I Chr.3: 138498035-138498035 on Build GRCh38
1039035 c.1728C>A p.I576I Chr.3: 138498035-138498035 on Build GRCh38
4615152 c.1688_1689insT p.A565fs*7 Chr.3: 138498074-138498075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763234775 Chr.3: 138498028-138498028 on Build GRCh38
rs908469117 Chr.3: 138497998-138497998 on Build GRCh38
rs200106960 Chr.3: 138498040-138498040 on Build GRCh38
rs896610473 Chr.3: 138498165-138498165 on Build GRCh38
rs781183839 Chr.3: 138498110-138498110 on Build GRCh38
rs768563992 Chr.3: 138497920-138497921 on Build GRCh38
rs186485050 Chr.3: 138497830-138497830 on Build GRCh38
rs34872441 Chr.3: 138497961-138497962 on Build GRCh38
rs377145448 Chr.3: 138498136-138498136 on Build GRCh38
rs770406461 Chr.3: 138498091-138498091 on Build GRCh38
rs891172835 Chr.3: 138498044-138498044 on Build GRCh38
rs553635613 Chr.3: 138498036-138498036 on Build GRCh38
rs748381537 Chr.3: 138498005-138498005 on Build GRCh38
rs749675278 Chr.3: 138498139-138498139 on Build GRCh38
rs932618410 Chr.3: 138497968-138497968 on Build GRCh38
rs760065622 Chr.3: 138498042-138498042 on Build GRCh38
rs7625183 Chr.3: 138497871-138497871 on Build GRCh38
rs112140750 Chr.3: 138498143-138498143 on Build GRCh38
rs770041827 Chr.3: 138498011-138498011 on Build GRCh38
rs1009748093 Chr.3: 138498212-138498212 on Build GRCh38
rs1038995929 Chr.3: 138497898-138497898 on Build GRCh38
rs139687895 Chr.3: 138497972-138497972 on Build GRCh38
rs771457397 Chr.3: 138498031-138498031 on Build GRCh38
rs529250069 Chr.3: 138498228-138498228 on Build GRCh38
rs151170111 Chr.3: 138498126-138498126 on Build GRCh38
rs774694406 Chr.3: 138498161-138498161 on Build GRCh38
rs779976251 Chr.3: 138498088-138498088 on Build GRCh38
rs201584232 Chr.3: 138497923-138497927 on Build GRCh38
rs148806908 Chr.3: 138498074-138498074 on Build GRCh38
rs372243600 Chr.3: 138498077-138498077 on Build GRCh38
rs199819372 Chr.3: 138498105-138498105 on Build GRCh38
rs933840609 Chr.3: 138497930-138497930 on Build GRCh38
rs1021212026 Chr.3: 138497810-138497810 on Build GRCh38
rs200041935 Chr.3: 138497928-138497928 on Build GRCh38
rs1056632806 Chr.3: 138498146-138498146 on Build GRCh38
rs763578313 Chr.3: 138498049-138498049 on Build GRCh38
rs116852071 Chr.3: 138497890-138497890 on Build GRCh38
rs142393409 Chr.3: 138498085-138498085 on Build GRCh38
rs533835670 Chr.3: 138497925-138497925 on Build GRCh38
rs964792283 Chr.3: 138497873-138497873 on Build GRCh38
rs968422443 Chr.3: 138497828-138497828 on Build GRCh38
rs758222104 Chr.3: 138498086-138498086 on Build GRCh38
rs576119443 Chr.3: 138497831-138497831 on Build GRCh38
rs773577268 Chr.3: 138498017-138498017 on Build GRCh38
rs143317911 Chr.3: 138497979-138497979 on Build GRCh38
rs368843375 Chr.3: 138497999-138497999 on Build GRCh38

View Full Product Details