Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138500566-138501116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP70
Entrez Gene ID
80321
Gene Name
centrosomal protein 70
Gene Aliases
BITE
UniGene
Hs.531962
Gene Chromosome Location
Chr.3: 138494339-138594383 on Build GRCh38
Transcripts
NM_001288964.1 NM_001288965.1 NM_001288966.1 NM_001288967.1 NM_001320598.1 NM_001320599.1 NM_024491.3 XM_005247805.3 XM_017007270.1 XM_017007271.1 XM_017007272.1 XM_017007273.1 XM_017007274.1 XM_017007275.1 XM_017007276.1 XM_017007277.1 XM_017007278.1 XM_017007279.1 XM_017007280.1 XM_017007281.1 XM_017007282.1 XM_017007283.1 XM_017007284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1616977 c.1245A>G p.T415T Chr.3: 138500858-138500858 on Build GRCh38
5346363 c.1278G>C p.L426F Chr.3: 138500825-138500825 on Build GRCh38
3915017 c.1264C>T p.P422S Chr.3: 138500839-138500839 on Build GRCh38
3587987 c.1260G>T p.L420L Chr.3: 138500843-138500843 on Build GRCh38
3587986 c.1365A>G p.E455E Chr.3: 138500738-138500738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs797020103 Chr.3: 138500594-138500595 on Build GRCh38
rs781674737 Chr.3: 138500692-138500692 on Build GRCh38
rs1009104883 Chr.3: 138500996-138500996 on Build GRCh38
rs774706923 Chr.3: 138500768-138500768 on Build GRCh38
rs371594314 Chr.3: 138500618-138500618 on Build GRCh38
rs769804831 Chr.3: 138500833-138500833 on Build GRCh38
rs34053101 Chr.3: 138500832-138500832 on Build GRCh38
rs113264564 Chr.3: 138500725-138500725 on Build GRCh38
rs576698082 Chr.3: 138500827-138500827 on Build GRCh38
rs746727577 Chr.3: 138500819-138500819 on Build GRCh38
rs962288722 Chr.3: 138500660-138500660 on Build GRCh38
rs879359645 Chr.3: 138500965-138500966 on Build GRCh38
rs917793279 Chr.3: 138501019-138501019 on Build GRCh38
rs1046226378 Chr.3: 138500780-138500780 on Build GRCh38
rs143913326 Chr.3: 138500807-138500807 on Build GRCh38
rs920858465 Chr.3: 138500687-138500687 on Build GRCh38
rs567096762 Chr.3: 138500699-138500699 on Build GRCh38
rs967755218 Chr.3: 138500973-138500977 on Build GRCh38
rs549748042 Chr.3: 138501063-138501063 on Build GRCh38
rs201668440 Chr.3: 138500912-138500912 on Build GRCh38
rs979181937 Chr.3: 138500975-138500977 on Build GRCh38
rs756649782 Chr.3: 138500793-138500793 on Build GRCh38
rs1027889307 Chr.3: 138501040-138501040 on Build GRCh38
rs556813754 Chr.3: 138500822-138500822 on Build GRCh38
rs937819123 Chr.3: 138501004-138501005 on Build GRCh38
rs971425024 Chr.3: 138501083-138501083 on Build GRCh38
rs867427641 Chr.3: 138500825-138500825 on Build GRCh38
rs893743552 Chr.3: 138500886-138500886 on Build GRCh38
rs768959881 Chr.3: 138500646-138500646 on Build GRCh38
rs1015543451 Chr.3: 138500653-138500653 on Build GRCh38
rs761953493 Chr.3: 138500785-138500785 on Build GRCh38
rs779631741 Chr.3: 138500717-138500717 on Build GRCh38
rs7634745 Chr.3: 138501081-138501081 on Build GRCh38
rs749229325 Chr.3: 138500848-138500848 on Build GRCh38
rs767791125 Chr.3: 138500874-138500874 on Build GRCh38
rs759896369 Chr.3: 138500870-138500870 on Build GRCh38
rs950212159 Chr.3: 138500779-138500779 on Build GRCh38
rs754340343 Chr.3: 138500931-138500931 on Build GRCh38
rs769555510 Chr.3: 138500741-138500741 on Build GRCh38
rs897909110 Chr.3: 138500801-138500801 on Build GRCh38
rs774569343 Chr.3: 138500857-138500857 on Build GRCh38
rs779505190 Chr.3: 138500816-138500816 on Build GRCh38
rs1050545457 Chr.3: 138500947-138500947 on Build GRCh38
rs988202300 Chr.3: 138500948-138500948 on Build GRCh38
rs529893769 Chr.3: 138501050-138501050 on Build GRCh38
rs200412030 Chr.3: 138500772-138500772 on Build GRCh38
rs538617017 Chr.3: 138500676-138500676 on Build GRCh38
rs1036344944 Chr.3: 138501028-138501028 on Build GRCh38
rs771176879 Chr.3: 138500850-138500850 on Build GRCh38
rs890206047 Chr.3: 138500989-138500989 on Build GRCh38
rs987645545 Chr.3: 138500629-138500629 on Build GRCh38
rs879472780 Chr.3: 138500956-138500956 on Build GRCh38
rs939988057 Chr.3: 138501024-138501024 on Build GRCh38
rs527701150 Chr.3: 138500981-138500981 on Build GRCh38
rs775954408 Chr.3: 138500881-138500881 on Build GRCh38
rs376931648 Chr.3: 138500790-138500790 on Build GRCh38
rs541494046 Chr.3: 138501011-138501011 on Build GRCh38
rs995020291 Chr.3: 138501037-138501037 on Build GRCh38
rs1049148579 Chr.3: 138501049-138501049 on Build GRCh38
rs759802823 Chr.3: 138500775-138500775 on Build GRCh38
rs764495879 Chr.3: 138500930-138500930 on Build GRCh38
rs762905167 Chr.3: 138500770-138500770 on Build GRCh38
rs866203232 Chr.3: 138501026-138501026 on Build GRCh38
rs774765506 Chr.3: 138500773-138500773 on Build GRCh38
rs150622817 Chr.3: 138500596-138500597 on Build GRCh38
rs148327991 Chr.3: 138501065-138501065 on Build GRCh38
rs138124965 Chr.3: 138500776-138500776 on Build GRCh38
rs370054530 Chr.3: 138500766-138500766 on Build GRCh38
rs777738211 Chr.3: 138500903-138500903 on Build GRCh38
rs138506086 Chr.3: 138500715-138500715 on Build GRCh38
rs767983476 Chr.3: 138500784-138500784 on Build GRCh38
rs771787285 Chr.3: 138500716-138500716 on Build GRCh38
rs772977093 Chr.3: 138500834-138500834 on Build GRCh38
rs756574638 Chr.3: 138500702-138500702 on Build GRCh38
rs377207926 Chr.3: 138500734-138500734 on Build GRCh38
rs534389214 Chr.3: 138500729-138500729 on Build GRCh38
rs764467823 Chr.3: 138500803-138500803 on Build GRCh38
rs1028719739 Chr.3: 138500617-138500617 on Build GRCh38
rs778543390 Chr.3: 138500708-138500708 on Build GRCh38
rs757893693 Chr.3: 138500813-138500813 on Build GRCh38
rs561226496 Chr.3: 138501032-138501032 on Build GRCh38
rs11920691 Chr.3: 138500958-138500958 on Build GRCh38
rs746639683 Chr.3: 138500718-138500718 on Build GRCh38
rs768061817 Chr.3: 138500784-138500784 on Build GRCh38
rs202216015 Chr.3: 138501043-138501043 on Build GRCh38
rs111605907 Chr.3: 138500595-138500596 on Build GRCh38
rs755347626 Chr.3: 138500686-138500686 on Build GRCh38
rs750459982 Chr.3: 138500593-138500593 on Build GRCh38
rs11920599 Chr.3: 138500675-138500675 on Build GRCh38
rs753111985 Chr.3: 138500791-138500792 on Build GRCh38

View Full Product Details