Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138508219-138508633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP70
Entrez Gene ID
80321
Gene Name
centrosomal protein 70
Gene Aliases
BITE
UniGene
Hs.531962
Gene Chromosome Location
Chr.3: 138494339-138594383 on Build GRCh38
Transcripts
NM_001288964.1 NM_001288965.1 NM_001288966.1 NM_001288967.1 NM_001320598.1 NM_001320599.1 NM_024491.3 XM_005247805.3 XM_011513209.2 XM_017007270.1 XM_017007271.1 XM_017007272.1 XM_017007273.1 XM_017007274.1 XM_017007275.1 XM_017007276.1 XM_017007277.1 XM_017007278.1 XM_017007279.1 XM_017007280.1 XM_017007281.1 XM_017007282.1 XM_017007283.1 XM_017007284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1327843 c.998A>C p.E333A Chr.3: 138508491-138508491 on Build GRCh38
6627149 c.1046T>A p.F349Y Chr.3: 138508443-138508443 on Build GRCh38
6305743 c.1005C>A p.S335R Chr.3: 138508484-138508484 on Build GRCh38
6302059 c.1043A>G p.Y348C Chr.3: 138508446-138508446 on Build GRCh38
6234144 c.983C>A p.T328K Chr.3: 138508506-138508506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755137638 Chr.3: 138508530-138508530 on Build GRCh38
rs973400966 Chr.3: 138508528-138508528 on Build GRCh38
rs78146857 Chr.3: 138508347-138508347 on Build GRCh38
rs1009228755 Chr.3: 138508378-138508378 on Build GRCh38
rs766295713 Chr.3: 138508515-138508515 on Build GRCh38
rs768788123 Chr.3: 138508438-138508438 on Build GRCh38
rs776463787 Chr.3: 138508496-138508496 on Build GRCh38
rs374098559 Chr.3: 138508562-138508562 on Build GRCh38
rs757165696 Chr.3: 138508405-138508405 on Build GRCh38
rs146347282 Chr.3: 138508503-138508503 on Build GRCh38
rs952092203 Chr.3: 138508601-138508601 on Build GRCh38
rs772159908 Chr.3: 138508556-138508556 on Build GRCh38
rs761550671 Chr.3: 138508500-138508500 on Build GRCh38
rs930745027 Chr.3: 138508263-138508263 on Build GRCh38
rs373053659 Chr.3: 138508478-138508478 on Build GRCh38
rs781430562 Chr.3: 138508442-138508442 on Build GRCh38
rs780007932 Chr.3: 138508414-138508414 on Build GRCh38
rs756319218 Chr.3: 138508575-138508575 on Build GRCh38
rs934996029 Chr.3: 138508300-138508300 on Build GRCh38
rs576459781 Chr.3: 138508299-138508299 on Build GRCh38
rs190147976 Chr.3: 138508538-138508538 on Build GRCh38
rs753707710 Chr.3: 138508397-138508397 on Build GRCh38
rs750182600 Chr.3: 138508508-138508508 on Build GRCh38
rs751609893 Chr.3: 138508523-138508523 on Build GRCh38
rs745332881 Chr.3: 138508523-138508523 on Build GRCh38
rs1046290836 Chr.3: 138508262-138508262 on Build GRCh38
rs919345418 Chr.3: 138508245-138508245 on Build GRCh38
rs149853784 Chr.3: 138508454-138508454 on Build GRCh38
rs748352321 Chr.3: 138508445-138508445 on Build GRCh38
rs919279375 Chr.3: 138508573-138508573 on Build GRCh38
rs961629273 Chr.3: 138508542-138508542 on Build GRCh38
rs903585726 Chr.3: 138508448-138508448 on Build GRCh38
rs1015169322 Chr.3: 138508485-138508485 on Build GRCh38
rs1033444481 Chr.3: 138508469-138508469 on Build GRCh38
rs1053963575 Chr.3: 138508301-138508301 on Build GRCh38
rs760260622 Chr.3: 138508390-138508390 on Build GRCh38
rs902216598 Chr.3: 138508269-138508269 on Build GRCh38
rs771413358 Chr.3: 138508462-138508462 on Build GRCh38
rs1049215472 Chr.3: 138508282-138508282 on Build GRCh38
rs774867298 Chr.3: 138508479-138508479 on Build GRCh38
rs749589519 Chr.3: 138508594-138508594 on Build GRCh38
rs1009185506 Chr.3: 138508466-138508466 on Build GRCh38
rs773565106 Chr.3: 138508458-138508458 on Build GRCh38
rs756354880 Chr.3: 138508456-138508456 on Build GRCh38
rs1029126404 Chr.3: 138508452-138508452 on Build GRCh38
rs781221552 Chr.3: 138508533-138508533 on Build GRCh38
rs185648153 Chr.3: 138508490-138508490 on Build GRCh38
rs1028163796 Chr.3: 138508553-138508553 on Build GRCh38
rs542557811 Chr.3: 138508266-138508266 on Build GRCh38
rs778125293 Chr.3: 138508584-138508584 on Build GRCh38
rs996756071 Chr.3: 138508435-138508435 on Build GRCh38
rs902111837 Chr.3: 138508376-138508376 on Build GRCh38
rs760192472 Chr.3: 138508484-138508484 on Build GRCh38
rs778459241 Chr.3: 138508517-138508518 on Build GRCh38
rs747040136 Chr.3: 138508420-138508420 on Build GRCh38
rs748438259 Chr.3: 138508574-138508574 on Build GRCh38
rs572473343 Chr.3: 138508460-138508460 on Build GRCh38
rs763735627 Chr.3: 138508394-138508394 on Build GRCh38
rs556908028 Chr.3: 138508294-138508294 on Build GRCh38
rs778817798 Chr.3: 138508406-138508406 on Build GRCh38
rs758270069 Chr.3: 138508514-138508514 on Build GRCh38
rs201271950 Chr.3: 138508408-138508408 on Build GRCh38

View Full Product Details