Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138525484-138525883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP70
Entrez Gene ID
80321
Gene Name
centrosomal protein 70
Gene Aliases
BITE
UniGene
Hs.531962
Gene Chromosome Location
Chr.3: 138494339-138594383 on Build GRCh38
Transcripts
NM_001288964.1 NM_001288965.1 NM_001288966.1 NM_001288967.1 NM_001320598.1 NM_001320599.1 NM_024491.3 XM_005247805.3 XM_011513209.2 XM_017007270.1 XM_017007271.1 XM_017007272.1 XM_017007273.1 XM_017007274.1 XM_017007275.1 XM_017007276.1 XM_017007277.1 XM_017007278.1 XM_017007279.1 XM_017007280.1 XM_017007281.1 XM_017007282.1 XM_017007283.1 XM_017007284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4157313 c.876G>A p.T292T Chr.3: 138525558-138525558 on Build GRCh38
4671701 c.890G>C p.R297T Chr.3: 138525544-138525544 on Build GRCh38
5469023 c.870-6C>T p.? Chr.3: 138525570-138525570 on Build GRCh38
6627147 c.894_895delTT p.Y299fs*1 Chr.3: 138525539-138525540 on Build GRCh38
4562036 c.909G>A p.V303V Chr.3: 138525525-138525525 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779140224 Chr.3: 138525522-138525522 on Build GRCh38
rs970160118 Chr.3: 138525749-138525749 on Build GRCh38
rs936058382 Chr.3: 138525609-138525609 on Build GRCh38
rs943138468 Chr.3: 138525839-138525839 on Build GRCh38
rs144266926 Chr.3: 138525770-138525770 on Build GRCh38
rs767377785 Chr.3: 138525591-138525591 on Build GRCh38
rs563089671 Chr.3: 138525771-138525771 on Build GRCh38
rs182607302 Chr.3: 138525763-138525763 on Build GRCh38
rs1022965064 Chr.3: 138525748-138525748 on Build GRCh38
rs144595685 Chr.3: 138525787-138525787 on Build GRCh38
rs200302654 Chr.3: 138525516-138525516 on Build GRCh38
rs777932606 Chr.3: 138525513-138525513 on Build GRCh38
rs544033926 Chr.3: 138525635-138525635 on Build GRCh38
rs1025957188 Chr.3: 138525739-138525739 on Build GRCh38
rs773044019 Chr.3: 138525559-138525559 on Build GRCh38
rs770830737 Chr.3: 138525581-138525581 on Build GRCh38
rs887468321 Chr.3: 138525745-138525745 on Build GRCh38
rs768567900 Chr.3: 138525608-138525608 on Build GRCh38
rs200516983 Chr.3: 138525553-138525553 on Build GRCh38
rs544891977 Chr.3: 138525626-138525626 on Build GRCh38
rs887786489 Chr.3: 138525697-138525697 on Build GRCh38
rs377620457 Chr.3: 138525520-138525520 on Build GRCh38
rs1052760344 Chr.3: 138525539-138525539 on Build GRCh38
rs199680079 Chr.3: 138525564-138525564 on Build GRCh38
rs140838830 Chr.3: 138525784-138525784 on Build GRCh38
rs910198115 Chr.3: 138525828-138525828 on Build GRCh38
rs190023105 Chr.3: 138525694-138525694 on Build GRCh38
rs993767412 Chr.3: 138525698-138525698 on Build GRCh38
rs979093722 Chr.3: 138525822-138525822 on Build GRCh38
rs993776906 Chr.3: 138525650-138525650 on Build GRCh38
rs116467036 Chr.3: 138525842-138525842 on Build GRCh38
rs571694056 Chr.3: 138525670-138525670 on Build GRCh38
rs752807296 Chr.3: 138525598-138525598 on Build GRCh38
rs759534621 Chr.3: 138525586-138525586 on Build GRCh38
rs774005804 Chr.3: 138525582-138525582 on Build GRCh38
rs990638129 Chr.3: 138525830-138525830 on Build GRCh38
rs964892399 Chr.3: 138525761-138525761 on Build GRCh38
rs772399583 Chr.3: 138525534-138525534 on Build GRCh38
rs977995023 Chr.3: 138525772-138525772 on Build GRCh38
rs943107450 Chr.3: 138525623-138525623 on Build GRCh38
rs560746308 Chr.3: 138525699-138525699 on Build GRCh38
rs1043459338 Chr.3: 138525585-138525585 on Build GRCh38
rs9878436 Chr.3: 138525558-138525558 on Build GRCh38
rs901985207 Chr.3: 138525636-138525636 on Build GRCh38
rs149616624 Chr.3: 138525599-138525599 on Build GRCh38
rs373284096 Chr.3: 138525532-138525532 on Build GRCh38
rs1006674397 Chr.3: 138525705-138525705 on Build GRCh38
rs1017391864 Chr.3: 138525759-138525759 on Build GRCh38
rs988085532 Chr.3: 138525782-138525782 on Build GRCh38
rs948954566 Chr.3: 138525658-138525658 on Build GRCh38
rs747619395 Chr.3: 138525557-138525557 on Build GRCh38

View Full Product Details