Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110873014-110873512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD53
Entrez Gene ID
963
Gene Name
CD53 molecule
Gene Aliases
MOX44, TSPAN25
UniGene
Hs.443057
Gene Chromosome Location
Chr.1: 110871199-110899936 on Build GRCh38
Transcripts
NM_000560.3 NM_001040033.1 NM_001320638.1

SNP Detail

SNP ID Chromosome Location
rs770823387 Chr.1: 110873484-110873484 on Build GRCh38
rs1002708485 Chr.1: 110873394-110873394 on Build GRCh38
rs918844554 Chr.1: 110873199-110873199 on Build GRCh38
rs897468626 Chr.1: 110873444-110873444 on Build GRCh38
rs377728213 Chr.1: 110873058-110873058 on Build GRCh38
rs891729230 Chr.1: 110873458-110873458 on Build GRCh38
rs892891529 Chr.1: 110873156-110873156 on Build GRCh38
rs563785003 Chr.1: 110873389-110873389 on Build GRCh38
rs529525604 Chr.1: 110873039-110873039 on Build GRCh38
rs769640696 Chr.1: 110873299-110873299 on Build GRCh38
rs865778991 Chr.1: 110873092-110873092 on Build GRCh38
rs1002698618 Chr.1: 110873080-110873080 on Build GRCh38
rs944245018 Chr.1: 110873175-110873175 on Build GRCh38
rs143697009 Chr.1: 110873103-110873103 on Build GRCh38
rs35289737 Chr.1: 110873211-110873212 on Build GRCh38
rs927534229 Chr.1: 110873091-110873091 on Build GRCh38
rs927553324 Chr.1: 110873379-110873379 on Build GRCh38
rs937005364 Chr.1: 110873117-110873117 on Build GRCh38
rs1494324 Chr.1: 110873219-110873219 on Build GRCh38
rs777527647 Chr.1: 110873386-110873386 on Build GRCh38
rs779128580 Chr.1: 110873078-110873080 on Build GRCh38
rs1022015752 Chr.1: 110873108-110873108 on Build GRCh38
rs566093860 Chr.1: 110873189-110873189 on Build GRCh38
rs571705022 Chr.1: 110873332-110873332 on Build GRCh38
rs537421220 Chr.1: 110873406-110873406 on Build GRCh38
rs981567913 Chr.1: 110873375-110873375 on Build GRCh38
rs1022692664 Chr.1: 110873057-110873057 on Build GRCh38
rs983520080 Chr.1: 110873045-110873045 on Build GRCh38
rs1011936058 Chr.1: 110873461-110873461 on Build GRCh38
rs749271005 Chr.1: 110873436-110873436 on Build GRCh38
rs952122484 Chr.1: 110873263-110873263 on Build GRCh38
rs545474338 Chr.1: 110873236-110873236 on Build GRCh38
rs1031599099 Chr.1: 110873087-110873087 on Build GRCh38
rs945829401 Chr.1: 110873169-110873169 on Build GRCh38
rs138436326 Chr.1: 110873449-110873449 on Build GRCh38
rs551896611 Chr.1: 110873260-110873260 on Build GRCh38
rs1041430684 Chr.1: 110873173-110873173 on Build GRCh38
rs899710837 Chr.1: 110873253-110873253 on Build GRCh38
rs907073127 Chr.1: 110873059-110873059 on Build GRCh38
rs990795374 Chr.1: 110873425-110873425 on Build GRCh38
rs931535204 Chr.1: 110873208-110873208 on Build GRCh38
rs985973025 Chr.1: 110873140-110873140 on Build GRCh38
rs914267511 Chr.1: 110873481-110873481 on Build GRCh38
rs553646071 Chr.1: 110873041-110873042 on Build GRCh38
rs950192875 Chr.1: 110873050-110873050 on Build GRCh38
rs906916222 Chr.1: 110873348-110873348 on Build GRCh38
rs971563305 Chr.1: 110873358-110873358 on Build GRCh38
rs955901788 Chr.1: 110873097-110873097 on Build GRCh38
rs758584438 Chr.1: 110873049-110873049 on Build GRCh38
rs878993498 Chr.1: 110873256-110873256 on Build GRCh38
rs1054103794 Chr.1: 110873129-110873129 on Build GRCh38

View Full Product Details