Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111182721-111183235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEPT1
Entrez Gene ID
10390
Gene Name
choline/ethanolamine phosphotransferase 1
Gene Aliases
-
UniGene
Hs.363572
Gene Chromosome Location
Chr.1: 111139436-111185102 on Build GRCh38
Transcripts
NM_001007794.1 NM_006090.3 XM_006710285.3 XM_011540496.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
893760 c.850A>G p.T284A Chr.1: 111182802-111182802 on Build GRCh38
3471156 c.865C>T p.P289S Chr.1: 111182817-111182817 on Build GRCh38
2119085 c.847-2A>G p.? Chr.1: 111182797-111182797 on Build GRCh38
2119086 c.847-1G>A p.? Chr.1: 111182798-111182798 on Build GRCh38
5596488 c.874C>T p.H292Y Chr.1: 111182826-111182826 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs369369892 Chr.1: 111182936-111182936 on Build GRCh38
rs543108380 Chr.1: 111183195-111183195 on Build GRCh38
rs923054307 Chr.1: 111182962-111182962 on Build GRCh38
rs921731365 Chr.1: 111182779-111182779 on Build GRCh38
rs780070304 Chr.1: 111182754-111182754 on Build GRCh38
rs1050480085 Chr.1: 111183034-111183034 on Build GRCh38
rs1045744622 Chr.1: 111183107-111183107 on Build GRCh38
rs770285994 Chr.1: 111182805-111182805 on Build GRCh38
rs780816910 Chr.1: 111183135-111183135 on Build GRCh38
rs920401433 Chr.1: 111183043-111183043 on Build GRCh38
rs775914989 Chr.1: 111182837-111182837 on Build GRCh38
rs777335868 Chr.1: 111182980-111182980 on Build GRCh38
rs760489127 Chr.1: 111182910-111182910 on Build GRCh38
rs199534942 Chr.1: 111182967-111182967 on Build GRCh38
rs566676716 Chr.1: 111182781-111182781 on Build GRCh38
rs756189253 Chr.1: 111182751-111182751 on Build GRCh38
rs562669681 Chr.1: 111183194-111183194 on Build GRCh38
rs746240964 Chr.1: 111182983-111182983 on Build GRCh38
rs777035244 Chr.1: 111182793-111182793 on Build GRCh38
rs747873117 Chr.1: 111182782-111182782 on Build GRCh38
rs780370490 Chr.1: 111183006-111183006 on Build GRCh38
rs546438290 Chr.1: 111182780-111182780 on Build GRCh38
rs770108074 Chr.1: 111182988-111182988 on Build GRCh38
rs768867524 Chr.1: 111183048-111183048 on Build GRCh38
rs766442401 Chr.1: 111182921-111182921 on Build GRCh38
rs771240192 Chr.1: 111182777-111182781 on Build GRCh38
rs574007011 Chr.1: 111183184-111183184 on Build GRCh38
rs750250181 Chr.1: 111182893-111182893 on Build GRCh38
rs375077360 Chr.1: 111183067-111183067 on Build GRCh38
rs774275456 Chr.1: 111182845-111182845 on Build GRCh38
rs1006524643 Chr.1: 111183049-111183049 on Build GRCh38
rs993316594 Chr.1: 111183079-111183079 on Build GRCh38
rs1025682250 Chr.1: 111183128-111183128 on Build GRCh38
rs778532958 Chr.1: 111182946-111182946 on Build GRCh38
rs575637034 Chr.1: 111183076-111183076 on Build GRCh38
rs148455686 Chr.1: 111183183-111183183 on Build GRCh38
rs1037635799 Chr.1: 111183053-111183053 on Build GRCh38
rs868091791 Chr.1: 111183116-111183116 on Build GRCh38
rs758133620 Chr.1: 111182972-111182972 on Build GRCh38
rs538907688 Chr.1: 111182843-111182843 on Build GRCh38
rs932964462 Chr.1: 111183020-111183020 on Build GRCh38
rs771600732 Chr.1: 111182784-111182784 on Build GRCh38
rs372073879 Chr.1: 111182942-111182942 on Build GRCh38
rs987172321 Chr.1: 111182846-111182846 on Build GRCh38
rs894588389 Chr.1: 111183101-111183101 on Build GRCh38
rs754496211 Chr.1: 111182883-111182883 on Build GRCh38
rs113123860 Chr.1: 111183046-111183046 on Build GRCh38
rs767630851 Chr.1: 111182876-111182876 on Build GRCh38
rs750231799 Chr.1: 111182750-111182750 on Build GRCh38
rs1024017634 Chr.1: 111183174-111183174 on Build GRCh38
rs751051585 Chr.1: 111182794-111182794 on Build GRCh38
rs1014207110 Chr.1: 111183104-111183105 on Build GRCh38
rs762156116 Chr.1: 111182856-111182856 on Build GRCh38
rs748821618 Chr.1: 111182764-111182764 on Build GRCh38
rs144547551 Chr.1: 111183161-111183161 on Build GRCh38

View Full Product Details