Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111191849-111192345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND2D
Entrez Gene ID
79961
Gene Name
DENN domain containing 2D
Gene Aliases
-
UniGene
Hs.557850
Gene Chromosome Location
Chr.1: 111185969-111204791 on Build GRCh38
Transcripts
NM_001271833.1 NM_024901.4 XM_006710921.2 XM_006710922.1 XM_006710923.1 XM_006710924.1 XM_011542187.1 XM_011542189.1 XM_011542190.2 XM_017002386.1 XM_017002387.1 XM_017002388.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1667804 c.917T>C p.M306T Chr.1: 111192195-111192195 on Build GRCh38
6318521 c.855C>A p.H285Q Chr.1: 111192257-111192257 on Build GRCh38
6642336 c.870T>C p.V290V Chr.1: 111192242-111192242 on Build GRCh38
6642333 c.852G>A p.A284A Chr.1: 111192260-111192260 on Build GRCh38
6238458 c.919G>C p.V307L Chr.1: 111192193-111192193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770670919 Chr.1: 111192257-111192257 on Build GRCh38
rs755448925 Chr.1: 111192320-111192320 on Build GRCh38
rs769717736 Chr.1: 111192288-111192288 on Build GRCh38
rs746851448 Chr.1: 111192246-111192246 on Build GRCh38
rs772438883 Chr.1: 111192233-111192233 on Build GRCh38
rs964703048 Chr.1: 111192013-111192013 on Build GRCh38
rs376513608 Chr.1: 111192294-111192294 on Build GRCh38
rs1027947414 Chr.1: 111192071-111192071 on Build GRCh38
rs773147957 Chr.1: 111192094-111192094 on Build GRCh38
rs758109616 Chr.1: 111192192-111192192 on Build GRCh38
rs749813179 Chr.1: 111192217-111192217 on Build GRCh38
rs200122035 Chr.1: 111192148-111192148 on Build GRCh38
rs751212764 Chr.1: 111192306-111192306 on Build GRCh38
rs202089781 Chr.1: 111192108-111192108 on Build GRCh38
rs367758355 Chr.1: 111192201-111192201 on Build GRCh38
rs140593688 Chr.1: 111192230-111192230 on Build GRCh38
rs72979803 Chr.1: 111192000-111192000 on Build GRCh38
rs368968192 Chr.1: 111192286-111192286 on Build GRCh38
rs761504696 Chr.1: 111192311-111192311 on Build GRCh38
rs750810960 Chr.1: 111192199-111192199 on Build GRCh38
rs151121538 Chr.1: 111192009-111192009 on Build GRCh38
rs555965792 Chr.1: 111192005-111192005 on Build GRCh38
rs779202042 Chr.1: 111192220-111192220 on Build GRCh38
rs753560422 Chr.1: 111192097-111192097 on Build GRCh38
rs952402704 Chr.1: 111192086-111192086 on Build GRCh38
rs974950107 Chr.1: 111192050-111192050 on Build GRCh38
rs376719159 Chr.1: 111192103-111192103 on Build GRCh38
rs930014875 Chr.1: 111192061-111192061 on Build GRCh38
rs759485695 Chr.1: 111192151-111192151 on Build GRCh38
rs142956255 Chr.1: 111191993-111191993 on Build GRCh38
rs187119617 Chr.1: 111192287-111192287 on Build GRCh38
rs764772273 Chr.1: 111192177-111192177 on Build GRCh38
rs374395836 Chr.1: 111192090-111192090 on Build GRCh38
rs113730801 Chr.1: 111192235-111192235 on Build GRCh38
rs763666689 Chr.1: 111192195-111192195 on Build GRCh38
rs763768849 Chr.1: 111192305-111192305 on Build GRCh38
rs111378970 Chr.1: 111192143-111192143 on Build GRCh38
rs905376736 Chr.1: 111192105-111192105 on Build GRCh38
rs1019963341 Chr.1: 111192197-111192197 on Build GRCh38
rs147784861 Chr.1: 111192178-111192178 on Build GRCh38
rs138266603 Chr.1: 111192260-111192260 on Build GRCh38
rs748439409 Chr.1: 111192089-111192089 on Build GRCh38
rs267597922 Chr.1: 111192193-111192193 on Build GRCh38
rs1010104638 Chr.1: 111191930-111191930 on Build GRCh38
rs893013619 Chr.1: 111192261-111192261 on Build GRCh38
rs760327963 Chr.1: 111192100-111192100 on Build GRCh38
rs913184937 Chr.1: 111191921-111191921 on Build GRCh38
rs773516777 Chr.1: 111192242-111192242 on Build GRCh38
rs61757211 Chr.1: 111192200-111192200 on Build GRCh38
rs766716172 Chr.1: 111192316-111192316 on Build GRCh38
rs751811163 Chr.1: 111191886-111191886 on Build GRCh38
rs754229487 Chr.1: 111192318-111192318 on Build GRCh38
rs752186052 Chr.1: 111192182-111192182 on Build GRCh38
rs780439490 Chr.1: 111192203-111192203 on Build GRCh38
rs988712979 Chr.1: 111191888-111191888 on Build GRCh38
rs949759884 Chr.1: 111192157-111192157 on Build GRCh38
rs553774664 Chr.1: 111191976-111191976 on Build GRCh38
rs959716871 Chr.1: 111191874-111191874 on Build GRCh38
rs560505714 Chr.1: 111192296-111192296 on Build GRCh38
rs1009943668 Chr.1: 111192196-111192196 on Build GRCh38
rs35742969 Chr.1: 111192267-111192267 on Build GRCh38
rs555153173 Chr.1: 111192218-111192218 on Build GRCh38

View Full Product Details