Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111238616-111239029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHI3L2
Entrez Gene ID
1117
Gene Name
chitinase 3 like 2
Gene Aliases
CHIL2, YKL-39, YKL39
UniGene
Hs.514840
Gene Chromosome Location
Chr.1: 111227659-111243440 on Build GRCh38
Transcripts
NM_001025197.1 NM_001025199.1 NM_004000.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4562203 c.916G>A p.E306K Chr.1: 111238930-111238930 on Build GRCh38
2119143 c.795C>T p.G265G Chr.1: 111238809-111238809 on Build GRCh38
3862194 c.819C>T p.F273F Chr.1: 111238833-111238833 on Build GRCh38
1332663 c.762T>C p.H254H Chr.1: 111238776-111238776 on Build GRCh38
4576115 c.843C>T p.T281T Chr.1: 111238857-111238857 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779614783 Chr.1: 111238880-111238880 on Build GRCh38
rs962124312 Chr.1: 111238685-111238685 on Build GRCh38
rs751297966 Chr.1: 111238929-111238929 on Build GRCh38
rs902605198 Chr.1: 111238702-111238702 on Build GRCh38
rs757541455 Chr.1: 111238936-111238936 on Build GRCh38
rs757122419 Chr.1: 111238866-111238866 on Build GRCh38
rs745539496 Chr.1: 111238804-111238804 on Build GRCh38
rs746377161 Chr.1: 111238973-111238973 on Build GRCh38
rs773645107 Chr.1: 111238884-111238884 on Build GRCh38
rs981127011 Chr.1: 111238875-111238875 on Build GRCh38
rs946800080 Chr.1: 111238661-111238661 on Build GRCh38
rs137918113 Chr.1: 111238882-111238882 on Build GRCh38
rs971122795 Chr.1: 111238867-111238867 on Build GRCh38
rs778778398 Chr.1: 111238735-111238735 on Build GRCh38
rs769519545 Chr.1: 111238878-111238878 on Build GRCh38
rs760824843 Chr.1: 111238786-111238786 on Build GRCh38
rs199844821 Chr.1: 111238914-111238914 on Build GRCh38
rs753799557 Chr.1: 111238852-111238852 on Build GRCh38
rs887910161 Chr.1: 111238802-111238802 on Build GRCh38
rs41281392 Chr.1: 111238857-111238857 on Build GRCh38
rs575989775 Chr.1: 111238984-111238984 on Build GRCh38
rs765334500 Chr.1: 111238855-111238855 on Build GRCh38
rs140247314 Chr.1: 111238858-111238858 on Build GRCh38
rs369049932 Chr.1: 111238814-111238814 on Build GRCh38
rs535477787 Chr.1: 111238997-111238997 on Build GRCh38
rs768926025 Chr.1: 111238720-111238721 on Build GRCh38
rs564734889 Chr.1: 111238657-111238657 on Build GRCh38
rs145737207 Chr.1: 111238757-111238757 on Build GRCh38
rs374279719 Chr.1: 111238945-111238945 on Build GRCh38
rs755637024 Chr.1: 111238940-111238940 on Build GRCh38
rs769834015 Chr.1: 111238974-111238974 on Build GRCh38
rs371597889 Chr.1: 111238759-111238759 on Build GRCh38
rs202224198 Chr.1: 111238933-111238933 on Build GRCh38
rs370690437 Chr.1: 111238977-111238977 on Build GRCh38
rs770490468 Chr.1: 111238781-111238781 on Build GRCh38
rs932226782 Chr.1: 111238985-111238985 on Build GRCh38
rs748952406 Chr.1: 111238953-111238953 on Build GRCh38
rs146436358 Chr.1: 111238932-111238932 on Build GRCh38
rs761260037 Chr.1: 111238830-111238830 on Build GRCh38
rs1019180555 Chr.1: 111238683-111238683 on Build GRCh38
rs541802513 Chr.1: 111238688-111238688 on Build GRCh38
rs769272863 Chr.1: 111238806-111238806 on Build GRCh38
rs1042437212 Chr.1: 111238696-111238696 on Build GRCh38
rs770164012 Chr.1: 111238878-111238878 on Build GRCh38
rs1027626759 Chr.1: 111238697-111238697 on Build GRCh38
rs780768576 Chr.1: 111238871-111238871 on Build GRCh38
rs779704460 Chr.1: 111238944-111238944 on Build GRCh38
rs143981389 Chr.1: 111238775-111238775 on Build GRCh38
rs750563082 Chr.1: 111238704-111238704 on Build GRCh38
rs759822331 Chr.1: 111238854-111238854 on Build GRCh38
rs201251272 Chr.1: 111238897-111238897 on Build GRCh38
rs777731709 Chr.1: 111238760-111238760 on Build GRCh38
rs199696199 Chr.1: 111238826-111238826 on Build GRCh38
rs747531579 Chr.1: 111238885-111238885 on Build GRCh38
rs754618815 Chr.1: 111238963-111238963 on Build GRCh38
rs900778952 Chr.1: 111238740-111238740 on Build GRCh38
rs778300632 Chr.1: 111238966-111238966 on Build GRCh38
rs528904328 Chr.1: 111238837-111238837 on Build GRCh38
rs781227516 Chr.1: 111238850-111238850 on Build GRCh38
rs745314218 Chr.1: 111238876-111238876 on Build GRCh38
rs766553749 Chr.1: 111238722-111238722 on Build GRCh38
rs763885151 Chr.1: 111238925-111238925 on Build GRCh38
rs748272432 Chr.1: 111238864-111238865 on Build GRCh38
rs752383236 Chr.1: 111238750-111238750 on Build GRCh38
rs762229305 Chr.1: 111238824-111238824 on Build GRCh38
rs373481051 Chr.1: 111238982-111238982 on Build GRCh38
rs771290048 Chr.1: 111238900-111238900 on Build GRCh38
rs145932809 Chr.1: 111238913-111238913 on Build GRCh38
rs754155781 Chr.1: 111238731-111238731 on Build GRCh38
rs747229577 Chr.1: 111238704-111238707 on Build GRCh38
rs777533544 Chr.1: 111238859-111238859 on Build GRCh38
rs747333558 Chr.1: 111238968-111238968 on Build GRCh38
rs760885803 Chr.1: 111238705-111238705 on Build GRCh38
rs377641964 Chr.1: 111238970-111238970 on Build GRCh38
rs374967581 Chr.1: 111238788-111238788 on Build GRCh38
rs996353137 Chr.1: 111238763-111238763 on Build GRCh38
rs751568385 Chr.1: 111238864-111238864 on Build GRCh38
rs202128382 Chr.1: 111238918-111238918 on Build GRCh38
rs775814264 Chr.1: 111238698-111238698 on Build GRCh38
rs200546876 Chr.1: 111238906-111238906 on Build GRCh38

View Full Product Details