Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111346814-111347318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIFO
Entrez Gene ID
128344
Gene Name
primary cilia formation
Gene Aliases
C1orf88, pitchfork
UniGene
Hs.172510
Gene Chromosome Location
Chr.1: 111324663-111353017 on Build GRCh38
Transcripts
NM_001300831.1 NM_181643.5 NR_125338.1 XM_005270472.1 XM_017000322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
893808 c.68T>G p.F23C Chr.1: 111346958-111346958 on Build GRCh38
6230324 c.140C>A p.P47H Chr.1: 111347030-111347030 on Build GRCh38
4020622 c.26C>T p.A9V Chr.1: 111346916-111346916 on Build GRCh38
1332673 c.28G>A p.D10N Chr.1: 111346918-111346918 on Build GRCh38
5719327 c.83C>T p.P28L Chr.1: 111346973-111346973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778858265 Chr.1: 111346882-111346882 on Build GRCh38
rs775864254 Chr.1: 111346894-111346894 on Build GRCh38
rs750658762 Chr.1: 111346927-111346927 on Build GRCh38
rs868436004 Chr.1: 111346993-111346993 on Build GRCh38
rs757899738 Chr.1: 111346987-111346987 on Build GRCh38
rs573634090 Chr.1: 111346985-111346985 on Build GRCh38
rs149275328 Chr.1: 111347252-111347252 on Build GRCh38
rs890282042 Chr.1: 111346952-111346952 on Build GRCh38
rs564605334 Chr.1: 111347153-111347153 on Build GRCh38
rs774777201 Chr.1: 111347047-111347047 on Build GRCh38
rs528857494 Chr.1: 111346837-111346837 on Build GRCh38
rs754993632 Chr.1: 111346877-111346877 on Build GRCh38
rs756373087 Chr.1: 111346938-111346938 on Build GRCh38
rs1032539914 Chr.1: 111346979-111346979 on Build GRCh38
rs1034276755 Chr.1: 111347088-111347088 on Build GRCh38
rs201094761 Chr.1: 111346924-111346924 on Build GRCh38
rs147382775 Chr.1: 111346929-111346929 on Build GRCh38
rs977834554 Chr.1: 111346846-111346846 on Build GRCh38
rs145940267 Chr.1: 111347014-111347014 on Build GRCh38
rs765061275 Chr.1: 111347073-111347073 on Build GRCh38
rs919154622 Chr.1: 111347103-111347103 on Build GRCh38
rs977865635 Chr.1: 111347226-111347226 on Build GRCh38
rs747623013 Chr.1: 111346884-111346884 on Build GRCh38
rs762358429 Chr.1: 111347048-111347048 on Build GRCh38
rs773368483 Chr.1: 111347063-111347063 on Build GRCh38
rs772747744 Chr.1: 111346954-111346954 on Build GRCh38
rs911756371 Chr.1: 111347280-111347280 on Build GRCh38
rs370840646 Chr.1: 111347118-111347118 on Build GRCh38
rs774856442 Chr.1: 111346915-111346915 on Build GRCh38
rs994818267 Chr.1: 111347045-111347045 on Build GRCh38
rs749659129 Chr.1: 111347022-111347022 on Build GRCh38
rs113245363 Chr.1: 111347242-111347242 on Build GRCh38
rs759464585 Chr.1: 111347071-111347071 on Build GRCh38
rs1045129942 Chr.1: 111347294-111347294 on Build GRCh38
rs555387038 Chr.1: 111347032-111347032 on Build GRCh38
rs371933921 Chr.1: 111347001-111347001 on Build GRCh38
rs550157522 Chr.1: 111346834-111346834 on Build GRCh38
rs773339102 Chr.1: 111347288-111347288 on Build GRCh38
rs201767445 Chr.1: 111346890-111346890 on Build GRCh38
rs555938468 Chr.1: 111347216-111347216 on Build GRCh38
rs758395725 Chr.1: 111347085-111347085 on Build GRCh38
rs754044405 Chr.1: 111347069-111347069 on Build GRCh38
rs1013668603 Chr.1: 111347265-111347265 on Build GRCh38
rs1030769783 Chr.1: 111347240-111347240 on Build GRCh38
rs767636933 Chr.1: 111346861-111346861 on Build GRCh38
rs556804855 Chr.1: 111346970-111346970 on Build GRCh38
rs770187933 Chr.1: 111346892-111346892 on Build GRCh38
rs748193266 Chr.1: 111347015-111347015 on Build GRCh38
rs894736320 Chr.1: 111347097-111347098 on Build GRCh38
rs774288415 Chr.1: 111347050-111347053 on Build GRCh38
rs186566800 Chr.1: 111346860-111346860 on Build GRCh38
rs746483931 Chr.1: 111347009-111347009 on Build GRCh38
rs929152147 Chr.1: 111347184-111347184 on Build GRCh38
rs139844072 Chr.1: 111347099-111347099 on Build GRCh38
rs923625158 Chr.1: 111346852-111346852 on Build GRCh38
rs766259465 Chr.1: 111347040-111347040 on Build GRCh38
rs191007120 Chr.1: 111346896-111346896 on Build GRCh38
rs11590153 Chr.1: 111346835-111346835 on Build GRCh38
rs957734227 Chr.1: 111347241-111347241 on Build GRCh38
rs538087365 Chr.1: 111346948-111346948 on Build GRCh38
rs200106671 Chr.1: 111347077-111347077 on Build GRCh38
rs762402190 Chr.1: 111346981-111346984 on Build GRCh38
rs899224724 Chr.1: 111347028-111347028 on Build GRCh38
rs779972072 Chr.1: 111346868-111346868 on Build GRCh38
rs756030755 Chr.1: 111346867-111346867 on Build GRCh38
rs565779259 Chr.1: 111346889-111346889 on Build GRCh38
rs758572502 Chr.1: 111346880-111346880 on Build GRCh38
rs571308247 Chr.1: 111346920-111346920 on Build GRCh38
rs368837713 Chr.1: 111346944-111346944 on Build GRCh38
rs1024517215 Chr.1: 111347144-111347144 on Build GRCh38
rs771609710 Chr.1: 111346885-111346885 on Build GRCh38
rs772083465 Chr.1: 111347061-111347061 on Build GRCh38
rs987773563 Chr.1: 111347275-111347275 on Build GRCh38
rs985792097 Chr.1: 111347024-111347024 on Build GRCh38
rs750651813 Chr.1: 111346863-111346863 on Build GRCh38
rs749273970 Chr.1: 111346875-111346875 on Build GRCh38
rs572491898 Chr.1: 111347064-111347064 on Build GRCh38
rs557846005 Chr.1: 111346902-111346902 on Build GRCh38
rs921848179 Chr.1: 111346900-111346901 on Build GRCh38
rs778785974 Chr.1: 111346977-111346977 on Build GRCh38
rs910214282 Chr.1: 111347033-111347033 on Build GRCh38

View Full Product Details