Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111351856-111352400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIFO
Entrez Gene ID
128344
Gene Name
primary cilia formation
Gene Aliases
C1orf88, pitchfork
UniGene
Hs.172510
Gene Chromosome Location
Chr.1: 111324663-111353017 on Build GRCh38
Transcripts
NM_001300831.1 NM_181643.5 NR_125338.1 XM_005270472.1 XM_017000322.1

SNP Detail

SNP ID Chromosome Location
rs757061148 Chr.1: 111352370-111352370 on Build GRCh38
rs959517024 Chr.1: 111351992-111351992 on Build GRCh38
rs1050791050 Chr.1: 111352278-111352278 on Build GRCh38
rs945633980 Chr.1: 111351926-111351926 on Build GRCh38
rs1056133922 Chr.1: 111352271-111352271 on Build GRCh38
rs140571952 Chr.1: 111352072-111352072 on Build GRCh38
rs1005712989 Chr.1: 111351909-111351909 on Build GRCh38
rs1038304392 Chr.1: 111352085-111352085 on Build GRCh38
rs1002049699 Chr.1: 111352096-111352096 on Build GRCh38
rs543824970 Chr.1: 111352116-111352116 on Build GRCh38
rs879070851 Chr.1: 111352283-111352283 on Build GRCh38
rs888239227 Chr.1: 111351901-111351901 on Build GRCh38
rs954344971 Chr.1: 111352351-111352351 on Build GRCh38
rs956058934 Chr.1: 111352108-111352108 on Build GRCh38
rs1050759830 Chr.1: 111352048-111352048 on Build GRCh38
rs571139313 Chr.1: 111352027-111352027 on Build GRCh38
rs72180703 Chr.1: 111352108-111352109 on Build GRCh38
rs886759373 Chr.1: 111352059-111352059 on Build GRCh38
rs9645352 Chr.1: 111352268-111352268 on Build GRCh38
rs986128119 Chr.1: 111352365-111352365 on Build GRCh38
rs966713520 Chr.1: 111352176-111352179 on Build GRCh38
rs1022156224 Chr.1: 111352300-111352300 on Build GRCh38
rs555448539 Chr.1: 111352185-111352185 on Build GRCh38
rs762229272 Chr.1: 111351960-111351960 on Build GRCh38
rs144535825 Chr.1: 111352221-111352221 on Build GRCh38
rs45595233 Chr.1: 111352304-111352304 on Build GRCh38
rs562102089 Chr.1: 111351929-111351929 on Build GRCh38
rs1020995318 Chr.1: 111352129-111352129 on Build GRCh38
rs979694890 Chr.1: 111351941-111351941 on Build GRCh38
rs186104770 Chr.1: 111352135-111352135 on Build GRCh38
rs79322001 Chr.1: 111352107-111352108 on Build GRCh38
rs968853161 Chr.1: 111352058-111352058 on Build GRCh38
rs976757176 Chr.1: 111352337-111352337 on Build GRCh38
rs1034935655 Chr.1: 111351987-111351987 on Build GRCh38
rs1022544416 Chr.1: 111352040-111352040 on Build GRCh38
rs1038407462 Chr.1: 111352356-111352356 on Build GRCh38
rs527891407 Chr.1: 111351954-111351954 on Build GRCh38
rs895102608 Chr.1: 111352280-111352280 on Build GRCh38
rs191008141 Chr.1: 111352169-111352169 on Build GRCh38
rs549830720 Chr.1: 111352073-111352073 on Build GRCh38
rs1012115513 Chr.1: 111352034-111352034 on Build GRCh38
rs1036799745 Chr.1: 111352228-111352228 on Build GRCh38
rs553271124 Chr.1: 111352306-111352306 on Build GRCh38
rs562234660 Chr.1: 111352369-111352369 on Build GRCh38
rs556181294 Chr.1: 111352299-111352299 on Build GRCh38
rs9645349 Chr.1: 111352100-111352100 on Build GRCh38
rs978803022 Chr.1: 111352099-111352099 on Build GRCh38
rs72693273 Chr.1: 111351975-111351975 on Build GRCh38
rs777158953 Chr.1: 111351948-111351948 on Build GRCh38
rs985686977 Chr.1: 111352209-111352209 on Build GRCh38
rs542541889 Chr.1: 111352202-111352202 on Build GRCh38
rs979816562 Chr.1: 111352188-111352188 on Build GRCh38
rs1030076242 Chr.1: 111352346-111352346 on Build GRCh38
rs898366023 Chr.1: 111352092-111352092 on Build GRCh38
rs9645350 Chr.1: 111352223-111352223 on Build GRCh38
rs1004568186 Chr.1: 111352068-111352068 on Build GRCh38
rs922866978 Chr.1: 111352189-111352189 on Build GRCh38
rs933734092 Chr.1: 111352031-111352031 on Build GRCh38
rs376405430 Chr.1: 111352109-111352110 on Build GRCh38
rs956093032 Chr.1: 111352326-111352326 on Build GRCh38
rs9645351 Chr.1: 111352248-111352248 on Build GRCh38
rs999292478 Chr.1: 111352305-111352305 on Build GRCh38

View Full Product Details