Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111475931-111476429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf162
Entrez Gene ID
128346
Gene Name
chromosome 1 open reading frame 162
Gene Aliases
-
UniGene
Hs.288010
Gene Chromosome Location
Chr.1: 111462694-111478512 on Build GRCh38
Transcripts
NM_001300834.1 NM_001300835.1 NM_174896.3 XM_011540712.2 XM_017000323.1 XM_017000324.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3930265 c.5G>C p.G2A Chr.1: 111476033-111476033 on Build GRCh38
6352221 c.7G>A p.G3S Chr.1: 111476035-111476035 on Build GRCh38
227682 c.31G>A p.D11N Chr.1: 111476059-111476059 on Build GRCh38
5353081 c.35C>T p.T12I Chr.1: 111476063-111476063 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs6703267 Chr.1: 111476035-111476035 on Build GRCh38
rs186900164 Chr.1: 111475973-111475973 on Build GRCh38
rs1027861740 Chr.1: 111476183-111476183 on Build GRCh38
rs113588502 Chr.1: 111475977-111475977 on Build GRCh38
rs1007953888 Chr.1: 111476111-111476111 on Build GRCh38
rs918561615 Chr.1: 111476202-111476202 on Build GRCh38
rs893007303 Chr.1: 111476355-111476355 on Build GRCh38
rs770391879 Chr.1: 111476084-111476084 on Build GRCh38
rs113897453 Chr.1: 111476066-111476066 on Build GRCh38
rs2800886 Chr.1: 111476370-111476370 on Build GRCh38
rs927358965 Chr.1: 111476298-111476298 on Build GRCh38
rs973428983 Chr.1: 111476186-111476186 on Build GRCh38
rs773679046 Chr.1: 111475981-111475981 on Build GRCh38
rs937451179 Chr.1: 111476305-111476305 on Build GRCh38
rs907829124 Chr.1: 111476286-111476286 on Build GRCh38
rs763144157 Chr.1: 111476104-111476104 on Build GRCh38
rs145690743 Chr.1: 111476100-111476100 on Build GRCh38
rs117009640 Chr.1: 111476369-111476369 on Build GRCh38
rs59547347 Chr.1: 111475970-111475970 on Build GRCh38
rs771214689 Chr.1: 111476079-111476079 on Build GRCh38
rs756043503 Chr.1: 111476062-111476062 on Build GRCh38
rs531678845 Chr.1: 111476391-111476391 on Build GRCh38
rs981522518 Chr.1: 111476292-111476292 on Build GRCh38
rs984179532 Chr.1: 111476270-111476270 on Build GRCh38
rs369208859 Chr.1: 111476058-111476058 on Build GRCh38
rs138266865 Chr.1: 111476351-111476352 on Build GRCh38
rs766726759 Chr.1: 111475998-111475998 on Build GRCh38
rs775840395 Chr.1: 111476148-111476148 on Build GRCh38
rs777011114 Chr.1: 111476006-111476006 on Build GRCh38
rs1054214462 Chr.1: 111476322-111476322 on Build GRCh38
rs760970825 Chr.1: 111475997-111475997 on Build GRCh38
rs1054248136 Chr.1: 111476005-111476005 on Build GRCh38
rs552151448 Chr.1: 111476313-111476313 on Build GRCh38
rs751732549 Chr.1: 111476113-111476113 on Build GRCh38
rs950114624 Chr.1: 111476287-111476287 on Build GRCh38
rs1053453844 Chr.1: 111476377-111476377 on Build GRCh38
rs1046237138 Chr.1: 111475971-111475971 on Build GRCh38
rs760184805 Chr.1: 111476080-111476080 on Build GRCh38
rs532323975 Chr.1: 111476150-111476150 on Build GRCh38
rs751752442 Chr.1: 111476039-111476039 on Build GRCh38
rs767285349 Chr.1: 111476117-111476117 on Build GRCh38
rs542041678 Chr.1: 111476023-111476023 on Build GRCh38
rs748777651 Chr.1: 111476068-111476068 on Build GRCh38
rs952822847 Chr.1: 111476253-111476253 on Build GRCh38
rs1018453302 Chr.1: 111476156-111476156 on Build GRCh38
rs755584221 Chr.1: 111476075-111476075 on Build GRCh38
rs907900519 Chr.1: 111475962-111475962 on Build GRCh38
rs138773319 Chr.1: 111476027-111476027 on Build GRCh38
rs974889168 Chr.1: 111476174-111476174 on Build GRCh38
rs762035740 Chr.1: 111476114-111476114 on Build GRCh38
rs963817347 Chr.1: 111476167-111476167 on Build GRCh38
rs879133307 Chr.1: 111476169-111476169 on Build GRCh38
rs190554894 Chr.1: 111476136-111476136 on Build GRCh38
rs376655836 Chr.1: 111476045-111476045 on Build GRCh38
rs879215501 Chr.1: 111476025-111476025 on Build GRCh38
rs768168213 Chr.1: 111476069-111476069 on Build GRCh38
rs760063987 Chr.1: 111476022-111476022 on Build GRCh38
rs1011215784 Chr.1: 111476392-111476392 on Build GRCh38
rs761198047 Chr.1: 111476336-111476336 on Build GRCh38
rs543135752 Chr.1: 111476291-111476291 on Build GRCh38
rs764396192 Chr.1: 111476015-111476015 on Build GRCh38
rs764191148 Chr.1: 111476106-111476106 on Build GRCh38
rs780970939 Chr.1: 111476056-111476056 on Build GRCh38
rs868249859 Chr.1: 111476059-111476059 on Build GRCh38
rs928499686 Chr.1: 111476336-111476336 on Build GRCh38
rs777475444 Chr.1: 111476076-111476076 on Build GRCh38
rs983710450 Chr.1: 111476277-111476277 on Build GRCh38
rs759207416 Chr.1: 111476115-111476117 on Build GRCh38
rs890767975 Chr.1: 111476386-111476386 on Build GRCh38
rs952264371 Chr.1: 111476185-111476185 on Build GRCh38

View Full Product Details