Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111755464-111755956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX20
Entrez Gene ID
11218
Gene Name
DEAD-box helicase 20
Gene Aliases
DP103, GEMIN3
UniGene
Hs.591405
Gene Chromosome Location
Chr.1: 111755568-111767577 on Build GRCh38
Transcripts
NM_007204.4 NM_198926.2 NR_125963.1

Gene Symbol
FAM212B
Entrez Gene ID
55924
Gene Name
family with sequence similarity 212 member B
Gene Aliases
C1orf183
UniGene
Hs.193406
Gene Chromosome Location
Chr.1: 111722064-111755797 on Build GRCh38
Transcripts
NM_007204.4 NM_198926.2 NR_125963.1

Gene Symbol
LOC101928718
Entrez Gene ID
101928718
Gene Name
uncharacterized LOC101928718
Gene Aliases
-
UniGene
Hs.715034
Gene Chromosome Location
Chr.1: 111745317-111755509 on Build GRCh38
Transcripts
NM_007204.4 NM_198926.2 NR_125963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7316625 c.16G>A p.E6K Chr.1: 111755940-111755940 on Build GRCh38
1717000 c.24G>T p.S8S Chr.1: 111755948-111755948 on Build GRCh38
1294730 c.23C>T p.S8L Chr.1: 111755947-111755947 on Build GRCh38
7363963 c.5C>G p.A2G Chr.1: 111755929-111755929 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759025169 Chr.1: 111755927-111755927 on Build GRCh38
rs894144915 Chr.1: 111755777-111755777 on Build GRCh38
rs879156360 Chr.1: 111755648-111755648 on Build GRCh38
rs781613582 Chr.1: 111755880-111755880 on Build GRCh38
rs773178091 Chr.1: 111755919-111755919 on Build GRCh38
rs778448245 Chr.1: 111755716-111755720 on Build GRCh38
rs976060578 Chr.1: 111755601-111755601 on Build GRCh38
rs772094207 Chr.1: 111755571-111755571 on Build GRCh38
rs115627960 Chr.1: 111755537-111755537 on Build GRCh38
rs746001617 Chr.1: 111755708-111755708 on Build GRCh38
rs986508638 Chr.1: 111755796-111755796 on Build GRCh38
rs757626633 Chr.1: 111755874-111755874 on Build GRCh38
rs768767734 Chr.1: 111755890-111755890 on Build GRCh38
rs200709002 Chr.1: 111755690-111755690 on Build GRCh38
rs540982657 Chr.1: 111755553-111755553 on Build GRCh38
rs1015737185 Chr.1: 111755846-111755846 on Build GRCh38
rs761602815 Chr.1: 111755740-111755740 on Build GRCh38
rs374422621 Chr.1: 111755759-111755759 on Build GRCh38
rs548831610 Chr.1: 111755693-111755693 on Build GRCh38
rs760249081 Chr.1: 111755746-111755746 on Build GRCh38
rs369610165 Chr.1: 111755679-111755679 on Build GRCh38
rs1024104611 Chr.1: 111755551-111755551 on Build GRCh38
rs1036177706 Chr.1: 111755732-111755732 on Build GRCh38
rs990843355 Chr.1: 111755889-111755889 on Build GRCh38
rs775894300 Chr.1: 111755718-111755718 on Build GRCh38
rs374658908 Chr.1: 111755921-111755921 on Build GRCh38
rs552136171 Chr.1: 111755508-111755508 on Build GRCh38
rs774576459 Chr.1: 111755910-111755910 on Build GRCh38
rs532406636 Chr.1: 111755600-111755600 on Build GRCh38
rs957043296 Chr.1: 111755591-111755591 on Build GRCh38
rs747954216 Chr.1: 111755918-111755918 on Build GRCh38
rs878880658 Chr.1: 111755593-111755593 on Build GRCh38
rs747356882 Chr.1: 111755694-111755694 on Build GRCh38
rs570351865 Chr.1: 111755512-111755512 on Build GRCh38
rs908683978 Chr.1: 111755605-111755605 on Build GRCh38
rs756309917 Chr.1: 111755885-111755885 on Build GRCh38
rs907122387 Chr.1: 111755748-111755748 on Build GRCh38
rs958452761 Chr.1: 111755547-111755547 on Build GRCh38
rs769877172 Chr.1: 111755506-111755506 on Build GRCh38
rs767373118 Chr.1: 111755743-111755743 on Build GRCh38
rs767450660 Chr.1: 111755658-111755658 on Build GRCh38
rs940238382 Chr.1: 111755507-111755507 on Build GRCh38
rs749500927 Chr.1: 111755722-111755722 on Build GRCh38
rs755566580 Chr.1: 111755667-111755667 on Build GRCh38
rs370244042 Chr.1: 111755596-111755596 on Build GRCh38
rs377714525 Chr.1: 111755785-111755785 on Build GRCh38
rs1014114946 Chr.1: 111755548-111755548 on Build GRCh38
rs765847654 Chr.1: 111755749-111755749 on Build GRCh38
rs539084519 Chr.1: 111755815-111755815 on Build GRCh38
rs377342526 Chr.1: 111755577-111755577 on Build GRCh38
rs376610400 Chr.1: 111755700-111755700 on Build GRCh38
rs567756344 Chr.1: 111755565-111755565 on Build GRCh38
rs753293565 Chr.1: 111755751-111755751 on Build GRCh38
rs955278788 Chr.1: 111755599-111755599 on Build GRCh38
rs375567168 Chr.1: 111755487-111755487 on Build GRCh38
rs768177374 Chr.1: 111755817-111755817 on Build GRCh38
rs370286029 Chr.1: 111755741-111755741 on Build GRCh38
rs571581810 Chr.1: 111755787-111755787 on Build GRCh38
rs774048368 Chr.1: 111755727-111755727 on Build GRCh38
rs973631913 Chr.1: 111755605-111755648 on Build GRCh38
rs978255293 Chr.1: 111755587-111755587 on Build GRCh38
rs932105112 Chr.1: 111755686-111755686 on Build GRCh38
rs182023622 Chr.1: 111755483-111755483 on Build GRCh38
rs28372703 Chr.1: 111755756-111755756 on Build GRCh38
rs576783970 Chr.1: 111755484-111755484 on Build GRCh38
rs746346172 Chr.1: 111755884-111755884 on Build GRCh38
rs751565975 Chr.1: 111755652-111755652 on Build GRCh38
rs1051178842 Chr.1: 111755492-111755492 on Build GRCh38
rs776121486 Chr.1: 111755924-111755924 on Build GRCh38
rs751880444 Chr.1: 111755762-111755762 on Build GRCh38
rs555736162 Chr.1: 111755830-111755830 on Build GRCh38
rs368179651 Chr.1: 111755929-111755929 on Build GRCh38
rs761678816 Chr.1: 111755655-111755655 on Build GRCh38
rs749942842 Chr.1: 111755666-111755666 on Build GRCh38
rs766259547 Chr.1: 111755923-111755923 on Build GRCh38
rs781681321 Chr.1: 111755704-111755704 on Build GRCh38
rs538998934 Chr.1: 111755491-111755491 on Build GRCh38
rs371151742 Chr.1: 111755888-111755888 on Build GRCh38
rs769990073 Chr.1: 111755710-111755710 on Build GRCh38
rs908819029 Chr.1: 111755494-111755494 on Build GRCh38
rs940315870 Chr.1: 111755607-111755607 on Build GRCh38
rs775286452 Chr.1: 111755843-111755843 on Build GRCh38
rs555405634 Chr.1: 111755920-111755920 on Build GRCh38
rs1038569540 Chr.1: 111755608-111755608 on Build GRCh38
rs887606194 Chr.1: 111755489-111755489 on Build GRCh38
rs938036379 Chr.1: 111755614-111755614 on Build GRCh38
rs760596848 Chr.1: 111755863-111755863 on Build GRCh38
rs197391 Chr.1: 111755544-111755544 on Build GRCh38
rs563012378 Chr.1: 111755683-111755683 on Build GRCh38
rs568546970 Chr.1: 111755726-111755726 on Build GRCh38
rs893017418 Chr.1: 111755751-111755752 on Build GRCh38
rs919948272 Chr.1: 111755612-111755612 on Build GRCh38
rs575759519 Chr.1: 111755853-111755853 on Build GRCh38
rs547775643 Chr.1: 111755773-111755773 on Build GRCh38
rs752387882 Chr.1: 111755932-111755932 on Build GRCh38
rs949488260 Chr.1: 111755926-111755926 on Build GRCh38
rs552565168 Chr.1: 111755623-111755623 on Build GRCh38
rs575720013 Chr.1: 111755524-111755524 on Build GRCh38
rs541667841 Chr.1: 111755530-111755530 on Build GRCh38
rs896570602 Chr.1: 111755649-111755650 on Build GRCh38

View Full Product Details