Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 112412113-112412606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTTNBP2NL
Entrez Gene ID
55917
Gene Name
CTTNBP2 N-terminal like
Gene Aliases
-
UniGene
Hs.744100
Gene Chromosome Location
Chr.1: 112391085-112461164 on Build GRCh38
Transcripts
NM_018704.2 XM_011541781.2 XM_017001806.1

SNP Detail

SNP ID Chromosome Location
rs187416143 Chr.1: 112412487-112412487 on Build GRCh38
rs1040841931 Chr.1: 112412322-112412322 on Build GRCh38
rs925795677 Chr.1: 112412293-112412293 on Build GRCh38
rs1005395711 Chr.1: 112412416-112412416 on Build GRCh38
rs1015913786 Chr.1: 112412377-112412377 on Build GRCh38
rs978399081 Chr.1: 112412491-112412491 on Build GRCh38
rs958969377 Chr.1: 112412477-112412477 on Build GRCh38
rs932350915 Chr.1: 112412569-112412569 on Build GRCh38
rs546512668 Chr.1: 112412233-112412233 on Build GRCh38
rs888364107 Chr.1: 112412371-112412371 on Build GRCh38
rs575071209 Chr.1: 112412560-112412560 on Build GRCh38
rs1040705889 Chr.1: 112412552-112412552 on Build GRCh38
rs556725512 Chr.1: 112412517-112412517 on Build GRCh38
rs912114852 Chr.1: 112412529-112412529 on Build GRCh38
rs977575636 Chr.1: 112412502-112412502 on Build GRCh38
rs970776312 Chr.1: 112412408-112412408 on Build GRCh38
rs995039804 Chr.1: 112412459-112412459 on Build GRCh38
rs962401 Chr.1: 112412144-112412144 on Build GRCh38
rs773945569 Chr.1: 112412381-112412381 on Build GRCh38
rs142503815 Chr.1: 112412573-112412573 on Build GRCh38
rs552994751 Chr.1: 112412467-112412467 on Build GRCh38
rs921078626 Chr.1: 112412533-112412533 on Build GRCh38
rs150306167 Chr.1: 112412442-112412442 on Build GRCh38
rs993069439 Chr.1: 112412478-112412478 on Build GRCh38
rs958938527 Chr.1: 112412177-112412177 on Build GRCh38
rs568978559 Chr.1: 112412568-112412568 on Build GRCh38
rs935744226 Chr.1: 112412528-112412534 on Build GRCh38
rs747753955 Chr.1: 112412475-112412475 on Build GRCh38
rs992536104 Chr.1: 112412182-112412182 on Build GRCh38
rs115837637 Chr.1: 112412355-112412355 on Build GRCh38
rs779840201 Chr.1: 112412306-112412306 on Build GRCh38
rs890434567 Chr.1: 112412413-112412413 on Build GRCh38
rs1034567414 Chr.1: 112412158-112412158 on Build GRCh38
rs115454178 Chr.1: 112412476-112412476 on Build GRCh38
rs1043027482 Chr.1: 112412273-112412273 on Build GRCh38
rs137977340 Chr.1: 112412505-112412505 on Build GRCh38
rs566318279 Chr.1: 112412335-112412335 on Build GRCh38
rs758294668 Chr.1: 112412275-112412275 on Build GRCh38
rs557004020 Chr.1: 112412409-112412409 on Build GRCh38
rs915498883 Chr.1: 112412146-112412146 on Build GRCh38
rs1412931 Chr.1: 112412342-112412342 on Build GRCh38
rs191510918 Chr.1: 112412434-112412434 on Build GRCh38
rs962402 Chr.1: 112412372-112412372 on Build GRCh38
rs943737759 Chr.1: 112412545-112412545 on Build GRCh38
rs945812662 Chr.1: 112412264-112412264 on Build GRCh38
rs866058720 Chr.1: 112412213-112412213 on Build GRCh38
rs188092668 Chr.1: 112412194-112412194 on Build GRCh38
rs549449336 Chr.1: 112412145-112412145 on Build GRCh38
rs182416061 Chr.1: 112412482-112412482 on Build GRCh38

View Full Product Details