Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 112619367-112619871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPZA1
Entrez Gene ID
829
Gene Name
capping actin protein of muscle Z-line alpha subunit 1
Gene Aliases
CAPPA1, CAPZ, CAZ1
UniGene
Hs.744974
Gene Chromosome Location
Chr.1: 112619453-112671619 on Build GRCh38
Transcripts
NM_006135.2 XM_011542225.2 XM_017002424.1 NM_017744.4 NM_138727.3 NM_138728.2 NM_138729.3 XM_011541630.2 XM_011541631.2

Gene Symbol
ST7L
Entrez Gene ID
54879
Gene Name
suppression of tumorigenicity 7 like
Gene Aliases
FAM4B, ST7R, STLR
UniGene
Hs.201921
Gene Chromosome Location
Chr.1: 112517444-112620838 on Build GRCh38
Transcripts
NM_006135.2 XM_011542225.2 XM_017002424.1 NM_017744.4 NM_138727.3 NM_138728.2 NM_138729.3 XM_011541630.2 XM_011541631.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6218934 c.27G>C p.S9S Chr.1: 112619871-112619871 on Build GRCh38
243478 c.6C>A p.A2A Chr.1: 112619850-112619850 on Build GRCh38
7207040 c.19C>T p.R7C Chr.1: 112619863-112619863 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs878978565 Chr.1: 112619411-112619411 on Build GRCh38
rs924140957 Chr.1: 112619791-112619791 on Build GRCh38
rs35979114 Chr.1: 112619550-112619551 on Build GRCh38
rs1025518467 Chr.1: 112619815-112619815 on Build GRCh38
rs933475236 Chr.1: 112619754-112619754 on Build GRCh38
rs865858132 Chr.1: 112619785-112619785 on Build GRCh38
rs1016080639 Chr.1: 112619487-112619487 on Build GRCh38
rs1005093057 Chr.1: 112619709-112619709 on Build GRCh38
rs949619465 Chr.1: 112619410-112619410 on Build GRCh38
rs548292767 Chr.1: 112619616-112619616 on Build GRCh38
rs930036241 Chr.1: 112619395-112619395 on Build GRCh38
rs1045058877 Chr.1: 112619670-112619670 on Build GRCh38
rs200729464 Chr.1: 112619803-112619803 on Build GRCh38
rs996057657 Chr.1: 112619417-112619418 on Build GRCh38
rs955149528 Chr.1: 112619546-112619546 on Build GRCh38
rs758100278 Chr.1: 112619821-112619821 on Build GRCh38
rs948169460 Chr.1: 112619659-112619659 on Build GRCh38
rs1000459600 Chr.1: 112619414-112619415 on Build GRCh38
rs371977187 Chr.1: 112619829-112619829 on Build GRCh38
rs1014776138 Chr.1: 112619720-112619720 on Build GRCh38
rs756265125 Chr.1: 112619805-112619805 on Build GRCh38
rs746143710 Chr.1: 112619678-112619679 on Build GRCh38
rs1042574375 Chr.1: 112619644-112619644 on Build GRCh38
rs761498923 Chr.1: 112619543-112619543 on Build GRCh38
rs12087862 Chr.1: 112619814-112619814 on Build GRCh38
rs145904439 Chr.1: 112619585-112619586 on Build GRCh38
rs868704553 Chr.1: 112619413-112619413 on Build GRCh38
rs973919241 Chr.1: 112619392-112619392 on Build GRCh38
rs3790597 Chr.1: 112619415-112619415 on Build GRCh38
rs772668991 Chr.1: 112619776-112619776 on Build GRCh38
rs34583754 Chr.1: 112619407-112619408 on Build GRCh38
rs983327159 Chr.1: 112619475-112619475 on Build GRCh38
rs115858905 Chr.1: 112619628-112619628 on Build GRCh38
rs919925973 Chr.1: 112619530-112619530 on Build GRCh38
rs1006551593 Chr.1: 112619564-112619564 on Build GRCh38
rs1030281356 Chr.1: 112619452-112619452 on Build GRCh38
rs983122611 Chr.1: 112619677-112619678 on Build GRCh38
rs577898492 Chr.1: 112619416-112619416 on Build GRCh38
rs752521238 Chr.1: 112619816-112619816 on Build GRCh38
rs959343633 Chr.1: 112619766-112619766 on Build GRCh38
rs892601183 Chr.1: 112619725-112619725 on Build GRCh38
rs995836961 Chr.1: 112619688-112619688 on Build GRCh38
rs962638406 Chr.1: 112619578-112619578 on Build GRCh38
rs527993035 Chr.1: 112619666-112619666 on Build GRCh38
rs898166353 Chr.1: 112619813-112619813 on Build GRCh38
rs539643061 Chr.1: 112619724-112619724 on Build GRCh38
rs142689145 Chr.1: 112619850-112619850 on Build GRCh38
rs995585125 Chr.1: 112619746-112619746 on Build GRCh38
rs200110073 Chr.1: 112619839-112619839 on Build GRCh38
rs868603161 Chr.1: 112619706-112619706 on Build GRCh38
rs117684232 Chr.1: 112619522-112619522 on Build GRCh38
rs371309950 Chr.1: 112619832-112619832 on Build GRCh38
rs1042514447 Chr.1: 112619412-112619413 on Build GRCh38
rs769590445 Chr.1: 112619831-112619831 on Build GRCh38
rs758438301 Chr.1: 112619707-112619707 on Build GRCh38
rs546364242 Chr.1: 112619453-112619453 on Build GRCh38
rs971022960 Chr.1: 112619782-112619782 on Build GRCh38
rs541292738 Chr.1: 112619414-112619414 on Build GRCh38
rs961496298 Chr.1: 112619500-112619500 on Build GRCh38
rs745316069 Chr.1: 112619830-112619830 on Build GRCh38
rs918388007 Chr.1: 112619587-112619587 on Build GRCh38
rs936427554 Chr.1: 112619665-112619666 on Build GRCh38
rs959397062 Chr.1: 112619535-112619535 on Build GRCh38
rs1055267837 Chr.1: 112619408-112619408 on Build GRCh38
rs531521553 Chr.1: 112619571-112619571 on Build GRCh38
rs549507726 Chr.1: 112619770-112619770 on Build GRCh38
rs539254227 Chr.1: 112619790-112619790 on Build GRCh38
rs570984636 Chr.1: 112619721-112619721 on Build GRCh38
rs547148420 Chr.1: 112619467-112619467 on Build GRCh38
rs375815657 Chr.1: 112619589-112619590 on Build GRCh38
rs779182044 Chr.1: 112619583-112619583 on Build GRCh38
rs961411422 Chr.1: 112619722-112619722 on Build GRCh38
rs923663073 Chr.1: 112619650-112619650 on Build GRCh38
rs1045511499 Chr.1: 112619420-112619420 on Build GRCh38
rs772131449 Chr.1: 112619777-112619777 on Build GRCh38
rs1052583206 Chr.1: 112619483-112619483 on Build GRCh38
rs1027113786 Chr.1: 112619755-112619755 on Build GRCh38
rs370756354 Chr.1: 112619809-112619809 on Build GRCh38
rs1004702835 Chr.1: 112619404-112619404 on Build GRCh38
rs1009956629 Chr.1: 112619537-112619537 on Build GRCh38
rs1029423872 Chr.1: 112619538-112619538 on Build GRCh38
rs951223521 Chr.1: 112619612-112619612 on Build GRCh38
rs765006137 Chr.1: 112619762-112619762 on Build GRCh38
rs374422270 Chr.1: 112619800-112619800 on Build GRCh38
rs368427318 Chr.1: 112619828-112619828 on Build GRCh38
rs572254518 Chr.1: 112619394-112619394 on Build GRCh38
rs187457672 Chr.1: 112619680-112619680 on Build GRCh38

View Full Product Details