Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 112619769-112620262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPZA1
Entrez Gene ID
829
Gene Name
capping actin protein of muscle Z-line alpha subunit 1
Gene Aliases
CAPPA1, CAPZ, CAZ1
UniGene
Hs.744974
Gene Chromosome Location
Chr.1: 112619453-112671619 on Build GRCh38
Transcripts
NM_006135.2 XM_011542225.2 XM_017002424.1 XM_011541630.2

Gene Symbol
ST7L
Entrez Gene ID
54879
Gene Name
suppression of tumorigenicity 7 like
Gene Aliases
FAM4B, ST7R, STLR
UniGene
Hs.201921
Gene Chromosome Location
Chr.1: 112517444-112620838 on Build GRCh38
Transcripts
NM_006135.2 XM_011542225.2 XM_017002424.1 XM_011541630.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5377212 c.39+1G>A p.? Chr.1: 112619884-112619884 on Build GRCh38
3975892 c.39+7G>A p.? Chr.1: 112619890-112619890 on Build GRCh38
5377211 c.39G>A p.K13K Chr.1: 112619883-112619883 on Build GRCh38
7359482 c.28G>A p.D10N Chr.1: 112619872-112619872 on Build GRCh38
6218934 c.27G>C p.S9S Chr.1: 112619871-112619871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749414612 Chr.1: 112619943-112619943 on Build GRCh38
rs924140957 Chr.1: 112619791-112619791 on Build GRCh38
rs971354853 Chr.1: 112619880-112619880 on Build GRCh38
rs199900757 Chr.1: 112619916-112619916 on Build GRCh38
rs542273761 Chr.1: 112619952-112619952 on Build GRCh38
rs1025518467 Chr.1: 112619815-112619815 on Build GRCh38
rs931749815 Chr.1: 112619934-112619934 on Build GRCh38
rs757436909 Chr.1: 112619944-112619944 on Build GRCh38
rs920488937 Chr.1: 112620227-112620227 on Build GRCh38
rs958185536 Chr.1: 112620075-112620075 on Build GRCh38
rs887011599 Chr.1: 112620020-112620020 on Build GRCh38
rs753166751 Chr.1: 112619895-112619895 on Build GRCh38
rs999957706 Chr.1: 112620121-112620121 on Build GRCh38
rs200729464 Chr.1: 112619803-112619803 on Build GRCh38
rs112717924 Chr.1: 112620158-112620158 on Build GRCh38
rs764315909 Chr.1: 112619918-112619918 on Build GRCh38
rs41283062 Chr.1: 112619872-112619872 on Build GRCh38
rs758100278 Chr.1: 112619821-112619821 on Build GRCh38
rs911656143 Chr.1: 112620196-112620196 on Build GRCh38
rs564431646 Chr.1: 112620046-112620046 on Build GRCh38
rs761047652 Chr.1: 112619897-112619898 on Build GRCh38
rs762550816 Chr.1: 112620201-112620201 on Build GRCh38
rs371977187 Chr.1: 112619829-112619829 on Build GRCh38
rs569887544 Chr.1: 112620159-112620159 on Build GRCh38
rs769413841 Chr.1: 112620129-112620129 on Build GRCh38
rs778060979 Chr.1: 112619930-112619930 on Build GRCh38
rs987500795 Chr.1: 112619935-112619935 on Build GRCh38
rs756265125 Chr.1: 112619805-112619805 on Build GRCh38
rs368745152 Chr.1: 112619929-112619929 on Build GRCh38
rs561974550 Chr.1: 112619982-112619982 on Build GRCh38
rs12087862 Chr.1: 112619814-112619814 on Build GRCh38
rs372337838 Chr.1: 112619865-112619865 on Build GRCh38
rs200758434 Chr.1: 112619870-112619870 on Build GRCh38
rs969636113 Chr.1: 112620047-112620047 on Build GRCh38
rs374051421 Chr.1: 112620127-112620127 on Build GRCh38
rs543649792 Chr.1: 112619890-112619890 on Build GRCh38
rs759574473 Chr.1: 112619856-112619856 on Build GRCh38
rs914151348 Chr.1: 112619863-112619863 on Build GRCh38
rs777092625 Chr.1: 112619899-112619899 on Build GRCh38
rs572539858 Chr.1: 112619971-112619971 on Build GRCh38
rs745918244 Chr.1: 112620029-112620029 on Build GRCh38
rs772918012 Chr.1: 112620078-112620078 on Build GRCh38
rs527661845 Chr.1: 112620016-112620016 on Build GRCh38
rs752521238 Chr.1: 112619816-112619816 on Build GRCh38
rs779097887 Chr.1: 112620024-112620024 on Build GRCh38
rs772555221 Chr.1: 112619889-112619889 on Build GRCh38
rs910684086 Chr.1: 112620177-112620177 on Build GRCh38
rs987734878 Chr.1: 112620169-112620169 on Build GRCh38
rs372431468 Chr.1: 112620183-112620183 on Build GRCh38
rs898166353 Chr.1: 112619813-112619813 on Build GRCh38
rs1029118409 Chr.1: 112620175-112620175 on Build GRCh38
rs144794798 Chr.1: 112619866-112619866 on Build GRCh38
rs202024562 Chr.1: 112619915-112619915 on Build GRCh38
rs142689145 Chr.1: 112619850-112619850 on Build GRCh38
rs200110073 Chr.1: 112619839-112619839 on Build GRCh38
rs528997047 Chr.1: 112620194-112620194 on Build GRCh38
rs930436515 Chr.1: 112620057-112620057 on Build GRCh38
rs751329458 Chr.1: 112619919-112619919 on Build GRCh38
rs549049248 Chr.1: 112620210-112620210 on Build GRCh38
rs371309950 Chr.1: 112619832-112619832 on Build GRCh38
rs765485560 Chr.1: 112619857-112619857 on Build GRCh38
rs769590445 Chr.1: 112619831-112619831 on Build GRCh38
rs766813977 Chr.1: 112619853-112619853 on Build GRCh38
rs377292807 Chr.1: 112619854-112619854 on Build GRCh38
rs1025514759 Chr.1: 112620115-112620115 on Build GRCh38
rs539918627 Chr.1: 112620062-112620062 on Build GRCh38
rs768827553 Chr.1: 112619896-112619897 on Build GRCh38
rs919661807 Chr.1: 112620212-112620212 on Build GRCh38
rs745316069 Chr.1: 112619830-112619830 on Build GRCh38
rs113823236 Chr.1: 112619860-112619860 on Build GRCh38
rs149653791 Chr.1: 112619878-112619878 on Build GRCh38
rs772936582 Chr.1: 112620131-112620131 on Build GRCh38
rs761052774 Chr.1: 112619891-112619891 on Build GRCh38
rs745399525 Chr.1: 112619873-112619873 on Build GRCh38
rs757075678 Chr.1: 112619920-112619920 on Build GRCh38
rs940391858 Chr.1: 112619992-112619992 on Build GRCh38
rs539254227 Chr.1: 112619790-112619790 on Build GRCh38
rs779900493 Chr.1: 112619874-112619874 on Build GRCh38
rs180902247 Chr.1: 112620019-112620019 on Build GRCh38
rs988128764 Chr.1: 112620116-112620116 on Build GRCh38
rs752843240 Chr.1: 112619858-112619858 on Build GRCh38
rs780986846 Chr.1: 112620056-112620056 on Build GRCh38
rs370756354 Chr.1: 112619809-112619809 on Build GRCh38
rs765290786 Chr.1: 112619910-112619910 on Build GRCh38
rs768770546 Chr.1: 112620051-112620051 on Build GRCh38
rs978359858 Chr.1: 112619906-112619907 on Build GRCh38
rs759664397 Chr.1: 112619909-112619909 on Build GRCh38
rs753937540 Chr.1: 112619855-112619855 on Build GRCh38
rs771461250 Chr.1: 112619893-112619893 on Build GRCh38
rs374422270 Chr.1: 112619800-112619800 on Build GRCh38
rs368427318 Chr.1: 112619828-112619828 on Build GRCh38

View Full Product Details