Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 113887292-113887801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4B1-AS1
Entrez Gene ID
100287722
Gene Name
AP4B1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.664669
Gene Chromosome Location
Chr.1: 113812612-113901237 on Build GRCh38
Transcripts
NR_037864.1 NR_125965.1 NM_001010922.2

Gene Symbol
BCL2L15
Entrez Gene ID
440603
Gene Name
BCL2 like 15
Gene Aliases
Bfk, C1orf178
UniGene
Hs.123106
Gene Chromosome Location
Chr.1: 113876814-113887547 on Build GRCh38
Transcripts
NR_037864.1 NR_125965.1 NM_001010922.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
894088 c.32C>T p.T11M Chr.1: 113887344-113887344 on Build GRCh38
894087 c.66C>A p.F22L Chr.1: 113887310-113887310 on Build GRCh38
4020798 c.52C>T p.L18L Chr.1: 113887324-113887324 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1539438 Chr.1: 113887734-113887734 on Build GRCh38
rs759047386 Chr.1: 113887363-113887363 on Build GRCh38
rs774300907 Chr.1: 113887339-113887339 on Build GRCh38
rs1009500901 Chr.1: 113887387-113887387 on Build GRCh38
rs141475527 Chr.1: 113887658-113887658 on Build GRCh38
rs941575161 Chr.1: 113887631-113887631 on Build GRCh38
rs111381432 Chr.1: 113887520-113887520 on Build GRCh38
rs778938769 Chr.1: 113887410-113887410 on Build GRCh38
rs1045012329 Chr.1: 113887663-113887663 on Build GRCh38
rs146604347 Chr.1: 113887448-113887448 on Build GRCh38
rs752305101 Chr.1: 113887366-113887366 on Build GRCh38
rs988936896 Chr.1: 113887492-113887492 on Build GRCh38
rs990619780 Chr.1: 113887698-113887698 on Build GRCh38
rs765988230 Chr.1: 113887347-113887347 on Build GRCh38
rs114576636 Chr.1: 113887466-113887466 on Build GRCh38
rs923908720 Chr.1: 113887757-113887757 on Build GRCh38
rs754929023 Chr.1: 113887674-113887674 on Build GRCh38
rs375501688 Chr.1: 113887332-113887332 on Build GRCh38
rs750058099 Chr.1: 113887533-113887533 on Build GRCh38
rs559723309 Chr.1: 113887635-113887635 on Build GRCh38
rs748282840 Chr.1: 113887415-113887415 on Build GRCh38
rs1046352653 Chr.1: 113887432-113887432 on Build GRCh38
rs751376707 Chr.1: 113887667-113887667 on Build GRCh38
rs776161431 Chr.1: 113887348-113887348 on Build GRCh38
rs372835450 Chr.1: 113887409-113887409 on Build GRCh38
rs34304177 Chr.1: 113887526-113887527 on Build GRCh38
rs189391190 Chr.1: 113887659-113887659 on Build GRCh38
rs1056456799 Chr.1: 113887648-113887648 on Build GRCh38
rs566458593 Chr.1: 113887539-113887539 on Build GRCh38
rs199687942 Chr.1: 113887343-113887343 on Build GRCh38
rs750753100 Chr.1: 113887388-113887388 on Build GRCh38
rs890672736 Chr.1: 113887585-113887585 on Build GRCh38
rs757827844 Chr.1: 113887367-113887367 on Build GRCh38
rs905785040 Chr.1: 113887679-113887679 on Build GRCh38
rs930105792 Chr.1: 113887644-113887644 on Build GRCh38
rs771990795 Chr.1: 113887425-113887425 on Build GRCh38
rs200731954 Chr.1: 113887342-113887342 on Build GRCh38
rs955004919 Chr.1: 113887510-113887510 on Build GRCh38
rs970587371 Chr.1: 113887708-113887708 on Build GRCh38
rs768208657 Chr.1: 113887382-113887382 on Build GRCh38
rs201837071 Chr.1: 113887395-113887395 on Build GRCh38
rs911467773 Chr.1: 113887608-113887608 on Build GRCh38
rs557280929 Chr.1: 113887563-113887563 on Build GRCh38
rs578023390 Chr.1: 113887489-113887489 on Build GRCh38
rs758034765 Chr.1: 113887620-113887620 on Build GRCh38
rs78095589 Chr.1: 113887364-113887364 on Build GRCh38
rs781046997 Chr.1: 113887702-113887702 on Build GRCh38
rs1022835495 Chr.1: 113887376-113887376 on Build GRCh38
rs182041526 Chr.1: 113887664-113887664 on Build GRCh38
rs767090830 Chr.1: 113887359-113887359 on Build GRCh38
rs1009562554 Chr.1: 113887591-113887591 on Build GRCh38
rs935379723 Chr.1: 113887457-113887457 on Build GRCh38
rs202039250 Chr.1: 113887335-113887335 on Build GRCh38
rs574085522 Chr.1: 113887580-113887580 on Build GRCh38
rs542939291 Chr.1: 113887634-113887634 on Build GRCh38
rs201294219 Chr.1: 113887402-113887402 on Build GRCh38
rs761350107 Chr.1: 113887344-113887344 on Build GRCh38
rs922290801 Chr.1: 113887493-113887493 on Build GRCh38
rs949291614 Chr.1: 113887424-113887424 on Build GRCh38

View Full Product Details