Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 113904844-113905342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4B1
Entrez Gene ID
10717
Gene Name
adaptor related protein complex 4 beta 1 subunit
Gene Aliases
BETA-4, CPSQ5, SPG47
UniGene
Hs.515048
Gene Chromosome Location
Chr.1: 113894194-113905202 on Build GRCh38
Transcripts
NM_001253852.2 NM_001253853.2 NM_001308312.1 NM_006594.4 XM_011540523.2 XM_011540524.2 XM_011540525.2 XM_011540528.2 XM_017000088.1 XM_017000090.1 XM_017000091.1 NM_001319946.1 NM_001319947.1 NM_022836.3

Gene Symbol
DCLRE1B
Entrez Gene ID
64858
Gene Name
DNA cross-link repair 1B
Gene Aliases
APOLLO, SNM1B, SNMIB
UniGene
Hs.591412
Gene Chromosome Location
Chr.1: 113904619-113914086 on Build GRCh38
Transcripts
NM_001253852.2 NM_001253853.2 NM_001308312.1 NM_006594.4 XM_011540523.2 XM_011540524.2 XM_011540525.2 XM_011540528.2 XM_017000088.1 XM_017000090.1 XM_017000091.1 NM_001319946.1 NM_001319947.1 NM_022836.3

SNP Detail

SNP ID Chromosome Location
rs939051175 Chr.1: 113905148-113905148 on Build GRCh38
rs539322999 Chr.1: 113905205-113905205 on Build GRCh38
rs1030268176 Chr.1: 113904935-113904935 on Build GRCh38
rs888219942 Chr.1: 113904951-113904951 on Build GRCh38
rs879814442 Chr.1: 113905258-113905258 on Build GRCh38
rs1036793185 Chr.1: 113905129-113905129 on Build GRCh38
rs1004788811 Chr.1: 113905170-113905170 on Build GRCh38
rs547372260 Chr.1: 113905193-113905193 on Build GRCh38
rs916230586 Chr.1: 113905317-113905317 on Build GRCh38
rs1011567985 Chr.1: 113905158-113905158 on Build GRCh38
rs957711537 Chr.1: 113905257-113905257 on Build GRCh38
rs1018945720 Chr.1: 113905056-113905056 on Build GRCh38
rs56246459 Chr.1: 113905285-113905285 on Build GRCh38
rs1043911312 Chr.1: 113905161-113905161 on Build GRCh38
rs1051594650 Chr.1: 113905289-113905289 on Build GRCh38
rs1048679777 Chr.1: 113904950-113904950 on Build GRCh38
rs552091954 Chr.1: 113905002-113905002 on Build GRCh38
rs189372881 Chr.1: 113905195-113905195 on Build GRCh38
rs1003787500 Chr.1: 113904863-113904863 on Build GRCh38
rs376810969 Chr.1: 113904914-113904914 on Build GRCh38
rs1054833220 Chr.1: 113905315-113905315 on Build GRCh38
rs1014427378 Chr.1: 113905222-113905222 on Build GRCh38
rs933183085 Chr.1: 113905144-113905144 on Build GRCh38
rs924924880 Chr.1: 113905256-113905256 on Build GRCh38
rs913493320 Chr.1: 113905025-113905025 on Build GRCh38
rs940593217 Chr.1: 113905298-113905298 on Build GRCh38
rs182022902 Chr.1: 113905200-113905200 on Build GRCh38
rs966328596 Chr.1: 113905069-113905069 on Build GRCh38
rs533659472 Chr.1: 113905192-113905192 on Build GRCh38
rs939085507 Chr.1: 113905313-113905313 on Build GRCh38
rs775396400 Chr.1: 113905005-113905005 on Build GRCh38
rs921738822 Chr.1: 113905134-113905134 on Build GRCh38
rs113338770 Chr.1: 113905000-113905000 on Build GRCh38
rs750919643 Chr.1: 113905015-113905023 on Build GRCh38
rs960252580 Chr.1: 113905014-113905014 on Build GRCh38
rs745544312 Chr.1: 113904980-113904980 on Build GRCh38
rs921209040 Chr.1: 113905083-113905083 on Build GRCh38
rs865904191 Chr.1: 113904945-113904945 on Build GRCh38
rs1022873895 Chr.1: 113905045-113905045 on Build GRCh38
rs971173 Chr.1: 113905292-113905292 on Build GRCh38
rs28364564 Chr.1: 113905020-113905020 on Build GRCh38
rs939737614 Chr.1: 113904963-113904963 on Build GRCh38
rs978093391 Chr.1: 113904927-113904927 on Build GRCh38
rs776451149 Chr.1: 113905207-113905207 on Build GRCh38
rs1217397 Chr.1: 113904943-113904943 on Build GRCh38
rs186281241 Chr.1: 113905187-113905187 on Build GRCh38
rs976698692 Chr.1: 113904944-113904944 on Build GRCh38
rs895429443 Chr.1: 113905155-113905155 on Build GRCh38
rs977750798 Chr.1: 113905073-113905073 on Build GRCh38
rs886353594 Chr.1: 113905167-113905167 on Build GRCh38
rs575828809 Chr.1: 113904874-113904874 on Build GRCh38
rs978145501 Chr.1: 113905055-113905055 on Build GRCh38
rs28364565 Chr.1: 113905234-113905234 on Build GRCh38
rs1037651415 Chr.1: 113905186-113905186 on Build GRCh38
rs974872037 Chr.1: 113905262-113905262 on Build GRCh38
rs983998489 Chr.1: 113905088-113905088 on Build GRCh38
rs909706800 Chr.1: 113905104-113905104 on Build GRCh38
rs561405964 Chr.1: 113904910-113904910 on Build GRCh38
rs975119117 Chr.1: 113905079-113905079 on Build GRCh38
rs536181728 Chr.1: 113904892-113904892 on Build GRCh38
rs1217398 Chr.1: 113905154-113905154 on Build GRCh38
rs966800143 Chr.1: 113905052-113905052 on Build GRCh38
rs766600719 Chr.1: 113905295-113905295 on Build GRCh38
rs1003178469 Chr.1: 113904994-113904994 on Build GRCh38
rs531565513 Chr.1: 113905133-113905133 on Build GRCh38
rs113177502 Chr.1: 113905242-113905242 on Build GRCh38
rs966749073 Chr.1: 113904904-113904904 on Build GRCh38
rs984440829 Chr.1: 113905269-113905269 on Build GRCh38
rs932513140 Chr.1: 113904949-113904949 on Build GRCh38
rs780530733 Chr.1: 113905087-113905087 on Build GRCh38
rs979038412 Chr.1: 113905243-113905243 on Build GRCh38
rs960449536 Chr.1: 113905201-113905201 on Build GRCh38
rs549325966 Chr.1: 113904872-113904872 on Build GRCh38
rs927671466 Chr.1: 113905130-113905130 on Build GRCh38
rs28364563 Chr.1: 113904937-113904937 on Build GRCh38
rs1022398381 Chr.1: 113904903-113904903 on Build GRCh38
rs939788537 Chr.1: 113905125-113905125 on Build GRCh38
rs909759243 Chr.1: 113905274-113905274 on Build GRCh38
rs189306022 Chr.1: 113904989-113904989 on Build GRCh38
rs1004841045 Chr.1: 113904939-113904939 on Build GRCh38
rs1036729758 Chr.1: 113904971-113904971 on Build GRCh38
rs954019430 Chr.1: 113905264-113905264 on Build GRCh38
rs966025265 Chr.1: 113905071-113905071 on Build GRCh38

View Full Product Details