Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114585336-114585833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND2C
Entrez Gene ID
163259
Gene Name
DENN domain containing 2C
Gene Aliases
dJ1156J9.1
UniGene
Hs.654928
Gene Chromosome Location
Chr.1: 114584575-114670111 on Build GRCh38
Transcripts
NM_001256404.1 NM_198459.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
894195 c.2610G>T p.K870N Chr.1: 114585606-114585606 on Build GRCh38
3720493 c.2784A>G p.K928K Chr.1: 114585603-114585603 on Build GRCh38
3720492 c.2613A>G p.K871K Chr.1: 114585603-114585603 on Build GRCh38
4874939 c.2781G>T p.K927N Chr.1: 114585606-114585606 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs908583689 Chr.1: 114585477-114585477 on Build GRCh38
rs201437846 Chr.1: 114585582-114585582 on Build GRCh38
rs754269161 Chr.1: 114585593-114585594 on Build GRCh38
rs149880335 Chr.1: 114585486-114585486 on Build GRCh38
rs922448202 Chr.1: 114585400-114585400 on Build GRCh38
rs1056611402 Chr.1: 114585507-114585507 on Build GRCh38
rs559101289 Chr.1: 114585531-114585531 on Build GRCh38
rs1043434521 Chr.1: 114585589-114585589 on Build GRCh38
rs560018064 Chr.1: 114585596-114585596 on Build GRCh38
rs901754912 Chr.1: 114585692-114585692 on Build GRCh38
rs561626330 Chr.1: 114585639-114585639 on Build GRCh38
rs577452520 Chr.1: 114585556-114585556 on Build GRCh38
rs1016945539 Chr.1: 114585803-114585803 on Build GRCh38
rs753192925 Chr.1: 114585655-114585655 on Build GRCh38
rs975341677 Chr.1: 114585766-114585767 on Build GRCh38
rs999894499 Chr.1: 114585684-114585684 on Build GRCh38
rs575062106 Chr.1: 114585623-114585623 on Build GRCh38
rs758888889 Chr.1: 114585661-114585661 on Build GRCh38
rs991384272 Chr.1: 114585481-114585481 on Build GRCh38
rs543887916 Chr.1: 114585511-114585511 on Build GRCh38
rs869052712 Chr.1: 114585370-114585371 on Build GRCh38
rs757754506 Chr.1: 114585669-114585669 on Build GRCh38
rs961342576 Chr.1: 114585808-114585808 on Build GRCh38
rs776660967 Chr.1: 114585578-114585578 on Build GRCh38
rs762809344 Chr.1: 114585591-114585591 on Build GRCh38
rs775459970 Chr.1: 114585586-114585586 on Build GRCh38
rs781220380 Chr.1: 114585710-114585713 on Build GRCh38
rs531405067 Chr.1: 114585369-114585369 on Build GRCh38
rs1028436497 Chr.1: 114585363-114585363 on Build GRCh38
rs754042107 Chr.1: 114585708-114585708 on Build GRCh38
rs139936260 Chr.1: 114585485-114585485 on Build GRCh38
rs933785378 Chr.1: 114585413-114585413 on Build GRCh38
rs984176734 Chr.1: 114585380-114585380 on Build GRCh38
rs963944612 Chr.1: 114585366-114585366 on Build GRCh38
rs146159704 Chr.1: 114585479-114585479 on Build GRCh38
rs576380433 Chr.1: 114585503-114585503 on Build GRCh38
rs889909388 Chr.1: 114585745-114585745 on Build GRCh38
rs369709860 Chr.1: 114585646-114585646 on Build GRCh38
rs222493 Chr.1: 114585466-114585466 on Build GRCh38
rs1029238891 Chr.1: 114585732-114585732 on Build GRCh38
rs909875259 Chr.1: 114585410-114585410 on Build GRCh38
rs542174209 Chr.1: 114585685-114585685 on Build GRCh38
rs767323244 Chr.1: 114585633-114585633 on Build GRCh38
rs769788635 Chr.1: 114585585-114585585 on Build GRCh38
rs998689485 Chr.1: 114585360-114585360 on Build GRCh38
rs1040239498 Chr.1: 114585681-114585681 on Build GRCh38
rs1006014057 Chr.1: 114585764-114585764 on Build GRCh38
rs553593163 Chr.1: 114585478-114585478 on Build GRCh38
rs935252412 Chr.1: 114585673-114585673 on Build GRCh38
rs961628669 Chr.1: 114585456-114585456 on Build GRCh38
rs375699722 Chr.1: 114585367-114585368 on Build GRCh38
rs778127344 Chr.1: 114585552-114585552 on Build GRCh38
rs571064176 Chr.1: 114585448-114585448 on Build GRCh38
rs180678695 Chr.1: 114585444-114585444 on Build GRCh38
rs938282815 Chr.1: 114585492-114585492 on Build GRCh38
rs765808299 Chr.1: 114585649-114585649 on Build GRCh38
rs771214011 Chr.1: 114585570-114585570 on Build GRCh38
rs963685709 Chr.1: 114585752-114585752 on Build GRCh38
rs761525996 Chr.1: 114585630-114585630 on Build GRCh38
rs902923915 Chr.1: 114585679-114585679 on Build GRCh38

View Full Product Details