Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114587604-114588115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND2C
Entrez Gene ID
163259
Gene Name
DENN domain containing 2C
Gene Aliases
dJ1156J9.1
UniGene
Hs.654928
Gene Chromosome Location
Chr.1: 114584575-114670111 on Build GRCh38
Transcripts
NM_001256404.1 NM_198459.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3700462 c.2450T>G p.F817C Chr.1: 114587763-114587763 on Build GRCh38
5377269 c.2646G>A p.E882E Chr.1: 114587738-114587738 on Build GRCh38
6352528 c.2481G>A p.R827R Chr.1: 114587732-114587732 on Build GRCh38
3471591 c.2565C>T p.S855S Chr.1: 114587819-114587819 on Build GRCh38
4815725 c.2501C>G p.S834C Chr.1: 114587883-114587883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916257909 Chr.1: 114588078-114588078 on Build GRCh38
rs772940406 Chr.1: 114587687-114587687 on Build GRCh38
rs1026774082 Chr.1: 114588019-114588019 on Build GRCh38
rs61753529 Chr.1: 114587804-114587804 on Build GRCh38
rs1009171450 Chr.1: 114587937-114587937 on Build GRCh38
rs933573451 Chr.1: 114587766-114587766 on Build GRCh38
rs370366211 Chr.1: 114587803-114587803 on Build GRCh38
rs748828925 Chr.1: 114587851-114587851 on Build GRCh38
rs374705348 Chr.1: 114587860-114587860 on Build GRCh38
rs1015106181 Chr.1: 114587685-114587685 on Build GRCh38
rs61753528 Chr.1: 114587887-114587887 on Build GRCh38
rs745525937 Chr.1: 114587774-114587774 on Build GRCh38
rs962498223 Chr.1: 114587706-114587706 on Build GRCh38
rs764242432 Chr.1: 114587913-114587913 on Build GRCh38
rs199932240 Chr.1: 114587742-114587742 on Build GRCh38
rs775792132 Chr.1: 114587988-114587988 on Build GRCh38
rs777596116 Chr.1: 114587743-114587743 on Build GRCh38
rs530792029 Chr.1: 114587638-114587638 on Build GRCh38
rs767958216 Chr.1: 114587800-114587800 on Build GRCh38
rs113198986 Chr.1: 114588035-114588035 on Build GRCh38
rs755306674 Chr.1: 114587814-114587814 on Build GRCh38
rs768244184 Chr.1: 114587862-114587862 on Build GRCh38
rs769473985 Chr.1: 114587859-114587859 on Build GRCh38
rs770533792 Chr.1: 114587695-114587695 on Build GRCh38
rs760374222 Chr.1: 114587693-114587693 on Build GRCh38
rs756336880 Chr.1: 114587722-114587722 on Build GRCh38
rs762905349 Chr.1: 114587912-114587912 on Build GRCh38
rs58553623 Chr.1: 114587628-114587628 on Build GRCh38
rs114832136 Chr.1: 114587852-114587852 on Build GRCh38
rs35704052 Chr.1: 114588061-114588062 on Build GRCh38
rs775046012 Chr.1: 114587778-114587778 on Build GRCh38
rs776108395 Chr.1: 114587765-114587765 on Build GRCh38
rs777114249 Chr.1: 114587969-114587969 on Build GRCh38
rs745509521 Chr.1: 114587855-114587855 on Build GRCh38
rs756014000 Chr.1: 114587872-114587873 on Build GRCh38
rs762323987 Chr.1: 114587712-114587712 on Build GRCh38
rs954706464 Chr.1: 114588067-114588067 on Build GRCh38
rs773525086 Chr.1: 114587871-114587871 on Build GRCh38
rs763253985 Chr.1: 114587990-114587990 on Build GRCh38
rs778541405 Chr.1: 114587963-114587963 on Build GRCh38
rs61000990 Chr.1: 114587732-114587732 on Build GRCh38
rs1029288385 Chr.1: 114587629-114587629 on Build GRCh38
rs767992970 Chr.1: 114587948-114587948 on Build GRCh38
rs150205392 Chr.1: 114587999-114587999 on Build GRCh38
rs770984060 Chr.1: 114587734-114587734 on Build GRCh38
rs751398782 Chr.1: 114587922-114587922 on Build GRCh38
rs758998101 Chr.1: 114587702-114587702 on Build GRCh38
rs143167338 Chr.1: 114587747-114587747 on Build GRCh38
rs990442553 Chr.1: 114587755-114587755 on Build GRCh38
rs780932484 Chr.1: 114587926-114587926 on Build GRCh38
rs141389827 Chr.1: 114587772-114587772 on Build GRCh38
rs1037855563 Chr.1: 114587822-114587822 on Build GRCh38
rs754052667 Chr.1: 114587808-114587808 on Build GRCh38
rs748975071 Chr.1: 114587944-114587944 on Build GRCh38
rs746114439 Chr.1: 114587979-114587979 on Build GRCh38
rs970329776 Chr.1: 114588080-114588080 on Build GRCh38
rs780596735 Chr.1: 114587836-114587836 on Build GRCh38
rs138334687 Chr.1: 114587781-114587781 on Build GRCh38
rs771607945 Chr.1: 114587968-114587968 on Build GRCh38
rs779144796 Chr.1: 114587733-114587733 on Build GRCh38
rs949288783 Chr.1: 114587805-114587805 on Build GRCh38
rs747550063 Chr.1: 114587671-114587671 on Build GRCh38
rs779637959 Chr.1: 114587934-114587934 on Build GRCh38
rs377505952 Chr.1: 114587971-114587971 on Build GRCh38
rs761257163 Chr.1: 114587873-114587873 on Build GRCh38
rs749081007 Chr.1: 114587965-114587970 on Build GRCh38
rs748049188 Chr.1: 114587839-114587839 on Build GRCh38
rs766638332 Chr.1: 114587875-114587875 on Build GRCh38
rs913872085 Chr.1: 114587761-114587761 on Build GRCh38
rs762275216 Chr.1: 114587779-114587779 on Build GRCh38
rs775437829 Chr.1: 114587988-114587990 on Build GRCh38
rs750791589 Chr.1: 114587716-114587716 on Build GRCh38
rs776187438 Chr.1: 114587697-114587697 on Build GRCh38
rs777205807 Chr.1: 114587877-114587877 on Build GRCh38
rs140629217 Chr.1: 114588036-114588036 on Build GRCh38
rs770756101 Chr.1: 114587965-114587966 on Build GRCh38
rs767412626 Chr.1: 114588056-114588056 on Build GRCh38
rs765606004 Chr.1: 114587896-114587896 on Build GRCh38
rs747463997 Chr.1: 114587965-114587965 on Build GRCh38
rs752865230 Chr.1: 114587827-114587827 on Build GRCh38
rs566142081 Chr.1: 114587850-114587850 on Build GRCh38
rs375051143 Chr.1: 114587710-114587710 on Build GRCh38
rs200162285 Chr.1: 114587816-114587816 on Build GRCh38
rs759523112 Chr.1: 114588051-114588051 on Build GRCh38
rs770314062 Chr.1: 114587979-114587979 on Build GRCh38
rs777741996 Chr.1: 114587906-114587906 on Build GRCh38
rs750264272 Chr.1: 114587927-114587927 on Build GRCh38
rs898949267 Chr.1: 114587824-114587824 on Build GRCh38
rs774538973 Chr.1: 114587971-114587972 on Build GRCh38
rs960235602 Chr.1: 114587656-114587656 on Build GRCh38
rs752991556 Chr.1: 114587911-114587911 on Build GRCh38
rs780612754 Chr.1: 114587726-114587726 on Build GRCh38
rs769238779 Chr.1: 114587776-114587776 on Build GRCh38
rs552522640 Chr.1: 114587670-114587670 on Build GRCh38
rs374656477 Chr.1: 114587713-114587713 on Build GRCh38
rs748098478 Chr.1: 114587667-114587667 on Build GRCh38
rs144990095 Chr.1: 114587746-114587746 on Build GRCh38
rs769236355 Chr.1: 114587711-114587711 on Build GRCh38
rs1023237043 Chr.1: 114587679-114587679 on Build GRCh38
rs772151411 Chr.1: 114587979-114587980 on Build GRCh38
rs373812311 Chr.1: 114588055-114588055 on Build GRCh38
rs770781912 Chr.1: 114587764-114587764 on Build GRCh38

View Full Product Details