Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114717200-114717708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSDE1
Entrez Gene ID
7812
Gene Name
cold shock domain containing E1
Gene Aliases
D1S155E, UNR
UniGene
Hs.69855
Gene Chromosome Location
Chr.1: 114716913-114758050 on Build GRCh38
Transcripts
NM_001007553.2 NM_001130523.2 NM_001242891.1 NM_001242892.1 NM_001242893.1 NM_007158.5

SNP Detail

SNP ID Chromosome Location
rs893478100 Chr.1: 114717311-114717312 on Build GRCh38
rs199596172 Chr.1: 114717404-114717407 on Build GRCh38
rs982321654 Chr.1: 114717447-114717449 on Build GRCh38
rs371539180 Chr.1: 114717464-114717464 on Build GRCh38
rs1046251132 Chr.1: 114717296-114717296 on Build GRCh38
rs542289182 Chr.1: 114717503-114717503 on Build GRCh38
rs949238241 Chr.1: 114717288-114717288 on Build GRCh38
rs1050112139 Chr.1: 114717604-114717604 on Build GRCh38
rs1028740158 Chr.1: 114717325-114717325 on Build GRCh38
rs746214615 Chr.1: 114717272-114717273 on Build GRCh38
rs991487651 Chr.1: 114717399-114717400 on Build GRCh38
rs557719829 Chr.1: 114717312-114717312 on Build GRCh38
rs758716380 Chr.1: 114717285-114717285 on Build GRCh38
rs942396836 Chr.1: 114717334-114717334 on Build GRCh38
rs957652301 Chr.1: 114717564-114717564 on Build GRCh38
rs1009462604 Chr.1: 114717350-114717350 on Build GRCh38
rs980090099 Chr.1: 114717510-114717510 on Build GRCh38
rs913395670 Chr.1: 114717250-114717250 on Build GRCh38
rs553336786 Chr.1: 114717388-114717388 on Build GRCh38
rs767793974 Chr.1: 114717403-114717407 on Build GRCh38
rs1054581411 Chr.1: 114717247-114717247 on Build GRCh38
rs923816075 Chr.1: 114717506-114717506 on Build GRCh38
rs1050870324 Chr.1: 114717269-114717269 on Build GRCh38
rs61764384 Chr.1: 114717273-114717274 on Build GRCh38
rs1008660858 Chr.1: 114717633-114717637 on Build GRCh38
rs1013797229 Chr.1: 114717449-114717449 on Build GRCh38
rs755356304 Chr.1: 114717618-114717618 on Build GRCh38
rs917312461 Chr.1: 114717405-114717407 on Build GRCh38
rs374332374 Chr.1: 114717398-114717398 on Build GRCh38
rs1053711100 Chr.1: 114717537-114717537 on Build GRCh38
rs890250544 Chr.1: 114717337-114717337 on Build GRCh38
rs1014209439 Chr.1: 114717260-114717274 on Build GRCh38
rs373000705 Chr.1: 114717399-114717399 on Build GRCh38
rs370476917 Chr.1: 114717397-114717397 on Build GRCh38
rs79884428 Chr.1: 114717273-114717273 on Build GRCh38
rs902413837 Chr.1: 114717299-114717299 on Build GRCh38
rs762842125 Chr.1: 114717452-114717452 on Build GRCh38
rs1027574515 Chr.1: 114717398-114717399 on Build GRCh38
rs752020623 Chr.1: 114717434-114717434 on Build GRCh38
rs574875543 Chr.1: 114717483-114717483 on Build GRCh38
rs560558972 Chr.1: 114717390-114717390 on Build GRCh38
rs935205831 Chr.1: 114717513-114717513 on Build GRCh38
rs916382276 Chr.1: 114717276-114717276 on Build GRCh38
rs575288337 Chr.1: 114717232-114717235 on Build GRCh38
rs771596885 Chr.1: 114717386-114717386 on Build GRCh38
rs950189312 Chr.1: 114717412-114717421 on Build GRCh38
rs946245432 Chr.1: 114717622-114717622 on Build GRCh38
rs750470887 Chr.1: 114717263-114717263 on Build GRCh38
rs563584650 Chr.1: 114717650-114717650 on Build GRCh38
rs886942022 Chr.1: 114717447-114717447 on Build GRCh38
rs61764383 Chr.1: 114717320-114717320 on Build GRCh38
rs1034301199 Chr.1: 114717560-114717560 on Build GRCh38
rs890836948 Chr.1: 114717307-114717307 on Build GRCh38
rs541309766 Chr.1: 114717321-114717321 on Build GRCh38
rs577033305 Chr.1: 114717313-114717313 on Build GRCh38
rs913435872 Chr.1: 114717597-114717597 on Build GRCh38
rs558730735 Chr.1: 114717235-114717235 on Build GRCh38
rs1039364864 Chr.1: 114717365-114717365 on Build GRCh38
rs990360457 Chr.1: 114717574-114717574 on Build GRCh38
rs753534125 Chr.1: 114717372-114717372 on Build GRCh38
rs897998418 Chr.1: 114717663-114717663 on Build GRCh38

View Full Product Details