Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138949428-138949922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXL2NB
Entrez Gene ID
401089
Gene Name
FOXL2 neighbor
Gene Aliases
C3orf72
UniGene
Hs.591303
Gene Chromosome Location
Chr.3: 138947234-138953992 on Build GRCh38
Transcripts
NM_001040061.2 XM_005247443.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6616074 c.193G>A p.V65I Chr.3: 138949612-138949612 on Build GRCh38
6544875 c.137C>T p.A46V Chr.3: 138949556-138949556 on Build GRCh38
3124141 c.198G>A p.R66R Chr.3: 138949617-138949617 on Build GRCh38
6163514 c.203C>G p.S68W Chr.3: 138949622-138949622 on Build GRCh38
1566232 c.162C>T p.I54I Chr.3: 138949581-138949581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750013386 Chr.3: 138949721-138949721 on Build GRCh38
rs747995258 Chr.3: 138949651-138949651 on Build GRCh38
rs766564440 Chr.3: 138949573-138949573 on Build GRCh38
rs768195932 Chr.3: 138949651-138949651 on Build GRCh38
rs201016461 Chr.3: 138949687-138949687 on Build GRCh38
rs112302431 Chr.3: 138949897-138949897 on Build GRCh38
rs374065217 Chr.3: 138949774-138949774 on Build GRCh38
rs756651663 Chr.3: 138949476-138949476 on Build GRCh38
rs776429015 Chr.3: 138949542-138949542 on Build GRCh38
rs766136474 Chr.3: 138949729-138949729 on Build GRCh38
rs770937890 Chr.3: 138949812-138949812 on Build GRCh38
rs748265780 Chr.3: 138949557-138949557 on Build GRCh38
rs866147006 Chr.3: 138949891-138949891 on Build GRCh38
rs761404406 Chr.3: 138949593-138949593 on Build GRCh38
rs550495513 Chr.3: 138949794-138949794 on Build GRCh38
rs975328176 Chr.3: 138949783-138949783 on Build GRCh38
rs904842716 Chr.3: 138949866-138949866 on Build GRCh38
rs753452514 Chr.3: 138949469-138949469 on Build GRCh38
rs1046823841 Chr.3: 138949565-138949565 on Build GRCh38
rs911684255 Chr.3: 138949459-138949459 on Build GRCh38
rs776150457 Chr.3: 138949747-138949747 on Build GRCh38
rs908325624 Chr.3: 138949836-138949836 on Build GRCh38
rs761130747 Chr.3: 138949696-138949696 on Build GRCh38
rs528584803 Chr.3: 138949570-138949570 on Build GRCh38
rs72976938 Chr.3: 138949558-138949558 on Build GRCh38
rs774486116 Chr.3: 138949688-138949688 on Build GRCh38
rs780888897 Chr.3: 138949770-138949770 on Build GRCh38
rs938273576 Chr.3: 138949871-138949871 on Build GRCh38
rs781202664 Chr.3: 138949642-138949642 on Build GRCh38
rs913572780 Chr.3: 138949508-138949508 on Build GRCh38
rs61408596 Chr.3: 138949850-138949850 on Build GRCh38
rs67626684 Chr.3: 138949740-138949740 on Build GRCh38
rs539321668 Chr.3: 138949880-138949880 on Build GRCh38
rs925909099 Chr.3: 138949489-138949489 on Build GRCh38
rs865982825 Chr.3: 138949556-138949556 on Build GRCh38
rs752514376 Chr.3: 138949771-138949771 on Build GRCh38
rs376474965 Chr.3: 138949587-138949587 on Build GRCh38
rs979443657 Chr.3: 138949722-138949722 on Build GRCh38
rs895868983 Chr.3: 138949652-138949652 on Build GRCh38
rs754843453 Chr.3: 138949759-138949759 on Build GRCh38
rs746781350 Chr.3: 138949517-138949517 on Build GRCh38
rs142253798 Chr.3: 138949667-138949667 on Build GRCh38
rs753084245 Chr.3: 138949589-138949589 on Build GRCh38
rs964335801 Chr.3: 138949753-138949753 on Build GRCh38
rs779126788 Chr.3: 138949816-138949816 on Build GRCh38
rs141282655 Chr.3: 138949616-138949625 on Build GRCh38
rs749372722 Chr.3: 138949798-138949798 on Build GRCh38
rs146404966 Chr.3: 138949887-138949887 on Build GRCh38
rs746225281 Chr.3: 138949484-138949484 on Build GRCh38
rs768774323 Chr.3: 138949532-138949532 on Build GRCh38
rs371549160 Chr.3: 138949867-138949867 on Build GRCh38
rs377658501 Chr.3: 138949564-138949564 on Build GRCh38
rs756026183 Chr.3: 138949582-138949582 on Build GRCh38
rs905481342 Chr.3: 138949609-138949609 on Build GRCh38
rs764643142 Chr.3: 138949705-138949705 on Build GRCh38
rs749921652 Chr.3: 138949598-138949598 on Build GRCh38
rs201726891 Chr.3: 138949495-138949495 on Build GRCh38
rs776099781 Chr.3: 138949694-138949694 on Build GRCh38
rs759903567 Chr.3: 138949689-138949689 on Build GRCh38
rs762550107 Chr.3: 138949726-138949726 on Build GRCh38
rs754753810 Chr.3: 138949623-138949623 on Build GRCh38
rs747049760 Chr.3: 138949478-138949478 on Build GRCh38
rs758177237 Chr.3: 138949599-138949599 on Build GRCh38
rs767995983 Chr.3: 138949463-138949463 on Build GRCh38
rs1020932563 Chr.3: 138949460-138949460 on Build GRCh38
rs764616976 Chr.3: 138949594-138949594 on Build GRCh38
rs559748613 Chr.3: 138949563-138949563 on Build GRCh38
rs987869293 Chr.3: 138949890-138949890 on Build GRCh38
rs1038034094 Chr.3: 138949603-138949603 on Build GRCh38
rs372389376 Chr.3: 138949877-138949877 on Build GRCh38
rs367754243 Chr.3: 138949661-138949661 on Build GRCh38
rs756062895 Chr.3: 138949793-138949793 on Build GRCh38
rs779841304 Chr.3: 138949605-138949605 on Build GRCh38
rs868781288 Chr.3: 138949637-138949637 on Build GRCh38
rs367968677 Chr.3: 138949507-138949507 on Build GRCh38
rs745912128 Chr.3: 138949590-138949591 on Build GRCh38
rs769739254 Chr.3: 138949655-138949655 on Build GRCh38
rs759887316 Chr.3: 138949586-138949586 on Build GRCh38
rs772271367 Chr.3: 138949691-138949691 on Build GRCh38
rs941085415 Chr.3: 138949846-138949846 on Build GRCh38
rs926438631 Chr.3: 138949574-138949574 on Build GRCh38
rs746653393 Chr.3: 138949630-138949630 on Build GRCh38
rs751468798 Chr.3: 138949619-138949619 on Build GRCh38
rs868263406 Chr.3: 138949815-138949815 on Build GRCh38

View Full Product Details