Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 140403337-140403840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLSTN2
Entrez Gene ID
64084
Gene Name
calsyntenin 2
Gene Aliases
ALC-GAMMA, CDHR13, CS2, CSTN2, alcagamma
UniGene
Hs.158529
Gene Chromosome Location
Chr.3: 139935185-140577397 on Build GRCh38
Transcripts
NM_022131.2 XM_017007022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6630751 c.428+1G>A p.? Chr.3: 140403825-140403825 on Build GRCh38
3588106 c.319C>T p.R107C Chr.3: 140403715-140403715 on Build GRCh38
1419496 c.378C>A p.A126A Chr.3: 140403774-140403774 on Build GRCh38
3588107 c.329G>A p.S110N Chr.3: 140403725-140403725 on Build GRCh38
5887804 c.233G>A p.G78E Chr.3: 140403629-140403629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371820039 Chr.3: 140403799-140403799 on Build GRCh38
rs375459786 Chr.3: 140403588-140403588 on Build GRCh38
rs1001542964 Chr.3: 140403456-140403456 on Build GRCh38
rs41265457 Chr.3: 140403580-140403580 on Build GRCh38
rs746385044 Chr.3: 140403620-140403620 on Build GRCh38
rs773081653 Chr.3: 140403579-140403580 on Build GRCh38
rs112339783 Chr.3: 140403609-140403609 on Build GRCh38
rs975254304 Chr.3: 140403708-140403708 on Build GRCh38
rs76323990 Chr.3: 140403372-140403372 on Build GRCh38
rs145666342 Chr.3: 140403716-140403716 on Build GRCh38
rs982965308 Chr.3: 140403751-140403751 on Build GRCh38
rs773769106 Chr.3: 140403776-140403776 on Build GRCh38
rs749052371 Chr.3: 140403736-140403736 on Build GRCh38
rs750342561 Chr.3: 140403809-140403809 on Build GRCh38
rs989512265 Chr.3: 140403439-140403439 on Build GRCh38
rs1051979987 Chr.3: 140403393-140403393 on Build GRCh38
rs753697687 Chr.3: 140403804-140403804 on Build GRCh38
rs1037916848 Chr.3: 140403783-140403783 on Build GRCh38
rs570060023 Chr.3: 140403729-140403729 on Build GRCh38
rs772186079 Chr.3: 140403746-140403746 on Build GRCh38
rs774326567 Chr.3: 140403682-140403682 on Build GRCh38
rs777898955 Chr.3: 140403674-140403674 on Build GRCh38
rs1005583741 Chr.3: 140403395-140403395 on Build GRCh38
rs767033379 Chr.3: 140403790-140403790 on Build GRCh38
rs778851963 Chr.3: 140403744-140403744 on Build GRCh38
rs765303330 Chr.3: 140403495-140403495 on Build GRCh38
rs959990248 Chr.3: 140403480-140403480 on Build GRCh38
rs112178546 Chr.3: 140403377-140403377 on Build GRCh38
rs992795786 Chr.3: 140403491-140403491 on Build GRCh38
rs142775951 Chr.3: 140403642-140403642 on Build GRCh38
rs747956181 Chr.3: 140403653-140403653 on Build GRCh38
rs751047596 Chr.3: 140403718-140403718 on Build GRCh38
rs755971554 Chr.3: 140403656-140403656 on Build GRCh38
rs770943686 Chr.3: 140403681-140403681 on Build GRCh38
rs753006773 Chr.3: 140403583-140403583 on Build GRCh38
rs745992339 Chr.3: 140403745-140403745 on Build GRCh38
rs4683493 Chr.3: 140403375-140403375 on Build GRCh38
rs1038371209 Chr.3: 140403556-140403556 on Build GRCh38
rs745893021 Chr.3: 140403693-140403693 on Build GRCh38
rs757587077 Chr.3: 140403572-140403572 on Build GRCh38
rs4683494 Chr.3: 140403499-140403499 on Build GRCh38
rs757148060 Chr.3: 140403741-140403741 on Build GRCh38
rs141317662 Chr.3: 140403753-140403753 on Build GRCh38
rs539832962 Chr.3: 140403709-140403709 on Build GRCh38
rs754501382 Chr.3: 140403727-140403727 on Build GRCh38
rs765781690 Chr.3: 140403616-140403616 on Build GRCh38
rs945617347 Chr.3: 140403464-140403464 on Build GRCh38
rs967318601 Chr.3: 140403501-140403501 on Build GRCh38
rs377236270 Chr.3: 140403806-140403806 on Build GRCh38
rs375333353 Chr.3: 140403710-140403710 on Build GRCh38
rs192364847 Chr.3: 140403538-140403538 on Build GRCh38
rs368696603 Chr.3: 140403798-140403798 on Build GRCh38
rs529034321 Chr.3: 140403641-140403641 on Build GRCh38
rs558249584 Chr.3: 140403432-140403432 on Build GRCh38
rs764641824 Chr.3: 140403594-140403594 on Build GRCh38
rs979110860 Chr.3: 140403502-140403502 on Build GRCh38
rs368785435 Chr.3: 140403457-140403457 on Build GRCh38
rs937947866 Chr.3: 140403662-140403662 on Build GRCh38
rs72981079 Chr.3: 140403621-140403621 on Build GRCh38
rs906371315 Chr.3: 140403552-140403552 on Build GRCh38
rs767319582 Chr.3: 140403383-140403383 on Build GRCh38
rs899356720 Chr.3: 140403453-140403453 on Build GRCh38
rs963887055 Chr.3: 140403513-140403513 on Build GRCh38
rs1011288697 Chr.3: 140403582-140403582 on Build GRCh38
rs565804116 Chr.3: 140403695-140403695 on Build GRCh38
rs749218371 Chr.3: 140403675-140403675 on Build GRCh38
rs1046760173 Chr.3: 140403535-140403535 on Build GRCh38
rs775766435 Chr.3: 140403696-140403696 on Build GRCh38
rs777898266 Chr.3: 140403534-140403534 on Build GRCh38
rs757061676 Chr.3: 140403805-140403805 on Build GRCh38
rs150165907 Chr.3: 140403715-140403715 on Build GRCh38
rs750347251 Chr.3: 140403394-140403394 on Build GRCh38
rs759011249 Chr.3: 140403787-140403787 on Build GRCh38
rs1014331442 Chr.3: 140403607-140403607 on Build GRCh38
rs980822493 Chr.3: 140403392-140403392 on Build GRCh38
rs768815689 Chr.3: 140403759-140403759 on Build GRCh38
rs74507604 Chr.3: 140403768-140403768 on Build GRCh38
rs775613715 Chr.3: 140403750-140403750 on Build GRCh38
rs76430510 Chr.3: 140403374-140403374 on Build GRCh38
rs777006836 Chr.3: 140403763-140403763 on Build GRCh38
rs541780328 Chr.3: 140403516-140403516 on Build GRCh38
rs138816883 Chr.3: 140403553-140403553 on Build GRCh38
rs559773847 Chr.3: 140403517-140403517 on Build GRCh38
rs777735378 Chr.3: 140403731-140403731 on Build GRCh38
rs145972974 Chr.3: 140403498-140403498 on Build GRCh38
rs200193660 Chr.3: 140403775-140403775 on Build GRCh38
rs780698105 Chr.3: 140403712-140403712 on Build GRCh38
rs574272572 Chr.3: 140403506-140403506 on Build GRCh38
rs564213262 Chr.3: 140403570-140403570 on Build GRCh38

View Full Product Details