Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 140403733-140404245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLSTN2
Entrez Gene ID
64084
Gene Name
calsyntenin 2
Gene Aliases
ALC-GAMMA, CDHR13, CS2, CSTN2, alcagamma
UniGene
Hs.158529
Gene Chromosome Location
Chr.3: 139935185-140577397 on Build GRCh38
Transcripts
NM_022131.2 XM_017007022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6630751 c.428+1G>A p.? Chr.3: 140403825-140403825 on Build GRCh38
212386 c.358T>G p.Y120D Chr.3: 140403754-140403754 on Build GRCh38
1419496 c.378C>A p.A126A Chr.3: 140403774-140403774 on Build GRCh38
5692836 c.345G>T p.E115D Chr.3: 140403741-140403741 on Build GRCh38
5615211 c.373C>G p.Q125E Chr.3: 140403769-140403769 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371820039 Chr.3: 140403799-140403799 on Build GRCh38
rs758454234 Chr.3: 140403826-140403826 on Build GRCh38
rs554196122 Chr.3: 140403989-140403989 on Build GRCh38
rs950283118 Chr.3: 140404029-140404029 on Build GRCh38
rs982526611 Chr.3: 140403950-140403950 on Build GRCh38
rs780128193 Chr.3: 140403827-140403827 on Build GRCh38
rs188137820 Chr.3: 140403852-140403852 on Build GRCh38
rs983881722 Chr.3: 140404112-140404112 on Build GRCh38
rs561420910 Chr.3: 140403972-140403972 on Build GRCh38
rs773769106 Chr.3: 140403776-140403776 on Build GRCh38
rs115217213 Chr.3: 140404150-140404150 on Build GRCh38
rs750342561 Chr.3: 140403809-140403809 on Build GRCh38
rs111820376 Chr.3: 140403992-140403992 on Build GRCh38
rs753697687 Chr.3: 140403804-140403804 on Build GRCh38
rs1037916848 Chr.3: 140403783-140403783 on Build GRCh38
rs770166493 Chr.3: 140403853-140403853 on Build GRCh38
rs1054972596 Chr.3: 140403889-140403889 on Build GRCh38
rs534712233 Chr.3: 140403926-140403926 on Build GRCh38
rs975283158 Chr.3: 140404041-140404041 on Build GRCh38
rs767033379 Chr.3: 140403790-140403790 on Build GRCh38
rs199793058 Chr.3: 140403864-140403864 on Build GRCh38
rs753770057 Chr.3: 140403952-140403952 on Build GRCh38
rs772922765 Chr.3: 140403899-140403899 on Build GRCh38
rs771791174 Chr.3: 140403840-140403840 on Build GRCh38
rs1055489988 Chr.3: 140404208-140404208 on Build GRCh38
rs760617131 Chr.3: 140403910-140403910 on Build GRCh38
rs538580855 Chr.3: 140403866-140403866 on Build GRCh38
rs952353701 Chr.3: 140404109-140404109 on Build GRCh38
rs937944522 Chr.3: 140404190-140404190 on Build GRCh38
rs184512372 Chr.3: 140404001-140404001 on Build GRCh38
rs137940141 Chr.3: 140404067-140404067 on Build GRCh38
rs908331696 Chr.3: 140404113-140404113 on Build GRCh38
rs181031528 Chr.3: 140403915-140403915 on Build GRCh38
rs1041766864 Chr.3: 140404063-140404063 on Build GRCh38
rs146951102 Chr.3: 140403943-140403943 on Build GRCh38
rs529070583 Chr.3: 140403973-140403973 on Build GRCh38
rs917387765 Chr.3: 140404000-140404000 on Build GRCh38
rs377236270 Chr.3: 140403806-140403806 on Build GRCh38
rs575617426 Chr.3: 140403958-140403958 on Build GRCh38
rs753534831 Chr.3: 140403869-140403869 on Build GRCh38
rs537419313 Chr.3: 140404140-140404140 on Build GRCh38
rs578152744 Chr.3: 140403905-140403905 on Build GRCh38
rs368696603 Chr.3: 140403798-140403798 on Build GRCh38
rs376362083 Chr.3: 140403865-140403865 on Build GRCh38
rs927850294 Chr.3: 140404159-140404159 on Build GRCh38
rs771304564 Chr.3: 140403859-140403859 on Build GRCh38
rs996735149 Chr.3: 140404149-140404149 on Build GRCh38
rs774155448 Chr.3: 140403934-140403934 on Build GRCh38
rs920777641 Chr.3: 140404106-140404106 on Build GRCh38
rs368435868 Chr.3: 140403987-140403987 on Build GRCh38
rs757061676 Chr.3: 140403805-140403805 on Build GRCh38
rs184823312 Chr.3: 140403836-140403836 on Build GRCh38
rs1054863575 Chr.3: 140404127-140404127 on Build GRCh38
rs763539458 Chr.3: 140403854-140403854 on Build GRCh38
rs543143873 Chr.3: 140403962-140403962 on Build GRCh38
rs759011249 Chr.3: 140403787-140403787 on Build GRCh38
rs768866966 Chr.3: 140403846-140403846 on Build GRCh38
rs558702153 Chr.3: 140403900-140403900 on Build GRCh38
rs762597835 Chr.3: 140403853-140403853 on Build GRCh38
rs189381889 Chr.3: 140404053-140404053 on Build GRCh38
rs768815689 Chr.3: 140403759-140403759 on Build GRCh38
rs74507604 Chr.3: 140403768-140403768 on Build GRCh38
rs777006836 Chr.3: 140403763-140403763 on Build GRCh38
rs1000608487 Chr.3: 140403877-140403877 on Build GRCh38
rs369452576 Chr.3: 140403918-140403918 on Build GRCh38
rs866235966 Chr.3: 140404114-140404114 on Build GRCh38
rs200193660 Chr.3: 140403775-140403775 on Build GRCh38
rs370042302 Chr.3: 140403847-140403847 on Build GRCh38
rs751763262 Chr.3: 140404189-140404189 on Build GRCh38

View Full Product Details