Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 140404466-140404962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLSTN2
Entrez Gene ID
64084
Gene Name
calsyntenin 2
Gene Aliases
ALC-GAMMA, CDHR13, CS2, CSTN2, alcagamma
UniGene
Hs.158529
Gene Chromosome Location
Chr.3: 139935185-140577397 on Build GRCh38
Transcripts
NM_022131.2 XM_017007022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
728129 c.548C>T p.A183V Chr.3: 140404677-140404677 on Build GRCh38
5599312 c.627C>T p.I209I Chr.3: 140404756-140404756 on Build GRCh38
728131 c.547G>T p.A183S Chr.3: 140404676-140404676 on Build GRCh38
3588108 c.438C>T p.V146V Chr.3: 140404567-140404567 on Build GRCh38
1248480 c.585C>A p.I195I Chr.3: 140404714-140404714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190766550 Chr.3: 140404840-140404840 on Build GRCh38
rs181844087 Chr.3: 140404906-140404906 on Build GRCh38
rs756363468 Chr.3: 140404651-140404651 on Build GRCh38
rs143019314 Chr.3: 140404819-140404819 on Build GRCh38
rs141176953 Chr.3: 140404639-140404639 on Build GRCh38
rs369597834 Chr.3: 140404529-140404529 on Build GRCh38
rs200729250 Chr.3: 140404591-140404591 on Build GRCh38
rs769497223 Chr.3: 140404727-140404727 on Build GRCh38
rs113602686 Chr.3: 140404756-140404756 on Build GRCh38
rs201089535 Chr.3: 140404510-140404510 on Build GRCh38
rs753973968 Chr.3: 140404768-140404768 on Build GRCh38
rs748384459 Chr.3: 140404561-140404561 on Build GRCh38
rs921355677 Chr.3: 140404745-140404745 on Build GRCh38
rs756136789 Chr.3: 140404757-140404757 on Build GRCh38
rs779601547 Chr.3: 140404685-140404685 on Build GRCh38
rs554988446 Chr.3: 140404835-140404835 on Build GRCh38
rs769158380 Chr.3: 140404805-140404805 on Build GRCh38
rs527836895 Chr.3: 140404850-140404850 on Build GRCh38
rs749551523 Chr.3: 140404583-140404583 on Build GRCh38
rs762594115 Chr.3: 140404628-140404628 on Build GRCh38
rs888036462 Chr.3: 140404630-140404630 on Build GRCh38
rs774984260 Chr.3: 140404592-140404592 on Build GRCh38
rs750132688 Chr.3: 140404526-140404526 on Build GRCh38
rs148587237 Chr.3: 140404684-140404684 on Build GRCh38
rs766373799 Chr.3: 140404638-140404638 on Build GRCh38
rs767610788 Chr.3: 140404645-140404645 on Build GRCh38
rs762779163 Chr.3: 140404730-140404730 on Build GRCh38
rs772657107 Chr.3: 140404719-140404719 on Build GRCh38
rs199631846 Chr.3: 140404641-140404641 on Build GRCh38
rs746191083 Chr.3: 140404786-140404786 on Build GRCh38
rs372597527 Chr.3: 140404814-140404814 on Build GRCh38
rs977728342 Chr.3: 140404675-140404675 on Build GRCh38
rs751648063 Chr.3: 140404544-140404544 on Build GRCh38
rs143858966 Chr.3: 140404614-140404614 on Build GRCh38
rs528619654 Chr.3: 140404598-140404598 on Build GRCh38
rs376407632 Chr.3: 140404815-140404815 on Build GRCh38
rs764442743 Chr.3: 140404765-140404765 on Build GRCh38
rs772969233 Chr.3: 140404809-140404809 on Build GRCh38
rs757744287 Chr.3: 140404771-140404771 on Build GRCh38
rs150691200 Chr.3: 140404653-140404653 on Build GRCh38
rs758387487 Chr.3: 140404528-140404528 on Build GRCh38
rs752679740 Chr.3: 140404648-140404648 on Build GRCh38
rs761552233 Chr.3: 140404509-140404509 on Build GRCh38
rs760327452 Chr.3: 140404597-140404597 on Build GRCh38
rs781435115 Chr.3: 140404559-140404559 on Build GRCh38
rs747748437 Chr.3: 140404801-140404801 on Build GRCh38
rs143006732 Chr.3: 140404611-140404611 on Build GRCh38
rs370132679 Chr.3: 140404794-140404794 on Build GRCh38
rs111391920 Chr.3: 140404747-140404747 on Build GRCh38
rs779441993 Chr.3: 140404785-140404785 on Build GRCh38
rs376084850 Chr.3: 140404735-140404735 on Build GRCh38
rs142111816 Chr.3: 140404696-140404696 on Build GRCh38
rs557248751 Chr.3: 140404734-140404734 on Build GRCh38
rs778232368 Chr.3: 140404570-140404570 on Build GRCh38
rs749721669 Chr.3: 140404668-140404668 on Build GRCh38
rs1050964638 Chr.3: 140404501-140404501 on Build GRCh38
rs972056662 Chr.3: 140404670-140404670 on Build GRCh38
rs915165888 Chr.3: 140404486-140404486 on Build GRCh38
rs146135578 Chr.3: 140404861-140404861 on Build GRCh38
rs530942353 Chr.3: 140404493-140404493 on Build GRCh38
rs891028198 Chr.3: 140404505-140404505 on Build GRCh38
rs755049824 Chr.3: 140404548-140404548 on Build GRCh38
rs759504921 Chr.3: 140404739-140404739 on Build GRCh38
rs137984856 Chr.3: 140404599-140404599 on Build GRCh38
rs907366666 Chr.3: 140404903-140404903 on Build GRCh38
rs772723832 Chr.3: 140404729-140404729 on Build GRCh38
rs779897798 Chr.3: 140404533-140404533 on Build GRCh38
rs866605205 Chr.3: 140404865-140404865 on Build GRCh38
rs772837759 Chr.3: 140404885-140404885 on Build GRCh38
rs777801919 Chr.3: 140404660-140404660 on Build GRCh38
rs769862663 Chr.3: 140404562-140404562 on Build GRCh38
rs59949679 Chr.3: 140404876-140404876 on Build GRCh38
rs1024591740 Chr.3: 140404884-140404884 on Build GRCh38
rs772422791 Chr.3: 140404528-140404528 on Build GRCh38
rs775847202 Chr.3: 140404778-140404779 on Build GRCh38
rs1003018609 Chr.3: 140404907-140404907 on Build GRCh38
rs747541327 Chr.3: 140404725-140404725 on Build GRCh38
rs577782961 Chr.3: 140404894-140404894 on Build GRCh38
rs143541782 Chr.3: 140404609-140404609 on Build GRCh38

View Full Product Details