Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 141234799-141235309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PXYLP1
Entrez Gene ID
92370
Gene Name
2-phosphoxylose phosphatase 1
Gene Aliases
ACPL2, HEL124, XYLP
UniGene
Hs.657887
Gene Chromosome Location
Chr.3: 141231825-141294903 on Build GRCh38
Transcripts
NM_001037172.2 NM_001282728.1 NM_152282.4 XM_017007511.1

SNP Detail

SNP ID Chromosome Location
rs550024188 Chr.3: 141235271-141235271 on Build GRCh38
rs1025454555 Chr.3: 141234880-141234880 on Build GRCh38
rs979405715 Chr.3: 141235189-141235189 on Build GRCh38
rs968918808 Chr.3: 141235187-141235187 on Build GRCh38
rs933365877 Chr.3: 141235116-141235116 on Build GRCh38
rs185829479 Chr.3: 141234902-141234902 on Build GRCh38
rs1043200961 Chr.3: 141234824-141234824 on Build GRCh38
rs1013253847 Chr.3: 141234951-141234951 on Build GRCh38
rs745725373 Chr.3: 141235212-141235212 on Build GRCh38
rs543993234 Chr.3: 141234998-141234998 on Build GRCh38
rs994083828 Chr.3: 141235250-141235250 on Build GRCh38
rs905083897 Chr.3: 141234997-141234997 on Build GRCh38
rs962172682 Chr.3: 141235040-141235040 on Build GRCh38
rs568504633 Chr.3: 141235274-141235274 on Build GRCh38
rs768970817 Chr.3: 141235252-141235252 on Build GRCh38
rs1052603243 Chr.3: 141235042-141235042 on Build GRCh38
rs548159349 Chr.3: 141235118-141235118 on Build GRCh38
rs993946421 Chr.3: 141234871-141234871 on Build GRCh38
rs916194847 Chr.3: 141234992-141234992 on Build GRCh38
rs954796266 Chr.3: 141235136-141235136 on Build GRCh38
rs2292400 Chr.3: 141235056-141235056 on Build GRCh38
rs910565025 Chr.3: 141235168-141235168 on Build GRCh38
rs538019433 Chr.3: 141235245-141235245 on Build GRCh38
rs1052598117 Chr.3: 141234885-141234885 on Build GRCh38
rs934815305 Chr.3: 141235233-141235233 on Build GRCh38
rs1037692517 Chr.3: 141235165-141235165 on Build GRCh38
rs780872515 Chr.3: 141235193-141235193 on Build GRCh38
rs774783644 Chr.3: 141235272-141235272 on Build GRCh38
rs560185154 Chr.3: 141235127-141235127 on Build GRCh38
rs564380758 Chr.3: 141234848-141234848 on Build GRCh38
rs924792072 Chr.3: 141234840-141234840 on Build GRCh38
rs925512397 Chr.3: 141235003-141235003 on Build GRCh38
rs552482116 Chr.3: 141235150-141235150 on Build GRCh38
rs1044760827 Chr.3: 141234953-141234953 on Build GRCh38
rs78793891 Chr.3: 141235036-141235036 on Build GRCh38
rs139726058 Chr.3: 141235143-141235143 on Build GRCh38
rs969101547 Chr.3: 141234994-141234994 on Build GRCh38
rs190245620 Chr.3: 141235240-141235240 on Build GRCh38

View Full Product Details