Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 142158247-142158767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GK5
Entrez Gene ID
256356
Gene Name
glycerol kinase 5 (putative)
Gene Aliases
-
UniGene
Hs.135904
Gene Chromosome Location
Chr.3: 142157527-142225607 on Build GRCh38
Transcripts
NM_001039547.2 NR_033289.1 XM_011512624.2 XM_017006089.1 XM_017006090.1

SNP Detail

SNP ID Chromosome Location
rs1054255049 Chr.3: 142158453-142158453 on Build GRCh38
rs907258566 Chr.3: 142158707-142158707 on Build GRCh38
rs1024825026 Chr.3: 142158345-142158345 on Build GRCh38
rs758279405 Chr.3: 142158571-142158571 on Build GRCh38
rs532445890 Chr.3: 142158653-142158653 on Build GRCh38
rs115930521 Chr.3: 142158522-142158522 on Build GRCh38
rs1034307896 Chr.3: 142158721-142158721 on Build GRCh38
rs965612661 Chr.3: 142158733-142158733 on Build GRCh38
rs995370993 Chr.3: 142158673-142158673 on Build GRCh38
rs374620066 Chr.3: 142158687-142158687 on Build GRCh38
rs912807175 Chr.3: 142158383-142158383 on Build GRCh38
rs1002778975 Chr.3: 142158618-142158618 on Build GRCh38
rs951005086 Chr.3: 142158275-142158275 on Build GRCh38
rs181197944 Chr.3: 142158333-142158333 on Build GRCh38
rs946261565 Chr.3: 142158454-142158454 on Build GRCh38
rs886927699 Chr.3: 142158656-142158656 on Build GRCh38
rs982460466 Chr.3: 142158366-142158366 on Build GRCh38
rs1025248743 Chr.3: 142158703-142158703 on Build GRCh38
rs1015037445 Chr.3: 142158289-142158289 on Build GRCh38
rs11707933 Chr.3: 142158520-142158520 on Build GRCh38
rs1039350176 Chr.3: 142158540-142158540 on Build GRCh38
rs144025140 Chr.3: 142158727-142158727 on Build GRCh38
rs866313069 Chr.3: 142158498-142158498 on Build GRCh38
rs981385325 Chr.3: 142158327-142158327 on Build GRCh38
rs188717524 Chr.3: 142158309-142158309 on Build GRCh38
rs535976183 Chr.3: 142158284-142158284 on Build GRCh38
rs550641339 Chr.3: 142158710-142158710 on Build GRCh38
rs527764968 Chr.3: 142158481-142158481 on Build GRCh38
rs560241570 Chr.3: 142158457-142158457 on Build GRCh38
rs374229447 Chr.3: 142158461-142158461 on Build GRCh38
rs1027121761 Chr.3: 142158688-142158688 on Build GRCh38
rs1039871609 Chr.3: 142158423-142158423 on Build GRCh38
rs762541416 Chr.3: 142158709-142158709 on Build GRCh38
rs1019331913 Chr.3: 142158732-142158732 on Build GRCh38
rs754644664 Chr.3: 142158530-142158531 on Build GRCh38
rs1034316136 Chr.3: 142158348-142158348 on Build GRCh38
rs151040990 Chr.3: 142158621-142158621 on Build GRCh38
rs914724182 Chr.3: 142158451-142158451 on Build GRCh38
rs936855491 Chr.3: 142158445-142158445 on Build GRCh38
rs116417634 Chr.3: 142158419-142158419 on Build GRCh38
rs1034279002 Chr.3: 142158651-142158651 on Build GRCh38
rs907125924 Chr.3: 142158607-142158607 on Build GRCh38
rs921396916 Chr.3: 142158441-142158441 on Build GRCh38
rs73236059 Chr.3: 142158565-142158565 on Build GRCh38
rs147320842 Chr.3: 142158740-142158740 on Build GRCh38
rs1046404185 Chr.3: 142158575-142158575 on Build GRCh38
rs977562574 Chr.3: 142158466-142158466 on Build GRCh38
rs143932924 Chr.3: 142158453-142158453 on Build GRCh38
rs138494020 Chr.3: 142158315-142158315 on Build GRCh38

View Full Product Details