Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 142163856-142164396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GK5
Entrez Gene ID
256356
Gene Name
glycerol kinase 5 (putative)
Gene Aliases
-
UniGene
Hs.135904
Gene Chromosome Location
Chr.3: 142157527-142225607 on Build GRCh38
Transcripts
NM_001039547.2 NR_033289.1 XM_011512624.2 XM_017006089.1 XM_017006090.1

SNP Detail

SNP ID Chromosome Location
rs543257379 Chr.3: 142164188-142164188 on Build GRCh38
rs891091787 Chr.3: 142164080-142164080 on Build GRCh38
rs571118244 Chr.3: 142163971-142163971 on Build GRCh38
rs987435757 Chr.3: 142163943-142163943 on Build GRCh38
rs760241328 Chr.3: 142164305-142164305 on Build GRCh38
rs911932838 Chr.3: 142163969-142163970 on Build GRCh38
rs11714396 Chr.3: 142164189-142164189 on Build GRCh38
rs927593383 Chr.3: 142163944-142163944 on Build GRCh38
rs971454798 Chr.3: 142164240-142164240 on Build GRCh38
rs538621034 Chr.3: 142164070-142164070 on Build GRCh38
rs949834846 Chr.3: 142164124-142164124 on Build GRCh38
rs148199274 Chr.3: 142164213-142164213 on Build GRCh38
rs1047905531 Chr.3: 142164105-142164105 on Build GRCh38
rs141408237 Chr.3: 142164031-142164031 on Build GRCh38
rs898580026 Chr.3: 142163986-142163986 on Build GRCh38
rs995956447 Chr.3: 142164160-142164160 on Build GRCh38
rs762247604 Chr.3: 142164115-142164115 on Build GRCh38
rs930801140 Chr.3: 142164058-142164058 on Build GRCh38
rs1008246745 Chr.3: 142164130-142164130 on Build GRCh38
rs570789959 Chr.3: 142164002-142164002 on Build GRCh38
rs1020138335 Chr.3: 142164276-142164276 on Build GRCh38
rs966777836 Chr.3: 142164288-142164288 on Build GRCh38
rs556828751 Chr.3: 142164099-142164099 on Build GRCh38
rs893061663 Chr.3: 142164206-142164206 on Build GRCh38
rs368604839 Chr.3: 142164280-142164280 on Build GRCh38
rs1032844237 Chr.3: 142164178-142164178 on Build GRCh38
rs1047150823 Chr.3: 142164055-142164055 on Build GRCh38
rs563405205 Chr.3: 142164072-142164074 on Build GRCh38
rs994215186 Chr.3: 142164026-142164026 on Build GRCh38
rs765752954 Chr.3: 142164322-142164322 on Build GRCh38
rs114991710 Chr.3: 142164155-142164155 on Build GRCh38
rs966787679 Chr.3: 142163899-142163899 on Build GRCh38
rs964772163 Chr.3: 142164359-142164359 on Build GRCh38
rs1019259161 Chr.3: 142164138-142164138 on Build GRCh38
rs927530287 Chr.3: 142164291-142164291 on Build GRCh38
rs1021026523 Chr.3: 142163890-142163890 on Build GRCh38
rs1020053729 Chr.3: 142164214-142164214 on Build GRCh38
rs1038583350 Chr.3: 142163979-142163979 on Build GRCh38
rs981161903 Chr.3: 142164262-142164262 on Build GRCh38

View Full Product Details