Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 142458866-142459370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATR
Entrez Gene ID
545
Gene Name
ATR serine/threonine kinase
Gene Aliases
FCTCS, FRP1, MEC1, SCKL, SCKL1
UniGene
Hs.271791
Gene Chromosome Location
Chr.3: 142449235-142578826 on Build GRCh38
Transcripts
NM_001184.3 XM_011512924.1 XM_011512925.1 XM_017006643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
728982 c.7226G>C p.C2409S Chr.3: 142459350-142459350 on Build GRCh38
728981 c.7206A>T p.T2402T Chr.3: 142459370-142459370 on Build GRCh38
6935023 c.7340C>A p.P2447H Chr.3: 142459236-142459236 on Build GRCh38
212642 c.7220G>A p.R2407H Chr.3: 142459356-142459356 on Build GRCh38
6297404 c.7379C>A p.S2460Y Chr.3: 142459082-142459082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2229032 Chr.3: 142459302-142459302 on Build GRCh38
rs940571167 Chr.3: 142459019-142459019 on Build GRCh38
rs756975919 Chr.3: 142459099-142459100 on Build GRCh38
rs999756372 Chr.3: 142459241-142459241 on Build GRCh38
rs775964355 Chr.3: 142459054-142459054 on Build GRCh38
rs773858387 Chr.3: 142459283-142459283 on Build GRCh38
rs754253403 Chr.3: 142459085-142459085 on Build GRCh38
rs759327414 Chr.3: 142459284-142459284 on Build GRCh38
rs746160642 Chr.3: 142459204-142459204 on Build GRCh38
rs749493772 Chr.3: 142459036-142459036 on Build GRCh38
rs751010998 Chr.3: 142459114-142459114 on Build GRCh38
rs756065629 Chr.3: 142459297-142459297 on Build GRCh38
rs368580370 Chr.3: 142458922-142458922 on Build GRCh38
rs1011313917 Chr.3: 142459189-142459189 on Build GRCh38
rs758030592 Chr.3: 142458950-142458950 on Build GRCh38
rs947466947 Chr.3: 142459012-142459012 on Build GRCh38
rs751057775 Chr.3: 142458961-142458961 on Build GRCh38
rs369200535 Chr.3: 142458973-142458973 on Build GRCh38
rs368562492 Chr.3: 142459190-142459190 on Build GRCh38
rs777776233 Chr.3: 142459034-142459034 on Build GRCh38
rs762621866 Chr.3: 142459086-142459086 on Build GRCh38
rs144901974 Chr.3: 142459161-142459161 on Build GRCh38
rs768716333 Chr.3: 142459147-142459148 on Build GRCh38
rs767102663 Chr.3: 142459286-142459286 on Build GRCh38
rs574699024 Chr.3: 142458991-142458991 on Build GRCh38
rs563169447 Chr.3: 142458895-142458895 on Build GRCh38
rs1015400677 Chr.3: 142459230-142459230 on Build GRCh38
rs33972295 Chr.3: 142459276-142459276 on Build GRCh38
rs764502964 Chr.3: 142458947-142458947 on Build GRCh38
rs752303030 Chr.3: 142459292-142459292 on Build GRCh38
rs150290264 Chr.3: 142459160-142459160 on Build GRCh38
rs756158487 Chr.3: 142459143-142459143 on Build GRCh38
rs753611213 Chr.3: 142459316-142459316 on Build GRCh38
rs772135904 Chr.3: 142459210-142459210 on Build GRCh38
rs761042012 Chr.3: 142458942-142458942 on Build GRCh38
rs756990664 Chr.3: 142459323-142459323 on Build GRCh38
rs868667144 Chr.3: 142459141-142459141 on Build GRCh38
rs373093280 Chr.3: 142458974-142458974 on Build GRCh38
rs767931735 Chr.3: 142458926-142458926 on Build GRCh38
rs150286172 Chr.3: 142459050-142459050 on Build GRCh38
rs532315419 Chr.3: 142459224-142459224 on Build GRCh38
rs775588375 Chr.3: 142459211-142459211 on Build GRCh38
rs777688157 Chr.3: 142459146-142459146 on Build GRCh38
rs370855362 Chr.3: 142459129-142459129 on Build GRCh38
rs556313656 Chr.3: 142458990-142458990 on Build GRCh38
rs771106421 Chr.3: 142459045-142459045 on Build GRCh38
rs144887641 Chr.3: 142459062-142459062 on Build GRCh38
rs765983327 Chr.3: 142459109-142459109 on Build GRCh38
rs752674877 Chr.3: 142459140-142459140 on Build GRCh38
rs761240123 Chr.3: 142459058-142459058 on Build GRCh38
rs904051578 Chr.3: 142459229-142459229 on Build GRCh38
rs200556378 Chr.3: 142459225-142459225 on Build GRCh38
rs1029034437 Chr.3: 142459279-142459279 on Build GRCh38
rs909033162 Chr.3: 142459311-142459311 on Build GRCh38
rs369748890 Chr.3: 142458917-142458917 on Build GRCh38
rs754456724 Chr.3: 142459122-142459122 on Build GRCh38
rs757460513 Chr.3: 142459195-142459195 on Build GRCh38
rs1031690432 Chr.3: 142459263-142459263 on Build GRCh38
rs140201292 Chr.3: 142459030-142459030 on Build GRCh38
rs376735982 Chr.3: 142459035-142459035 on Build GRCh38
rs113377909 Chr.3: 142459258-142459258 on Build GRCh38
rs752993414 Chr.3: 142458934-142458934 on Build GRCh38
rs779016953 Chr.3: 142459203-142459203 on Build GRCh38
rs199551428 Chr.3: 142459223-142459223 on Build GRCh38
rs200700656 Chr.3: 142459049-142459049 on Build GRCh38
rs750073869 Chr.3: 142458949-142458949 on Build GRCh38
rs866711636 Chr.3: 142458897-142458897 on Build GRCh38
rs376727616 Chr.3: 142458953-142458953 on Build GRCh38
rs377119675 Chr.3: 142459313-142459313 on Build GRCh38
rs149045116 Chr.3: 142459273-142459273 on Build GRCh38

View Full Product Details