Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 143990091-143990601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf58
Entrez Gene ID
205428
Gene Name
chromosome 3 open reading frame 58
Gene Aliases
DIA1, GoPro49, HASF
UniGene
Hs.288954
Gene Chromosome Location
Chr.3: 143971798-143992368 on Build GRCh38
Transcripts
NM_001134470.1 NM_173552.3 XM_011512543.2 XM_017005869.1

SNP Detail

SNP ID Chromosome Location
rs1009836165 Chr.3: 143990430-143990430 on Build GRCh38
rs545143818 Chr.3: 143990428-143990428 on Build GRCh38
rs967417198 Chr.3: 143990151-143990151 on Build GRCh38
rs543997947 Chr.3: 143990265-143990265 on Build GRCh38
rs1029508494 Chr.3: 143990221-143990221 on Build GRCh38
rs891005364 Chr.3: 143990469-143990469 on Build GRCh38
rs891396009 Chr.3: 143990172-143990172 on Build GRCh38
rs899899759 Chr.3: 143990448-143990448 on Build GRCh38
rs903226468 Chr.3: 143990556-143990556 on Build GRCh38
rs933552092 Chr.3: 143990332-143990332 on Build GRCh38
rs865976233 Chr.3: 143990546-143990546 on Build GRCh38
rs747341563 Chr.3: 143990196-143990196 on Build GRCh38
rs781490506 Chr.3: 143990226-143990226 on Build GRCh38
rs1047311604 Chr.3: 143990159-143990159 on Build GRCh38
rs1012108109 Chr.3: 143990479-143990479 on Build GRCh38
rs17667256 Chr.3: 143990526-143990526 on Build GRCh38
rs992120709 Chr.3: 143990578-143990578 on Build GRCh38
rs7642533 Chr.3: 143990555-143990555 on Build GRCh38
rs572153742 Chr.3: 143990427-143990427 on Build GRCh38
rs927103801 Chr.3: 143990380-143990380 on Build GRCh38
rs557551663 Chr.3: 143990550-143990550 on Build GRCh38
rs959911628 Chr.3: 143990176-143990176 on Build GRCh38
rs757791852 Chr.3: 143990200-143990200 on Build GRCh38
rs998817471 Chr.3: 143990568-143990568 on Build GRCh38
rs951377503 Chr.3: 143990554-143990554 on Build GRCh38
rs576457993 Chr.3: 143990129-143990129 on Build GRCh38
rs1040299309 Chr.3: 143990421-143990421 on Build GRCh38
rs200201063 Chr.3: 143990294-143990295 on Build GRCh38
rs1010096200 Chr.3: 143990173-143990173 on Build GRCh38
rs995576682 Chr.3: 143990537-143990537 on Build GRCh38
rs891476340 Chr.3: 143990427-143990428 on Build GRCh38
rs191951426 Chr.3: 143990264-143990264 on Build GRCh38
rs1040694963 Chr.3: 143990306-143990306 on Build GRCh38
rs918570705 Chr.3: 143990297-143990297 on Build GRCh38
rs935735968 Chr.3: 143990385-143990385 on Build GRCh38
rs7642337 Chr.3: 143990360-143990360 on Build GRCh38
rs559598108 Chr.3: 143990399-143990399 on Build GRCh38
rs918140271 Chr.3: 143990310-143990310 on Build GRCh38
rs992423756 Chr.3: 143990304-143990304 on Build GRCh38
rs368566375 Chr.3: 143990426-143990427 on Build GRCh38
rs879563410 Chr.3: 143990246-143990246 on Build GRCh38
rs529568914 Chr.3: 143990161-143990161 on Build GRCh38
rs1018519040 Chr.3: 143990489-143990489 on Build GRCh38
rs769755991 Chr.3: 143990294-143990294 on Build GRCh38
rs575536190 Chr.3: 143990544-143990544 on Build GRCh38
rs1050606795 Chr.3: 143990480-143990480 on Build GRCh38
rs377662843 Chr.3: 143990367-143990367 on Build GRCh38
rs116580203 Chr.3: 143990283-143990283 on Build GRCh38
rs963108335 Chr.3: 143990289-143990289 on Build GRCh38
rs749487430 Chr.3: 143990528-143990528 on Build GRCh38
rs551084063 Chr.3: 143990434-143990434 on Build GRCh38
rs563398379 Chr.3: 143990524-143990524 on Build GRCh38
rs577772731 Chr.3: 143990326-143990326 on Build GRCh38
rs182271050 Chr.3: 143990300-143990300 on Build GRCh38
rs568774882 Chr.3: 143990135-143990135 on Build GRCh38
rs1050973787 Chr.3: 143990442-143990442 on Build GRCh38
rs1045606149 Chr.3: 143990338-143990338 on Build GRCh38
rs931081217 Chr.3: 143990281-143990281 on Build GRCh38

View Full Product Details