Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 143991266-143991808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf58
Entrez Gene ID
205428
Gene Name
chromosome 3 open reading frame 58
Gene Aliases
DIA1, GoPro49, HASF
UniGene
Hs.288954
Gene Chromosome Location
Chr.3: 143971798-143992368 on Build GRCh38
Transcripts
NM_001134470.1 NM_173552.3 XM_011512543.2 XM_017005869.1

SNP Detail

SNP ID Chromosome Location
rs879354070 Chr.3: 143991612-143991612 on Build GRCh38
rs1029492560 Chr.3: 143991591-143991591 on Build GRCh38
rs967126450 Chr.3: 143991337-143991337 on Build GRCh38
rs905244900 Chr.3: 143991453-143991453 on Build GRCh38
rs374347585 Chr.3: 143991753-143991753 on Build GRCh38
rs750860959 Chr.3: 143991336-143991336 on Build GRCh38
rs530651142 Chr.3: 143991322-143991322 on Build GRCh38
rs980605907 Chr.3: 143991696-143991696 on Build GRCh38
rs76073851 Chr.3: 143991316-143991316 on Build GRCh38
rs952329359 Chr.3: 143991608-143991608 on Build GRCh38
rs185739883 Chr.3: 143991646-143991646 on Build GRCh38
rs779049252 Chr.3: 143991581-143991581 on Build GRCh38
rs997995920 Chr.3: 143991338-143991338 on Build GRCh38
rs909944304 Chr.3: 143991697-143991697 on Build GRCh38
rs969019564 Chr.3: 143991400-143991400 on Build GRCh38
rs1044757299 Chr.3: 143991303-143991303 on Build GRCh38
rs114925678 Chr.3: 143991420-143991420 on Build GRCh38
rs1009513282 Chr.3: 143991710-143991710 on Build GRCh38
rs942362402 Chr.3: 143991363-143991363 on Build GRCh38
rs578148002 Chr.3: 143991299-143991299 on Build GRCh38
rs914887208 Chr.3: 143991399-143991399 on Build GRCh38
rs917340453 Chr.3: 143991519-143991519 on Build GRCh38
rs955062481 Chr.3: 143991557-143991557 on Build GRCh38
rs866598756 Chr.3: 143991652-143991652 on Build GRCh38
rs912288540 Chr.3: 143991355-143991355 on Build GRCh38
rs193041630 Chr.3: 143991642-143991642 on Build GRCh38
rs534890127 Chr.3: 143991655-143991655 on Build GRCh38
rs985174476 Chr.3: 143991379-143991379 on Build GRCh38
rs571523220 Chr.3: 143991549-143991549 on Build GRCh38
rs897035500 Chr.3: 143991589-143991589 on Build GRCh38
rs931215353 Chr.3: 143991387-143991387 on Build GRCh38
rs986967304 Chr.3: 143991342-143991342 on Build GRCh38
rs115250100 Chr.3: 143991750-143991750 on Build GRCh38
rs868520169 Chr.3: 143991465-143991465 on Build GRCh38
rs949251897 Chr.3: 143991639-143991639 on Build GRCh38
rs578206027 Chr.3: 143991780-143991780 on Build GRCh38
rs948798657 Chr.3: 143991529-143991529 on Build GRCh38
rs377382591 Chr.3: 143991321-143991321 on Build GRCh38
rs1044442993 Chr.3: 143991534-143991534 on Build GRCh38
rs998026972 Chr.3: 143991590-143991590 on Build GRCh38
rs187972259 Chr.3: 143991568-143991568 on Build GRCh38
rs917414058 Chr.3: 143991689-143991689 on Build GRCh38
rs948855893 Chr.3: 143991693-143991693 on Build GRCh38
rs766967790 Chr.3: 143991300-143991314 on Build GRCh38
rs1042619981 Chr.3: 143991659-143991659 on Build GRCh38
rs1029420667 Chr.3: 143991353-143991353 on Build GRCh38
rs888427807 Chr.3: 143991538-143991538 on Build GRCh38
rs941360999 Chr.3: 143991726-143991726 on Build GRCh38
rs756233404 Chr.3: 143991351-143991351 on Build GRCh38
rs116183819 Chr.3: 143991494-143991494 on Build GRCh38
rs569924970 Chr.3: 143991592-143991592 on Build GRCh38
rs748043118 Chr.3: 143991596-143991596 on Build GRCh38
rs1050938468 Chr.3: 143991782-143991782 on Build GRCh38
rs953739910 Chr.3: 143991374-143991374 on Build GRCh38
rs780128425 Chr.3: 143991481-143991481 on Build GRCh38
rs908388003 Chr.3: 143991402-143991402 on Build GRCh38
rs551768623 Chr.3: 143991573-143991573 on Build GRCh38
rs768104090 Chr.3: 143991648-143991648 on Build GRCh38
rs975071889 Chr.3: 143991366-143991366 on Build GRCh38
rs1046961370 Chr.3: 143991395-143991395 on Build GRCh38
rs888405950 Chr.3: 143991314-143991314 on Build GRCh38
rs961212150 Chr.3: 143991333-143991333 on Build GRCh38
rs773898455 Chr.3: 143991728-143991728 on Build GRCh38
rs563100573 Chr.3: 143991318-143991318 on Build GRCh38
rs1979910 Chr.3: 143991426-143991426 on Build GRCh38
rs773251611 Chr.3: 143991631-143991631 on Build GRCh38
rs563570831 Chr.3: 143991611-143991611 on Build GRCh38
rs528434025 Chr.3: 143991640-143991640 on Build GRCh38
rs549127622 Chr.3: 143991377-143991377 on Build GRCh38

View Full Product Details