Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 148878306-148878841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA3
Entrez Gene ID
1359
Gene Name
carboxypeptidase A3
Gene Aliases
MC-CPA
UniGene
Hs.646
Gene Chromosome Location
Chr.3: 148865256-148897091 on Build GRCh38
Transcripts
NM_001870.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1308700 c.313G>C p.D105H Chr.3: 148878484-148878484 on Build GRCh38
127706 c.420C>T p.V140V Chr.3: 148878694-148878694 on Build GRCh38
4449579 c.270-1G>A p.? Chr.3: 148878440-148878440 on Build GRCh38
1308701 c.463T>C p.Y155H Chr.3: 148878737-148878737 on Build GRCh38
1419812 c.273C>A p.I91I Chr.3: 148878444-148878444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755072183 Chr.3: 148878416-148878416 on Build GRCh38
rs373995016 Chr.3: 148878623-148878623 on Build GRCh38
rs774374687 Chr.3: 148878633-148878633 on Build GRCh38
rs182570216 Chr.3: 148878563-148878563 on Build GRCh38
rs775837205 Chr.3: 148878651-148878651 on Build GRCh38
rs751872495 Chr.3: 148878774-148878774 on Build GRCh38
rs748436457 Chr.3: 148878679-148878679 on Build GRCh38
rs370991073 Chr.3: 148878534-148878534 on Build GRCh38
rs769345179 Chr.3: 148878547-148878547 on Build GRCh38
rs770430908 Chr.3: 148878717-148878717 on Build GRCh38
rs766795653 Chr.3: 148878696-148878696 on Build GRCh38
rs753245625 Chr.3: 148878713-148878713 on Build GRCh38
rs368273938 Chr.3: 148878581-148878581 on Build GRCh38
rs75229139 Chr.3: 148878657-148878657 on Build GRCh38
rs754441144 Chr.3: 148878520-148878520 on Build GRCh38
rs761338426 Chr.3: 148878452-148878452 on Build GRCh38
rs746262682 Chr.3: 148878531-148878531 on Build GRCh38
rs372036286 Chr.3: 148878582-148878582 on Build GRCh38
rs770652989 Chr.3: 148878691-148878691 on Build GRCh38
rs376999632 Chr.3: 148878505-148878505 on Build GRCh38
rs891390161 Chr.3: 148878490-148878490 on Build GRCh38
rs539944476 Chr.3: 148878653-148878653 on Build GRCh38
rs764428082 Chr.3: 148878656-148878656 on Build GRCh38
rs765799527 Chr.3: 148878615-148878615 on Build GRCh38
rs747626973 Chr.3: 148878545-148878545 on Build GRCh38
rs775116134 Chr.3: 148878574-148878575 on Build GRCh38
rs566975700 Chr.3: 148878333-148878333 on Build GRCh38
rs146235151 Chr.3: 148878675-148878675 on Build GRCh38
rs773590504 Chr.3: 148878763-148878763 on Build GRCh38
rs778445496 Chr.3: 148878426-148878426 on Build GRCh38
rs761780745 Chr.3: 148878758-148878758 on Build GRCh38
rs371556836 Chr.3: 148878337-148878337 on Build GRCh38
rs763383662 Chr.3: 148878764-148878764 on Build GRCh38
rs779731268 Chr.3: 148878443-148878443 on Build GRCh38
rs187835267 Chr.3: 148878625-148878625 on Build GRCh38
rs529614942 Chr.3: 148878626-148878626 on Build GRCh38
rs781635243 Chr.3: 148878619-148878619 on Build GRCh38
rs776955071 Chr.3: 148878749-148878749 on Build GRCh38
rs775892136 Chr.3: 148878590-148878590 on Build GRCh38
rs777424403 Chr.3: 148878712-148878712 on Build GRCh38
rs202121687 Chr.3: 148878396-148878396 on Build GRCh38
rs75696101 Chr.3: 148878566-148878566 on Build GRCh38
rs775949190 Chr.3: 148878451-148878451 on Build GRCh38
rs990832586 Chr.3: 148878668-148878668 on Build GRCh38
rs752773216 Chr.3: 148878493-148878493 on Build GRCh38
rs769681649 Chr.3: 148878455-148878455 on Build GRCh38
rs374102630 Chr.3: 148878425-148878425 on Build GRCh38
rs376921076 Chr.3: 148878583-148878583 on Build GRCh38
rs575542115 Chr.3: 148878609-148878609 on Build GRCh38
rs372213795 Chr.3: 148878731-148878731 on Build GRCh38
rs757650636 Chr.3: 148878614-148878614 on Build GRCh38
rs750582936 Chr.3: 148878470-148878470 on Build GRCh38
rs367753806 Chr.3: 148878721-148878721 on Build GRCh38
rs745618307 Chr.3: 148878635-148878635 on Build GRCh38
rs368209657 Chr.3: 148878666-148878666 on Build GRCh38
rs751779500 Chr.3: 148878415-148878415 on Build GRCh38
rs762715744 Chr.3: 148878458-148878458 on Build GRCh38
rs61733456 Chr.3: 148878739-148878739 on Build GRCh38
rs61733455 Chr.3: 148878530-148878530 on Build GRCh38
rs61504164 Chr.3: 148878358-148878359 on Build GRCh38
rs192753847 Chr.3: 148878650-148878650 on Build GRCh38
rs201079313 Chr.3: 148878413-148878413 on Build GRCh38
rs755500166 Chr.3: 148878617-148878617 on Build GRCh38
rs76952562 Chr.3: 148878780-148878780 on Build GRCh38
rs1020527945 Chr.3: 148878796-148878796 on Build GRCh38
rs1020117522 Chr.3: 148878559-148878559 on Build GRCh38
rs780857203 Chr.3: 148878537-148878537 on Build GRCh38
rs755705481 Chr.3: 148878709-148878709 on Build GRCh38
rs754388058 Chr.3: 148878613-148878613 on Build GRCh38
rs555034185 Chr.3: 148878427-148878427 on Build GRCh38
rs768364468 Chr.3: 148878450-148878450 on Build GRCh38
rs111881854 Chr.3: 148878719-148878719 on Build GRCh38
rs763703343 Chr.3: 148878785-148878785 on Build GRCh38
rs746826544 Chr.3: 148878736-148878736 on Build GRCh38
rs748842657 Chr.3: 148878627-148878627 on Build GRCh38
rs766145394 Chr.3: 148878465-148878465 on Build GRCh38
rs949502733 Chr.3: 148878336-148878336 on Build GRCh38
rs767793731 Chr.3: 148878486-148878486 on Build GRCh38
rs746693413 Chr.3: 148878444-148878444 on Build GRCh38
rs369546683 Chr.3: 148878766-148878766 on Build GRCh38
rs768964496 Chr.3: 148878738-148878738 on Build GRCh38
rs762770539 Chr.3: 148878412-148878412 on Build GRCh38
rs866956006 Chr.3: 148878454-148878454 on Build GRCh38
rs114722178 Chr.3: 148878368-148878368 on Build GRCh38
rs750988849 Chr.3: 148878682-148878682 on Build GRCh38
rs753574446 Chr.3: 148878794-148878794 on Build GRCh38
rs769976312 Chr.3: 148878761-148878761 on Build GRCh38
rs977803874 Chr.3: 148878334-148878334 on Build GRCh38
rs200383775 Chr.3: 148878718-148878718 on Build GRCh38
rs770724109 Chr.3: 148878629-148878629 on Build GRCh38
rs138751736 Chr.3: 148878519-148878519 on Build GRCh38
rs377284175 Chr.3: 148878733-148878733 on Build GRCh38
rs544079869 Chr.3: 148878618-148878618 on Build GRCh38
rs781195825 Chr.3: 148878419-148878419 on Build GRCh38
rs553566790 Chr.3: 148878561-148878561 on Build GRCh38
rs576567278 Chr.3: 148878558-148878558 on Build GRCh38
rs745587427 Chr.3: 148878403-148878403 on Build GRCh38
rs1050032879 Chr.3: 148878480-148878480 on Build GRCh38
rs759608831 Chr.3: 148878481-148878481 on Build GRCh38
rs559987225 Chr.3: 148878403-148878403 on Build GRCh38
rs771236993 Chr.3: 148878439-148878439 on Build GRCh38
rs757624575 Chr.3: 148878528-148878528 on Build GRCh38
rs750715856 Chr.3: 148878616-148878616 on Build GRCh38
rs558461338 Chr.3: 148878591-148878591 on Build GRCh38
rs374800491 Chr.3: 148878699-148878699 on Build GRCh38
rs771611187 Chr.3: 148878565-148878566 on Build GRCh38

View Full Product Details