Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 148878506-148879010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA3
Entrez Gene ID
1359
Gene Name
carboxypeptidase A3
Gene Aliases
MC-CPA
UniGene
Hs.646
Gene Chromosome Location
Chr.3: 148865256-148897091 on Build GRCh38
Transcripts
NM_001870.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6096380 c.441T>C p.S147S Chr.3: 148878715-148878715 on Build GRCh38
7226291 c.372+4C>T p.? Chr.3: 148878547-148878547 on Build GRCh38
127706 c.420C>T p.V140V Chr.3: 148878694-148878694 on Build GRCh38
6163344 c.340C>T p.H114Y Chr.3: 148878511-148878511 on Build GRCh38
1308701 c.463T>C p.Y155H Chr.3: 148878737-148878737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531704427 Chr.3: 148878957-148878957 on Build GRCh38
rs76952562 Chr.3: 148878780-148878780 on Build GRCh38
rs1020527945 Chr.3: 148878796-148878796 on Build GRCh38
rs373995016 Chr.3: 148878623-148878623 on Build GRCh38
rs1020117522 Chr.3: 148878559-148878559 on Build GRCh38
rs763806984 Chr.3: 148878908-148878908 on Build GRCh38
rs774374687 Chr.3: 148878633-148878633 on Build GRCh38
rs182570216 Chr.3: 148878563-148878563 on Build GRCh38
rs775837205 Chr.3: 148878651-148878651 on Build GRCh38
rs748436457 Chr.3: 148878679-148878679 on Build GRCh38
rs751872495 Chr.3: 148878774-148878774 on Build GRCh38
rs1034298563 Chr.3: 148878870-148878870 on Build GRCh38
rs370991073 Chr.3: 148878534-148878534 on Build GRCh38
rs770430908 Chr.3: 148878717-148878717 on Build GRCh38
rs769345179 Chr.3: 148878547-148878547 on Build GRCh38
rs766795653 Chr.3: 148878696-148878696 on Build GRCh38
rs753245625 Chr.3: 148878713-148878713 on Build GRCh38
rs368273938 Chr.3: 148878581-148878581 on Build GRCh38
rs75229139 Chr.3: 148878657-148878657 on Build GRCh38
rs970795213 Chr.3: 148878906-148878906 on Build GRCh38
rs780857203 Chr.3: 148878537-148878537 on Build GRCh38
rs755705481 Chr.3: 148878709-148878709 on Build GRCh38
rs989251299 Chr.3: 148878823-148878823 on Build GRCh38
rs754388058 Chr.3: 148878613-148878613 on Build GRCh38
rs746262682 Chr.3: 148878531-148878531 on Build GRCh38
rs372036286 Chr.3: 148878582-148878582 on Build GRCh38
rs770652989 Chr.3: 148878691-148878691 on Build GRCh38
rs111881854 Chr.3: 148878719-148878719 on Build GRCh38
rs763703343 Chr.3: 148878785-148878785 on Build GRCh38
rs539944476 Chr.3: 148878653-148878653 on Build GRCh38
rs764428082 Chr.3: 148878656-148878656 on Build GRCh38
rs746826544 Chr.3: 148878736-148878736 on Build GRCh38
rs747626973 Chr.3: 148878545-148878545 on Build GRCh38
rs765799527 Chr.3: 148878615-148878615 on Build GRCh38
rs748842657 Chr.3: 148878627-148878627 on Build GRCh38
rs775116134 Chr.3: 148878574-148878575 on Build GRCh38
rs113737792 Chr.3: 148878817-148878817 on Build GRCh38
rs146235151 Chr.3: 148878675-148878675 on Build GRCh38
rs1000502447 Chr.3: 148878849-148878849 on Build GRCh38
rs773590504 Chr.3: 148878763-148878763 on Build GRCh38
rs908019813 Chr.3: 148878931-148878931 on Build GRCh38
rs759781055 Chr.3: 148878828-148878828 on Build GRCh38
rs768964496 Chr.3: 148878738-148878738 on Build GRCh38
rs369546683 Chr.3: 148878766-148878766 on Build GRCh38
rs761780745 Chr.3: 148878758-148878758 on Build GRCh38
rs548864291 Chr.3: 148878901-148878901 on Build GRCh38
rs763383662 Chr.3: 148878764-148878764 on Build GRCh38
rs750988849 Chr.3: 148878682-148878682 on Build GRCh38
rs753574446 Chr.3: 148878794-148878794 on Build GRCh38
rs769976312 Chr.3: 148878761-148878761 on Build GRCh38
rs200383775 Chr.3: 148878718-148878718 on Build GRCh38
rs770724109 Chr.3: 148878629-148878629 on Build GRCh38
rs187835267 Chr.3: 148878625-148878625 on Build GRCh38
rs529614942 Chr.3: 148878626-148878626 on Build GRCh38
rs781635243 Chr.3: 148878619-148878619 on Build GRCh38
rs776955071 Chr.3: 148878749-148878749 on Build GRCh38
rs914730914 Chr.3: 148878847-148878847 on Build GRCh38
rs777424403 Chr.3: 148878712-148878712 on Build GRCh38
rs775892136 Chr.3: 148878590-148878590 on Build GRCh38
rs377284175 Chr.3: 148878733-148878733 on Build GRCh38
rs544079869 Chr.3: 148878618-148878618 on Build GRCh38
rs75696101 Chr.3: 148878566-148878566 on Build GRCh38
rs973990700 Chr.3: 148878983-148878983 on Build GRCh38
rs553566790 Chr.3: 148878561-148878561 on Build GRCh38
rs576567278 Chr.3: 148878558-148878558 on Build GRCh38
rs990832586 Chr.3: 148878668-148878668 on Build GRCh38
rs532080433 Chr.3: 148878894-148878894 on Build GRCh38
rs2331448 Chr.3: 148878844-148878844 on Build GRCh38
rs376921076 Chr.3: 148878583-148878583 on Build GRCh38
rs369341487 Chr.3: 148878885-148878885 on Build GRCh38
rs575542115 Chr.3: 148878609-148878609 on Build GRCh38
rs990780200 Chr.3: 148878891-148878891 on Build GRCh38
rs372213795 Chr.3: 148878731-148878731 on Build GRCh38
rs757650636 Chr.3: 148878614-148878614 on Build GRCh38
rs367753806 Chr.3: 148878721-148878721 on Build GRCh38
rs745618307 Chr.3: 148878635-148878635 on Build GRCh38
rs368209657 Chr.3: 148878666-148878666 on Build GRCh38
rs757624575 Chr.3: 148878528-148878528 on Build GRCh38
rs750715856 Chr.3: 148878616-148878616 on Build GRCh38
rs61733456 Chr.3: 148878739-148878739 on Build GRCh38
rs374800491 Chr.3: 148878699-148878699 on Build GRCh38
rs558461338 Chr.3: 148878591-148878591 on Build GRCh38
rs771611187 Chr.3: 148878565-148878566 on Build GRCh38
rs61733455 Chr.3: 148878530-148878530 on Build GRCh38
rs904394628 Chr.3: 148878819-148878819 on Build GRCh38
rs192753847 Chr.3: 148878650-148878650 on Build GRCh38
rs568485250 Chr.3: 148878902-148878902 on Build GRCh38
rs755500166 Chr.3: 148878617-148878617 on Build GRCh38

View Full Product Details