Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 148882418-148882916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA3
Entrez Gene ID
1359
Gene Name
carboxypeptidase A3
Gene Aliases
MC-CPA
UniGene
Hs.646
Gene Chromosome Location
Chr.3: 148865256-148897091 on Build GRCh38
Transcripts
NM_001870.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6511640 c.735C>A p.C245* Chr.3: 148882552-148882552 on Build GRCh38
1693569 c.709C>T p.R237C Chr.3: 148882526-148882526 on Build GRCh38
6240069 c.688-4G>T p.? Chr.3: 148882501-148882501 on Build GRCh38
1039674 c.761T>G p.F254C Chr.3: 148882578-148882578 on Build GRCh38
6346786 c.696G>T p.M232I Chr.3: 148882513-148882513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765086873 Chr.3: 148882591-148882591 on Build GRCh38
rs373378775 Chr.3: 148882487-148882487 on Build GRCh38
rs778693111 Chr.3: 148882555-148882555 on Build GRCh38
rs773067485 Chr.3: 148882598-148882598 on Build GRCh38
rs1010815991 Chr.3: 148882797-148882797 on Build GRCh38
rs1037721761 Chr.3: 148882855-148882855 on Build GRCh38
rs968452281 Chr.3: 148882768-148882768 on Build GRCh38
rs769211015 Chr.3: 148882462-148882462 on Build GRCh38
rs773123811 Chr.3: 148882460-148882460 on Build GRCh38
rs751674825 Chr.3: 148882532-148882532 on Build GRCh38
rs1001496080 Chr.3: 148882809-148882809 on Build GRCh38
rs753387651 Chr.3: 148882658-148882658 on Build GRCh38
rs774801756 Chr.3: 148882567-148882567 on Build GRCh38
rs952912939 Chr.3: 148882886-148882886 on Build GRCh38
rs768538060 Chr.3: 148882584-148882584 on Build GRCh38
rs918783005 Chr.3: 148882614-148882614 on Build GRCh38
rs781772953 Chr.3: 148882538-148882538 on Build GRCh38
rs545467403 Chr.3: 148882613-148882613 on Build GRCh38
rs1009873812 Chr.3: 148882725-148882725 on Build GRCh38
rs370838199 Chr.3: 148882580-148882580 on Build GRCh38
rs1001790285 Chr.3: 148882689-148882689 on Build GRCh38
rs35603555 Chr.3: 148882620-148882621 on Build GRCh38
rs35324938 Chr.3: 148882591-148882592 on Build GRCh38
rs775602948 Chr.3: 148882488-148882488 on Build GRCh38
rs774186299 Chr.3: 148882499-148882499 on Build GRCh38
rs755494551 Chr.3: 148882537-148882537 on Build GRCh38
rs922338136 Chr.3: 148882757-148882757 on Build GRCh38
rs185926770 Chr.3: 148882616-148882616 on Build GRCh38
rs942107667 Chr.3: 148882851-148882851 on Build GRCh38
rs374150898 Chr.3: 148882527-148882527 on Build GRCh38
rs75723270 Chr.3: 148882775-148882775 on Build GRCh38
rs905724569 Chr.3: 148882620-148882620 on Build GRCh38
rs116686228 Chr.3: 148882511-148882511 on Build GRCh38
rs747539518 Chr.3: 148882460-148882460 on Build GRCh38
rs999344158 Chr.3: 148882452-148882452 on Build GRCh38
rs1036106367 Chr.3: 148882831-148882831 on Build GRCh38
rs1025790689 Chr.3: 148882871-148882871 on Build GRCh38
rs745573726 Chr.3: 148882556-148882556 on Build GRCh38
rs764564236 Chr.3: 148882634-148882634 on Build GRCh38
rs905128048 Chr.3: 148882808-148882808 on Build GRCh38
rs1052267592 Chr.3: 148882786-148882786 on Build GRCh38
rs765426086 Chr.3: 148882517-148882517 on Build GRCh38
rs34561950 Chr.3: 148882773-148882773 on Build GRCh38
rs113080458 Chr.3: 148882685-148882685 on Build GRCh38
rs370488172 Chr.3: 148882481-148882481 on Build GRCh38
rs759848761 Chr.3: 148882518-148882519 on Build GRCh38
rs1057026384 Chr.3: 148882716-148882716 on Build GRCh38
rs1044973417 Chr.3: 148882807-148882807 on Build GRCh38
rs910877396 Chr.3: 148882686-148882686 on Build GRCh38
rs766647006 Chr.3: 148882612-148882612 on Build GRCh38
rs907905350 Chr.3: 148882848-148882848 on Build GRCh38
rs199600994 Chr.3: 148882526-148882526 on Build GRCh38
rs935084753 Chr.3: 148882760-148882760 on Build GRCh38
rs767600438 Chr.3: 148882524-148882525 on Build GRCh38
rs979586698 Chr.3: 148882500-148882500 on Build GRCh38
rs756542652 Chr.3: 148882645-148882645 on Build GRCh38
rs955136367 Chr.3: 148882822-148882822 on Build GRCh38
rs199944830 Chr.3: 148882525-148882525 on Build GRCh38
rs1041128110 Chr.3: 148882724-148882724 on Build GRCh38
rs763028364 Chr.3: 148882606-148882606 on Build GRCh38
rs76223378 Chr.3: 148882773-148882773 on Build GRCh38
rs753312147 Chr.3: 148882631-148882631 on Build GRCh38
rs761624530 Chr.3: 148882512-148882512 on Build GRCh38
rs150822877 Chr.3: 148882589-148882589 on Build GRCh38
rs752921459 Chr.3: 148882546-148882547 on Build GRCh38
rs770330448 Chr.3: 148882553-148882553 on Build GRCh38
rs984358752 Chr.3: 148882660-148882660 on Build GRCh38
rs768114633 Chr.3: 148882630-148882630 on Build GRCh38
rs968465448 Chr.3: 148882489-148882489 on Build GRCh38
rs147449316 Chr.3: 148882857-148882857 on Build GRCh38
rs757614589 Chr.3: 148882870-148882870 on Build GRCh38
rs141428447 Chr.3: 148882719-148882719 on Build GRCh38
rs1019994563 Chr.3: 148882726-148882726 on Build GRCh38
rs748494421 Chr.3: 148882546-148882546 on Build GRCh38
rs760251410 Chr.3: 148882571-148882571 on Build GRCh38
rs1032908357 Chr.3: 148882779-148882779 on Build GRCh38
rs370747287 Chr.3: 148882508-148882508 on Build GRCh38
rs141391874 Chr.3: 148882564-148882564 on Build GRCh38
rs74413758 Chr.3: 148882790-148882790 on Build GRCh38
rs765856914 Chr.3: 148882509-148882509 on Build GRCh38
rs975049062 Chr.3: 148882524-148882524 on Build GRCh38
rs550240191 Chr.3: 148882875-148882875 on Build GRCh38
rs528603640 Chr.3: 148882588-148882588 on Build GRCh38
rs563615985 Chr.3: 148882815-148882815 on Build GRCh38
rs999968490 Chr.3: 148882776-148882776 on Build GRCh38
rs376534861 Chr.3: 148882501-148882501 on Build GRCh38

View Full Product Details