Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 154275581-154276095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX36
Entrez Gene ID
170506
Gene Name
DEAH-box helicase 36
Gene Aliases
DDX36, G4R1, MLEL1, RHAU
UniGene
Hs.446270
Gene Chromosome Location
Chr.3: 154275668-154324497 on Build GRCh38
Transcripts
NM_001114397.1 NM_020865.2

SNP Detail

SNP ID Chromosome Location
rs746689850 Chr.3: 154275659-154275659 on Build GRCh38
rs922793118 Chr.3: 154275740-154275740 on Build GRCh38
rs932078713 Chr.3: 154276053-154276053 on Build GRCh38
rs1034652825 Chr.3: 154275699-154275699 on Build GRCh38
rs763460864 Chr.3: 154275797-154275797 on Build GRCh38
rs529462021 Chr.3: 154275753-154275753 on Build GRCh38
rs969744491 Chr.3: 154275681-154275681 on Build GRCh38
rs544360633 Chr.3: 154275874-154275874 on Build GRCh38
rs1001179189 Chr.3: 154275983-154275983 on Build GRCh38
rs144399858 Chr.3: 154275700-154275700 on Build GRCh38
rs921994961 Chr.3: 154276044-154276044 on Build GRCh38
rs931942996 Chr.3: 154275803-154275803 on Build GRCh38
rs1010747646 Chr.3: 154275978-154275978 on Build GRCh38
rs887999444 Chr.3: 154275849-154275849 on Build GRCh38
rs114735328 Chr.3: 154275910-154275910 on Build GRCh38
rs759175630 Chr.3: 154276026-154276026 on Build GRCh38
rs908014009 Chr.3: 154275893-154275893 on Build GRCh38
rs112801501 Chr.3: 154275732-154275732 on Build GRCh38
rs988633616 Chr.3: 154275734-154275734 on Build GRCh38
rs71790534 Chr.3: 154275979-154275980 on Build GRCh38
rs72996619 Chr.3: 154275731-154275731 on Build GRCh38
rs142215991 Chr.3: 154275678-154275678 on Build GRCh38
rs1010761743 Chr.3: 154275647-154275647 on Build GRCh38
rs1015518697 Chr.3: 154275644-154275644 on Build GRCh38
rs868084327 Chr.3: 154275981-154275981 on Build GRCh38
rs983649758 Chr.3: 154275783-154275783 on Build GRCh38
rs979773101 Chr.3: 154275692-154275692 on Build GRCh38
rs1032691699 Chr.3: 154275998-154275998 on Build GRCh38
rs113647894 Chr.3: 154276000-154276000 on Build GRCh38
rs944793060 Chr.3: 154275760-154275760 on Build GRCh38
rs570340635 Chr.3: 154275976-154275976 on Build GRCh38
rs944775695 Chr.3: 154275897-154275897 on Build GRCh38
rs112560810 Chr.3: 154276033-154276033 on Build GRCh38
rs568119530 Chr.3: 154276002-154276002 on Build GRCh38
rs147857747 Chr.3: 154275967-154275967 on Build GRCh38
rs1049067520 Chr.3: 154275845-154275845 on Build GRCh38
rs1003226195 Chr.3: 154275685-154275685 on Build GRCh38
rs1004981598 Chr.3: 154275632-154275632 on Build GRCh38
rs1014444635 Chr.3: 154275662-154275662 on Build GRCh38
rs1003256312 Chr.3: 154276000-154276001 on Build GRCh38
rs77982843 Chr.3: 154275947-154275947 on Build GRCh38
rs1042222934 Chr.3: 154275980-154275980 on Build GRCh38
rs932040089 Chr.3: 154275948-154275948 on Build GRCh38
rs879087769 Chr.3: 154276068-154276068 on Build GRCh38
rs966233871 Chr.3: 154276061-154276061 on Build GRCh38
rs535647169 Chr.3: 154276010-154276010 on Build GRCh38
rs1027721593 Chr.3: 154276024-154276024 on Build GRCh38
rs573966628 Chr.3: 154276039-154276039 on Build GRCh38
rs1040899900 Chr.3: 154275762-154275762 on Build GRCh38
rs759294351 Chr.3: 154275979-154275982 on Build GRCh38
rs896685974 Chr.3: 154275929-154275929 on Build GRCh38
rs1005573798 Chr.3: 154275873-154275873 on Build GRCh38
rs35550732 Chr.3: 154276000-154276001 on Build GRCh38
rs762720222 Chr.3: 154275981-154275982 on Build GRCh38
rs79342407 Chr.3: 154275977-154275977 on Build GRCh38

View Full Product Details