Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 154276635-154277162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX36
Entrez Gene ID
170506
Gene Name
DEAH-box helicase 36
Gene Aliases
DDX36, G4R1, MLEL1, RHAU
UniGene
Hs.446270
Gene Chromosome Location
Chr.3: 154275668-154324497 on Build GRCh38
Transcripts
NM_001114397.1 NM_020865.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7227792 c.2799G>T p.W933C Chr.3: 154276789-154276789 on Build GRCh38
3365050 c.2657A>G p.Y886C Chr.3: 154276889-154276889 on Build GRCh38
4677406 c.2697delT p.F899fs*18 Chr.3: 154276849-154276849 on Build GRCh38
387380 c.2754C>A p.S918S Chr.3: 154276834-154276834 on Build GRCh38
4677404 c.2727C>T p.N909N Chr.3: 154276819-154276819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758737721 Chr.3: 154276792-154276792 on Build GRCh38
rs892897184 Chr.3: 154276692-154276692 on Build GRCh38
rs889373577 Chr.3: 154277085-154277085 on Build GRCh38
rs189113801 Chr.3: 154277099-154277099 on Build GRCh38
rs750565904 Chr.3: 154277031-154277031 on Build GRCh38
rs967450501 Chr.3: 154277109-154277113 on Build GRCh38
rs757479472 Chr.3: 154276860-154276860 on Build GRCh38
rs967419464 Chr.3: 154276902-154276902 on Build GRCh38
rs113028874 Chr.3: 154276900-154276900 on Build GRCh38
rs1016722808 Chr.3: 154276861-154276861 on Build GRCh38
rs998872933 Chr.3: 154277112-154277112 on Build GRCh38
rs142222613 Chr.3: 154276758-154276758 on Build GRCh38
rs1041756284 Chr.3: 154277049-154277049 on Build GRCh38
rs759342254 Chr.3: 154276858-154276858 on Build GRCh38
rs376578943 Chr.3: 154277016-154277017 on Build GRCh38
rs752614061 Chr.3: 154276722-154276722 on Build GRCh38
rs780306001 Chr.3: 154276933-154276933 on Build GRCh38
rs762149375 Chr.3: 154276702-154276702 on Build GRCh38
rs754013959 Chr.3: 154276873-154276873 on Build GRCh38
rs199632295 Chr.3: 154276690-154276690 on Build GRCh38
rs573838822 Chr.3: 154276962-154276962 on Build GRCh38
rs977421480 Chr.3: 154276903-154276903 on Build GRCh38
rs533945423 Chr.3: 154276954-154276954 on Build GRCh38
rs756907366 Chr.3: 154276757-154276757 on Build GRCh38
rs992446400 Chr.3: 154276952-154276952 on Build GRCh38
rs374164936 Chr.3: 154276798-154276798 on Build GRCh38
rs755136663 Chr.3: 154276937-154276937 on Build GRCh38
rs756170390 Chr.3: 154277038-154277038 on Build GRCh38
rs73875340 Chr.3: 154276661-154276661 on Build GRCh38
rs151188512 Chr.3: 154276818-154276818 on Build GRCh38
rs386667254 Chr.3: 154276713-154276714 on Build GRCh38
rs763527830 Chr.3: 154276885-154276885 on Build GRCh38
rs781520161 Chr.3: 154277002-154277002 on Build GRCh38
rs777290837 Chr.3: 154276712-154276713 on Build GRCh38
rs998150992 Chr.3: 154276817-154276817 on Build GRCh38
rs764846473 Chr.3: 154276917-154276917 on Build GRCh38
rs902108724 Chr.3: 154276812-154276812 on Build GRCh38
rs758751767 Chr.3: 154276932-154276932 on Build GRCh38
rs138447229 Chr.3: 154276831-154276831 on Build GRCh38
rs748759274 Chr.3: 154276714-154276715 on Build GRCh38
rs745719588 Chr.3: 154276934-154276934 on Build GRCh38
rs1010291911 Chr.3: 154276694-154276694 on Build GRCh38
rs560623270 Chr.3: 154277120-154277120 on Build GRCh38
rs947920534 Chr.3: 154276975-154276975 on Build GRCh38
rs355787 Chr.3: 154276713-154276713 on Build GRCh38
rs997410014 Chr.3: 154276742-154276742 on Build GRCh38
rs578119150 Chr.3: 154277065-154277065 on Build GRCh38
rs550827541 Chr.3: 154276741-154276741 on Build GRCh38
rs752048217 Chr.3: 154276936-154276936 on Build GRCh38
rs940391219 Chr.3: 154277013-154277013 on Build GRCh38
rs781438310 Chr.3: 154276811-154276811 on Build GRCh38
rs917954517 Chr.3: 154276926-154276926 on Build GRCh38
rs760190079 Chr.3: 154276739-154276739 on Build GRCh38
rs377323319 Chr.3: 154276819-154276819 on Build GRCh38
rs947836764 Chr.3: 154276678-154276678 on Build GRCh38
rs750771753 Chr.3: 154276790-154276790 on Build GRCh38
rs937073650 Chr.3: 154276659-154276659 on Build GRCh38
rs894949618 Chr.3: 154276667-154276667 on Build GRCh38
rs753427207 Chr.3: 154276756-154276756 on Build GRCh38
rs201901768 Chr.3: 154276915-154276915 on Build GRCh38
rs866726380 Chr.3: 154276830-154276830 on Build GRCh38
rs555340044 Chr.3: 154276955-154276955 on Build GRCh38
rs537936210 Chr.3: 154276998-154276998 on Build GRCh38
rs1012184888 Chr.3: 154277089-154277089 on Build GRCh38
rs113420308 Chr.3: 154276901-154276901 on Build GRCh38
rs768734415 Chr.3: 154276801-154276801 on Build GRCh38
rs997783876 Chr.3: 154277056-154277056 on Build GRCh38
rs776952795 Chr.3: 154276696-154276696 on Build GRCh38
rs199866859 Chr.3: 154276896-154276896 on Build GRCh38
rs184648358 Chr.3: 154277051-154277051 on Build GRCh38
rs750085532 Chr.3: 154276928-154276928 on Build GRCh38
rs9289943 Chr.3: 154276714-154276714 on Build GRCh38
rs945842801 Chr.3: 154277038-154277038 on Build GRCh38
rs866299469 Chr.3: 154276989-154276989 on Build GRCh38
rs371212695 Chr.3: 154276717-154276717 on Build GRCh38
rs200951715 Chr.3: 154276713-154276714 on Build GRCh38
rs759318435 Chr.3: 154276715-154276715 on Build GRCh38
rs771792767 Chr.3: 154276867-154276867 on Build GRCh38
rs748228490 Chr.3: 154276951-154276951 on Build GRCh38
rs753346374 Chr.3: 154276987-154276990 on Build GRCh38
rs781782625 Chr.3: 154276941-154276941 on Build GRCh38
rs760538965 Chr.3: 154276875-154276875 on Build GRCh38
rs914276002 Chr.3: 154277019-154277019 on Build GRCh38

View Full Product Details